Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm01195479_m1
          See other GTF2IRD1 GE Assays ›
          Gene Symbol:
          Gtf2ird1
          Gene Name:
          general transcription factor II I repeat domain-containing 1
          Gene Aliases:
          1700012P16Rik, BEN, Cream1, ESTM9, GTF3, Gtf2il, MusTRD1, Tg(Alb1-Myc)166.8Sst, WBSCR11, X83320
          Chromosome Location:
          Chr.5: 134357661 - 134456716 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm01195479_m1
          Size
          Availability Made To Order
          Catalog # 4351372
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Gtf2ird1
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001081462.2 NP_001074931.1 4 - 5 640 85
          NM_001081463.2 NP_001074932.1 4 - 5 640 85
          NM_001081464.2 NP_001074933.1 4 - 5 640 85
          NM_001081465.1 NP_001074934.1 4 - 5 583 85
          NM_001081466.2 NP_001074935.1 4 - 5 640 85
          NM_001081467.2 NP_001074936.1 4 - 5 640 85
          NM_001081468.2 NP_001074937.1 4 - 5 640 85
          NM_001081469.2 NP_001074938.1 4 - 5 640 85
          NM_001081470.2 NP_001074939.1 4 - 5 640 85
          NM_001244936.1 NP_001231865.1 4 - 5 640 85
          NM_020331.3 NP_065064.2 4 - 5 640 85
          GenBank mRNA AF247161.1 - 4 - 5 500 85
          AF257475.1 - 4 - 5 616 85
          AF260133.1 - 4 - 5 611 85
          AF343349.1 - 4 - 5 514 85
          AF343350.1 - 4 - 5 510 85
          AF343351.1 - 4 - 5 508 85
          AF497637.1 - 4 - 5 500 85
          AF497638.1 - 4 - 5 500 85
          AF497639.1 - 4 - 5 500 85
          AF497640.1 - 4 - 5 508 85
          AF497641.1 - 4 - 5 514 85
          AF497642.1 - 4 - 5 510 85
          AK078488.1 - 4 - 5 508 85
          AK079487.1 - 4 - 5 580 85
          AK147196.1 - 4 - 5 623 85
          AY030287.1 - 4 - 5 611 85
          AY030288.1 - 4 - 5 611 85
          AY030289.1 - 4 - 5 611 85
          BC050822.1 - 4 - 5 635 6393328 85
          BC057091.1 - 4 - 5 462 5703300 85
          BC058964.1 - 4 - 5 462 5703300 85
          BC083150.1 - 4 - 5 542 30459984 85

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC0500003371.mm.1

          Set Membership:

          Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Mus musculus Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Gtf2ird1

          Gene Ontology Categories:

          Function(s) Process(es)

          transcription, DNA-templated
          regulation of transcription, DNA-templated
          multicellular organism development
          embryo development
          transition between slow and fast fiber
          DNA binding
          DNA-binding transcription factor activity

          MicroArray Probes:

          Affymetrix

          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Clariom S Array PSR0500030395.mm.1 TC0500003371.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0500030395.mm.1 NM_001081462.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0500030395.mm.1 NM_001081462.2 Refseq
          Affymetrix Mouse Clariom S Array PSR0500030395.mm.1 NM_001081463.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0500030395.mm.1 NM_001081465.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0500030395.mm.1 NM_001081463.2 Refseq
          Affymetrix Mouse Clariom S Array PSR0500030395.mm.1 NM_001081464.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0500030395.mm.1 NM_001081464.2 Refseq
          Affymetrix Mouse Clariom S Array PSR0500030395.mm.1 NM_001081465.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0500030395.mm.1 NM_001081466.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0500030395.mm.1 NM_001081466.2 Refseq
          Affymetrix Mouse Clariom S Array PSR0500030395.mm.1 NM_001081467.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0500030395.mm.1 NM_001081467.2 Refseq
          Affymetrix Mouse Clariom S Array PSR0500030395.mm.1 NM_001081468.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0500030395.mm.1 NM_001081468.2 Refseq
          Affymetrix Mouse Clariom S Array PSR0500030395.mm.1 NM_001081469.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0500030395.mm.1 NM_001081469.2 Refseq
          Affymetrix Mouse Clariom S Array PSR0500030395.mm.1 NM_001081470.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0500030395.mm.1 NM_001081470.2 Refseq
          Affymetrix Mouse Clariom S Array PSR0500030395.mm.1 NM_001244936.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0500030395.mm.1 NM_001244936.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0500030395.mm.1 NM_020331.3.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0500030395.mm.1 NM_020331.3 Refseq
          Affymetrix Mouse Clariom S Array PSR0500030394.mm.1 TC0500003371.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0500030394.mm.1 NM_001081462.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0500030394.mm.1 NM_001081462.2 Refseq
          Affymetrix Mouse Clariom S Array PSR0500030394.mm.1 NM_001081463.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0500030394.mm.1 NM_001081463.2 Refseq
          Affymetrix Mouse Clariom S Array PSR0500030394.mm.1 NM_001081464.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0500030394.mm.1 NM_001081464.2 Refseq
          Affymetrix Mouse Clariom S Array PSR0500030394.mm.1 NM_001081465.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0500030394.mm.1 NM_001081465.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0500030394.mm.1 NM_001081466.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0500030394.mm.1 NM_001081466.2 Refseq
          Affymetrix Mouse Clariom S Array PSR0500030394.mm.1 NM_001081467.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0500030394.mm.1 NM_001081467.2 Refseq
          Affymetrix Mouse Clariom S Array PSR0500030394.mm.1 NM_001081468.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0500030394.mm.1 NM_001081468.2 Refseq
          Affymetrix Mouse Clariom S Array PSR0500030394.mm.1 NM_001081469.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0500030394.mm.1 NM_001081469.2 Refseq
          Affymetrix Mouse Clariom S Array PSR0500030394.mm.1 NM_001081470.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0500030394.mm.1 NM_001081470.2 Refseq
          Affymetrix Mouse Clariom S Array PSR0500030394.mm.1 NM_001244936.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0500030394.mm.1 NM_001244936.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0500030394.mm.1 NM_020331.3.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0500030394.mm.1 NM_020331.3 Refseq

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-97865cc7b-9fgnx:80/100.66.128.131:80.
          git-commit: d53bc399386730d6c5c6d01fe94b21e0ad3fdbfa
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.33.0-2025.07.30-1.0