Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm01195480_m1
          See other GTF2IRD1 GE Assays ›
          Gene Symbol:
          Gtf2ird1
          Gene Name:
          general transcription factor II I repeat domain-containing 1
          Gene Aliases:
          1700012P16Rik, BEN, Cream1, ESTM9, GTF3, Gtf2il, MusTRD1, Tg(Alb1-Myc)166.8Sst, WBSCR11, X83320
          Chromosome Location:
          Chr.5: 134357661 - 134456716 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm01195480_m1
          Size
          Availability Made To Order
          Catalog # 4351372
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Gtf2ird1
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001081462.2 NP_001074931.1 5 - 6 819 60
          NM_001081463.2 NP_001074932.1 5 - 6 819 60
          NM_001081464.2 NP_001074933.1 5 - 6 819 60
          NM_001081465.1 NP_001074934.1 5 - 6 762 60
          NM_001081466.2 NP_001074935.1 5 - 6 819 60
          NM_001081467.2 NP_001074936.1 5 - 6 819 60
          NM_001081468.2 NP_001074937.1 5 - 6 819 60
          NM_001081469.2 NP_001074938.1 5 - 6 819 60
          NM_001081470.2 NP_001074939.1 5 - 6 819 60
          NM_001244936.1 NP_001231865.1 5 - 6 819 60
          NM_020331.3 NP_065064.2 5 - 6 819 60
          GenBank mRNA AF247161.1 - 5 - 6 679 60
          AF257475.1 - 5 - 6 795 60
          AF260133.1 - 5 - 6 790 60
          AF343348.1 - 5 - 6 687 60
          AF343349.1 - 5 - 6 693 60
          AF343350.1 - 5 - 6 689 60
          AF343351.1 - 5 - 6 687 60
          AF497637.1 - 5 - 6 679 60
          AF497638.1 - 5 - 6 679 60
          AF497639.1 - 5 - 6 679 60
          AF497640.1 - 5 - 6 687 60
          AF497641.1 - 5 - 6 693 60
          AF497642.1 - 5 - 6 689 60
          AK078488.1 - 5 - 6 687 60
          AK079487.1 - 5 - 6 759 60
          AK147196.1 - 5 - 6 802 60
          AY030287.1 - 5 - 6 790 60
          AY030288.1 - 5 - 6 790 60
          AY030289.1 - 5 - 6 790 60
          BC050822.1 - 5 - 6 814 6393328 60
          BC057091.1 - 5 - 6 641 5703300 60
          BC058964.1 - 5 - 6 641 5703300 60
          BC083150.1 - 5 - 6 721 30459984 60

          Back To Top

          More Information


          Set Membership:

          Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Mus musculus Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Gtf2ird1

          Gene Ontology Categories:

          Function(s) Process(es)

          transcription, DNA-templated
          regulation of transcription, DNA-templated
          multicellular organism development
          embryo development
          transition between slow and fast fiber
          DNA binding
          DNA-binding transcription factor activity

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-65cb46898d-nlgdw:80/100.66.128.131:80.
          git-commit: e5493f66f4597fc66e516c8a379ca3b6bed48f5b
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.33.0-2025.07.30-1.0