Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm01195805_m1
          See other TAOK3 GE Assays ›
          Gene Symbol:
          Taok3
          Gene Name:
          TAO kinase 3
          Gene Aliases:
          2900006A08Rik, A130052D22, A430105I05Rik
          Chromosome Location:
          Chr.5: 117120112 - 117275219 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm01195805_m1
          Size
          Availability Made To Order
          Catalog # 4351372
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Taok3
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001081308.2 NP_001074777.1 15 - 16 1976 110
          NM_001199685.1 NP_001186614.1 16 - 17 2084 110
          NM_183306.2 NP_899129.2 16 - 17 2061 110
          GenBank mRNA AK039665.1 - 3 - 4 634 110
          AK155208.1 - 15 - 16 1972 110
          AK188887.1 - 1 - 2 115 110
          BC158072.1 - 16 - 17 2026 9088167 110

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC0500001288.mm.1

          Set Membership:

          Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Mus musculus Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Taok3

          Gene Ontology Categories:

          Function(s) Process(es)

          MAPK cascade
          DNA repair
          protein phosphorylation
          cellular response to DNA damage stimulus
          mitotic G2 DNA damage checkpoint signaling
          regulation of mitotic cell cycle
          phosphorylation
          stress-activated protein kinase signaling cascade
          positive regulation of stress-activated MAPK cascade
          regulation of apoptotic process
          positive regulation of JUN kinase activity
          negative regulation of JNK cascade
          positive regulation of JNK cascade
          protein autophosphorylation
          nucleotide binding
          protein kinase activity
          protein serine/threonine kinase activity
          MAP kinase kinase kinase activity
          ATP binding
          kinase activity
          transferase activity
          transferase activity, transferring phosphorus-containing groups

          MicroArray Probes:

          Affymetrix

          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Clariom S Array PSR0500011239.mm.1 TC0500001288.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0500011239.mm.1 NM_001081308.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0500011239.mm.1 NM_001081308.2 Refseq
          Affymetrix Mouse Clariom S Array PSR0500011239.mm.1 NM_001199685.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0500011239.mm.1 NM_001199685.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0500011239.mm.1 NM_183306.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0500011239.mm.1 NM_183306.2 Refseq
          Affymetrix Mouse Clariom S Array PSR0500011240.mm.1 TC0500001288.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0500011240.mm.1 NM_001081308.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0500011240.mm.1 NM_001081308.2 Refseq
          Affymetrix Mouse Clariom S Array PSR0500011240.mm.1 NM_001199685.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0500011240.mm.1 NM_001199685.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0500011240.mm.1 NM_183306.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0500011240.mm.1 NM_183306.2 Refseq

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-74594944d-pk6zc:80/100.66.130.62:80.
          git-commit: ab2b2261c2971e97b1ae408e3ad10520b8413932
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.31.0-2025.06.23-1.0