Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm01196328_g1
          See other NCF1 GE Assays ›
          Gene Symbol:
          Ncf1
          Gene Name:
          neutrophil cytosolic factor 1
          Gene Aliases:
          NCF-47K, NOXO2, Ncf-1, p47-phox, p47<phox>, p47phox
          Chromosome Location:
          Chr.5: 134220053 - 134229625 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm01196328_g1
          Size
          Availability Made To Order
          Catalog # 4351372
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Ncf1
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001286037.1 NP_001272966.1 2 - 3 207 85
          NM_010876.4 NP_035006.3 2 - 3 207 85
          NR_104383.1 2 - 3 207 85
          GenBank mRNA AB002663.1 - 2 - 3 186 85
          AK149619.1 - 2 - 3 208 85
          AK149668.1 - 2 - 3 174 85
          AK149732.1 - 2 - 3 208 85
          AK151316.1 - 2 - 3 174 85
          AK151593.1 - 2 - 3 172 85
          AK152386.1 - 2 - 3 170 85
          AK152992.1 - 2 - 3 174 85
          AK162308.1 - 2 - 3 174 85
          AK162623.1 - 2 - 3 177 85
          AK170462.1 - 2 - 3 174 85
          AK171559.1 - 2 - 3 173 85
          BC055836.1 - 2 - 3 161 3983481 85
          L11455.1 - 2 - 3 144 85

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC0500003368.mm.1

          Set Membership:

          Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Ncf1

          Gene Ontology Categories:

          Function(s) Process(es)

          response to yeast
          leukocyte mediated cytotoxicity
          respiratory burst involved in defense response
          protein targeting to membrane
          leukotriene metabolic process
          NADP catabolic process
          apoptotic process
          inflammatory response
          cellular defense response
          cell population proliferation
          response to bacterium
          superoxide anion generation
          defense response to bacterium
          respiratory burst
          negative regulation of smooth muscle contraction
          hydrogen peroxide biosynthetic process
          defense response to Gram-positive bacterium
          defense response to fungus
          neutrophil-mediated killing of gram-positive bacterium
          neutrophil-mediated killing of fungus
          protein binding
          lipid binding
          superoxide-generating NAD(P)H oxidase activity
          SH3 domain binding
          phosphatidylinositol binding
          phosphatidylinositol-3,4-bisphosphate binding

          MicroArray Probes:

          Affymetrix

          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Clariom S Array PSR0500030267.mm.1 TC0500003368.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0500030267.mm.1 NM_001286037.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0500030267.mm.1 NM_010876.4 Refseq
          Affymetrix Mouse Clariom S Array PSR0500030267.mm.1 NR_104383.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0500030266.mm.1 TC0500003368.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0500030266.mm.1 NM_001286037.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0500030266.mm.1 NM_010876.4 Refseq
          Affymetrix Mouse Clariom S Array PSR0500030266.mm.1 NR_104383.1 Refseq

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-65cb46898d-nlgdw:80/100.66.128.131:80.
          git-commit: e5493f66f4597fc66e516c8a379ca3b6bed48f5b
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.33.0-2025.07.30-1.0