Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm01196421_m1
          See other MIAT GE Assays ›
          Gene Symbol:
          Miat
          Gene Name:
          myocardial infarction associated transcript (non-protein coding)
          Gene Aliases:
          3632434I06, A230057G18Rik, Rncr2, gomafu
          Chromosome Location:
          Chr.5: 112213228 - 112228948 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm01196421_m1
          Size
          Availability Made To Order
          Catalog # 4426961
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Miat
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NR_003718.2 4 - 5 659 80
          NR_033657.1 4 - 5 703 80
          GenBank mRNA AB294525.1 - 4 - 5 453 80
          AB294526.1 - 4 - 5 417 80
          AB294527.1 - 4 - 5 409 80
          AB294528.1 - 3 - 4 361 80
          AB300594.1 - 4 - 5 646 80
          AK038726.1 - 4 - 5 704 80
          AK053540.1 - 4 - 5 647 80
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Transcript Location Amplicon Length
          GenBankmRNA CJ098960.1 - 1 - 2 90 Chr. 5 - 112221511 - 112224299 [-] on Build GRCm38 80
          GenBankmRNA CJ103439.1 - 1 - 2 90 Chr. 5 - 112221502 - 112224299 [-] on Build GRCm38 80
          GenBankmRNA BY252337.1 - 4 - 5 402 Chr. 5 - 112221826 - 112228691 [-] on Build GRCm38 80
          GenBankmRNA CJ098394.1 - 1 - 2 90 Chr. 5 - 112221424 - 112224299 [-] on Build GRCm38 80
          GenBankmRNA CJ103735.1 - 1 - 2 90 Chr. 5 - 112221518 - 112224299 [-] on Build GRCm38 80
          GenBankmRNA CJ099017.1 - 1 - 2 90 Chr. 5 - 112221519 - 112224299 [-] on Build GRCm38 80
          GenBankmRNA CJ099062.1 - 1 - 2 90 Chr. 5 - 112221510 - 112224299 [-] on Build GRCm38 80
          GenBankmRNA CJ103281.1 - 1 - 2 90 Chr. 5 - 112221495 - 112224299 [-] on Build GRCm38 80
          GenBankmRNA CJ103381.1 - 1 - 2 90 Chr. 5 - 112221509 - 112224299 [-] on Build GRCm38 80
          GenBankmRNA CJ099362.1 - 1 - 2 90 Chr. 5 - 112221499 - 112224299 [-] on Build GRCm38 80
          GenBankmRNA CF534566.1 - 3 - 4 239 Chr. 5 - 112221369 - 112228644 [-] on Build GRCm38 80

          Adjacent Gene Details

          Gene name Gene Symbol Alias Distance Relative Direction Location Chromsome
          Upstream RIKEN cDNA 1700028D13 gene 1700028D13Rik -11.5kb Anti-sense Chr. 5 - 112206763 - 112210002 on NCBI Build GRCm38
          Downstream crystallin,
          beta A4
          Cryba4 22.2kb Sense Chr. 5 - 112246492 - 112263555 on NCBI Build GRCm38

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC0500003012.mm.1

          Set Membership:

          Amplicon length between 71 and 85 Mus musculus Probe spans exons

          Gene Ontology Categories:

          Function(s) Process(es)

          glial cell differentiation
          stem cell population maintenance
          amacrine cell differentiation
          retinal rod cell development
          molecular_function

          MicroArray Probes:

          Affymetrix

          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Clariom S Array PSR0500026705.mm.1 TC0500003012.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0500026705.mm.1 NR_003718.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0500026705.mm.1 NR_003718.2 Refseq
          Affymetrix Mouse Clariom S Array PSR0500026705.mm.1 NR_033657.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0500026705.mm.1 NR_033657.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0500026704.mm.1 TC0500003012.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0500026704.mm.1 NR_003718.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0500026704.mm.1 NR_003718.2 Refseq
          Affymetrix Mouse Clariom S Array PSR0500026704.mm.1 NR_033657.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0500026704.mm.1 NR_033657.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0500026710.mm.1 TC0500003012.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0500026710.mm.1 NR_003718.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0500026710.mm.1 NR_003718.2 Refseq
          Affymetrix Mouse Clariom S Array PSR0500026710.mm.1 NR_033657.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0500026710.mm.1 NR_033657.1 Refseq

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-74594944d-pk6zc:80/100.66.130.62:80.
          git-commit: ab2b2261c2971e97b1ae408e3ad10520b8413932
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.31.0-2025.06.23-1.0