Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Clariom S Array |
PSR1900005441.mm.1 |
TC1900000609.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1900005441.mm.1 |
NM_145501.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1900005441.mm.1 |
NM_145501.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1900005443.mm.1 |
TC1900000609.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1900005443.mm.1 |
NM_145501.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1900005443.mm.1 |
NM_145501.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1900005445.mm.1 |
TC1900000609.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1900005445.mm.1 |
NM_145501.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1900005445.mm.1 |
NM_145501.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1900005440.mm.1 |
TC1900000609.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1900005440.mm.1 |
NM_145501.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1900005440.mm.1 |
NM_145501.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1900005446.mm.1 |
TC1900000609.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1900005446.mm.1 |
NM_145501.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1900005446.mm.1 |
NM_145501.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1900005444.mm.1 |
TC1900000609.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1900005444.mm.1 |
NM_145501.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1900005444.mm.1 |
NM_145501.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1900005442.mm.1 |
TC1900000609.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1900005442.mm.1 |
NM_145501.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1900005442.mm.1 |
NM_145501.2 |
Refseq |