Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Clariom S Array |
PSR1900010748.mm.1 |
TC1900001144.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1900010748.mm.1 |
NM_001025307.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1900010748.mm.1 |
NM_001025307.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1900010748.mm.1 |
NM_001286543.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1900010748.mm.1 |
NM_011502.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1900010748.mm.1 |
NM_152220.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1900010747.mm.1 |
TC1900001144.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1900010747.mm.1 |
NM_001025307.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1900010747.mm.1 |
NM_001025307.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1900010747.mm.1 |
NM_001286543.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1900010747.mm.1 |
NM_011502.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1900010747.mm.1 |
NM_152220.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1900010745.mm.1 |
TC1900001144.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1900010745.mm.1 |
NM_001025307.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1900010745.mm.1 |
NM_001025307.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1900010745.mm.1 |
NM_001286543.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1900010745.mm.1 |
NM_011502.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1900010745.mm.1 |
NM_152220.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1900010746.mm.1 |
TC1900001144.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1900010746.mm.1 |
NM_001025307.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1900010746.mm.1 |
NM_001025307.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1900010746.mm.1 |
NM_001286543.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1900010746.mm.1 |
NM_011502.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1900010746.mm.1 |
NM_152220.2 |
Refseq |