Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm01199175_m1
          See other 4933430A20RIK GE Assays ›
          Gene Symbol:
          4933430A20Rik
          Gene Name:
          RIKEN cDNA 4933430A20 gene
          Gene Aliases:
          -
          Chromosome Location:
          -
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm01199175_m1
          Size
          Availability Made To Order
          Catalog # 4426961
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: 4933430A20Rik
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          GenBank mRNA AK016987.1 - 1 - 2 866 78

          Adjacent Gene Details

          Gene name Gene Symbol Alias Distance Relative Direction Location Chromsome
          Upstream predicted gene,
          39164
          Gm39164 -66.0 Anti-sense Chr. 8 - 36174140 - 36176075 on NCBI Build GRCm38
          Downstream LON peptidase N-terminal domain and ring finger 1 Lonrf1 37.6kb Anti-sense Chr. 8 - 36216064 - 36249516 on NCBI Build GRCm38

          Back To Top

          More Information


          Set Membership:

          Amplicon length between 71 and 85 3' Most Mus musculus 1700 Probe spans exons

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2400101 AK016987.1 mouse_transcriptome
          Agilent Six_ChIP_02_of_03_v2 A_68_P25784298 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P07362904 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06717449 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P07362901 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25784300 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25784309 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25784329 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06746208 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25784316 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P07272538 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25784294 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25784320 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25784313 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25784333 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P07088367 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P07173569 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25784321 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25784296 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25784306 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25784327 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06886237 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25784315 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25784286 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25784326 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25784314 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P07080038 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25784301 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25784311 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25784334 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P07362907 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P07173571 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix HT MG-430 PM Array Plate 1432025_PM_at - -
          Mouse Genome 430 2.0 Array 1432025_at - -
          Mouse Genome 430 2.0 Array 1432025_PM_at - -
          Mouse Expression Set 430 1432025_at - -
          Mouse Expression Set 430 1432025_PM_at - -
          Affymetrix HT Mouse Genome 430B Array 1432025_at - -
          Affymetrix HT Mouse Genome 430B Array 1432025_PM_at - -
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 888901 - -
          Array Name Probe IDs Targets Target Type
          MouseWG-6 v2 Expression BeadChip 101410440 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-65cb46898d-nlgdw:80/100.66.128.131:80.
          git-commit: e5493f66f4597fc66e516c8a379ca3b6bed48f5b
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.33.0-2025.07.30-1.0