Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm01199402_m1
          See other GM3916 GE Assays ›
          Gene Symbol:
          Gm3916
          Gene Name:
          predicted gene 3916
          Gene Aliases:
          -
          Chromosome Location:
          -
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm01199402_m1
          Size
          Availability Made To Order
          Catalog # 4426961
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Gm3916
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          GenBank mRNA AK086006.1 - 1 - 2 91 117
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Transcript Location Amplicon Length
          GenBankmRNA BB662335.1 - 1 - 2 88 Chr. 19 - 27260732 - 27262321 [-] on Build GRCm38 117

          Adjacent Gene Details

          Gene name Gene Symbol Alias Distance Relative Direction Location Chromsome
          Upstream predicted gene,
          35438
          Gm35438 Sense Chr. 19 - 27101951 - 27316698 on NCBI Build GRCm38
          Downstream potassium channel,
          subfamily V,
          member 2
          Kcnv2 KV11.1 59.7kb Anti-sense Chr. 19 - 27322060 - 27337179 on NCBI Build GRCm38

          Back To Top

          More Information


          Additional Information:

          Based on assay re-evaluation, this assay may detect transcript(s) from off target gene(s). Please contact Tech Support if you need more information.

          Set Membership:

          Amplicon length greater than or equal to 101 3' Most Mus musculus 1700 Probe spans exons

          MicroArray Probes:

          Agilent Affymetrix ABI

          Array Name Probe IDs Targets Target Type
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P105293 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_66_P106983 BB520468.2 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_66_P106983 AK086006.1 mouse_transcriptome
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P105293 - -
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P106983 BB520468.2 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P106983 AK086006.1 mouse_transcriptome
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P105293 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32039948 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P14441168 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32039941 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32039963 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32039966 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32039965 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P14304508 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32039961 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32039962 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P14255561 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32039990 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P14260375 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32039968 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32039953 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P14304511 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32039951 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P14304509 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P14403218 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32039954 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32039945 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32039958 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32039943 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P14403213 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32039987 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P14436574 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32039957 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32039976 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P14304510 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P14436570 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32039978 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32039981 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32039949 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32039979 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix HT MG-430 PM Array Plate 1442304_PM_at - -
          Mouse Genome 430 2.0 Array 1442304_at - -
          Mouse Genome 430 2.0 Array 1442304_PM_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 100042588_PM_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 100042588_at - -
          Mouse Expression Set 430 1442304_PM_at - -
          Mouse Expression Set 430 1442304_at - -
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 868077 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-865d7754c7-mcq8j:80/100.66.130.62:80.
          git-commit: 7ff36f61b459eb8f02ba5a263f0e3ce367ca82d3
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.32.0-2025.07.28-1.0