Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm01199903_m1
          See other RAD9B GE Assays ›
          Gene Symbol:
          Rad9b
          Gene Name:
          RAD9 homolog B
          Gene Aliases:
          A630082N15Rik, BC021784
          Chromosome Location:
          Chr.5: 122323223 - 122354272 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm01199903_m1
          Size
          Availability Made To Order
          Catalog # 4351372
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Rad9b
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_144912.3 NP_659161.2 10 - 11 1120 80
          GenBank mRNA AK136504.1 - 10 - 11 1156 80
          AY297460.1 - 10 - 11 1120 80
          BC021784.1 - 10 - 11 1120 4162418 80

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC0500003206.mm.1

          Set Membership:

          Ambion siRNA available Amplicon length between 71 and 85 3' Most Mammalian Gene Collection Mus musculus 1700 Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Rad9b

          Gene Ontology Categories:

          Function(s) Process(es)

          cell cycle checkpoint signaling
          DNA replication checkpoint signaling
          DNA damage checkpoint signaling
          DNA repair
          mitotic intra-S DNA damage checkpoint signaling
          cellular response to ionizing radiation
          nucleic acid phosphodiester bond hydrolysis
          3'-5' exonuclease activity

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P24049901 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P24049898 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P24049900 - -
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P513013 AY297460.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P513013 BC021784.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P513013 AK136504.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P513013 NM_144912.3 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P513013 AY297460.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P513013 BC021784.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P513013 AK136504.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P513013 NM_144912.3 mouse_transcriptome
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 767159 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 263120 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 215667 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 125594 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 514787 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 437503 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 617136 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 117189 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 156582 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 154450 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 351679 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 935171 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 83995 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 874143 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 567652 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 838850 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 74507 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 145827 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 11032 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P116298 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P152661 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P513013 AY297460.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P513013 BC021784.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P513013 AK136504.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P513013 NM_144912.3 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P513013 AY297460.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P513013 BC021784.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P513013 AK136504.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P513013 NM_144912.3 mouse_transcriptome
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24049888 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24049879 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24049887 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24049896 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24049875 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24049883 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24049877 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24049876 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24049891 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24049882 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24049884 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24049899 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24049898 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24049870 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24049873 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24049886 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24049871 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24049893 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24049894 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24049881 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24049901 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P01207260 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P01207288 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P01207278 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P152661 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P05724285 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P116298 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P01207269 - -
          Whole Mouse Genome G4122A A_51_P513013 AY297460.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P513013 BC021784.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P513013 AK136504.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P513013 NM_144912.3 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P513013 AY297460.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P513013 BC021784.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P513013 AK136504.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P513013 NM_144912.3 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P106341 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P116298 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P152661 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_67_P01207278 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P04782194 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P04430154 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P04430155 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P04782182 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P04782185 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P24049898 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P24049901 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P24049900 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24049792 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24049816 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24049765 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24049694 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24049769 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24049774 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24049815 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24049817 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24049886 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24049761 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24049689 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24049687 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24049800 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24049870 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24049824 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24049781 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24049901 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24049877 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24049882 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24049884 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24049889 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24049899 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24049867 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24049818 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24049811 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24049865 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24049770 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24049820 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24049875 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24049821 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24049750 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24049839 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24049894 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24049810 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24049872 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24049796 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24049748 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24049680 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24049848 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24049759 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24049746 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24049757 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24049690 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24049888 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24049857 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24049822 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24049832 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24049845 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24049749 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24049766 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24049790 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24049871 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24049763 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24049879 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24049773 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24049684 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24049873 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24049698 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24049853 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24049856 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24049847 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24049859 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24049813 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24049838 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24049771 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24049688 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24049755 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24049812 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24049686 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24049830 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24049697 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24049860 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24049809 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24049891 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24049682 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24049786 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24049784 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24049840 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24049854 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24049753 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24049874 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24049869 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P24049896 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P24049886 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P24049869 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P24049884 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P04686878 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P04686900 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P04686909 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P24049748 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P24049873 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P24049860 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P24049881 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P24049879 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P24049872 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P24049870 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P24049899 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P24049749 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P24049875 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P24049874 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P24049894 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P24049889 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P24049882 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P24049893 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P24049866 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P24049888 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P24049877 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P04779435 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P24049862 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P04750120 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P24049897 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P04779439 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P04686895 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P24049891 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P04686875 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P04779445 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10533471 - -
          Affymetrix Mouse Gene 1.0 ST Array 10533468 - -
          Affymetrix Mouse Gene 1.0 ST Array 10533465 - -
          Affymetrix Mouse Gene 1.0 ST Array 10533470 - -
          Affymetrix Mouse Gene 1.0 ST Array 10533473 - -
          Affymetrix Mouse Gene 1.0 ST Array 10533472 - -
          Affymetrix Mouse Gene 1.0 ST Array 10533464 - -
          Affymetrix Mouse Gene 1.0 ST Array 10533466 - -
          Affymetrix Mouse Gene 1.0 ST Array 10533467 - -
          Affymetrix Mouse Gene 1.0 ST Array 10533469 - -
          Affymetrix Mouse Gene 1.0 ST Array 10533463 - -
          Affymetrix HT MG-430 PM Array Plate 1439955_PM_at - -
          Affymetrix HT MG-430 PM Array Plate 1425800_PM_at - -
          Affymetrix Mouse Gene 1.1 ST Array 10533462 - -
          Mouse Genome 430 2.0 Array 1439955_PM_at - -
          Mouse Genome 430 2.0 Array 1425800_at - -
          Mouse Genome 430 2.0 Array 1425800_PM_at - -
          Mouse Genome 430 2.0 Array 1439955_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 231724_PM_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 231724_at - -
          Affymetrix Mouse Clariom S Array PSR0500028653.mm.1 TC0500003206.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0500028653.mm.1 NM_144912.3.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0500028653.mm.1 NM_144912.3 Refseq
          Affymetrix Mouse Clariom S Array PSR0500028652.mm.1 TC0500003206.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0500028652.mm.1 NM_144912.3.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0500028652.mm.1 NM_144912.3 Refseq
          Mouse Expression Set 430 1425800_PM_at - -
          Mouse Expression Set 430 1425800_at - -
          Mouse Expression Set 430 1439955_PM_at - -
          Mouse Expression Set 430 1439955_at - -
          Mouse Genome 430A 2.0 Array 1425800_at - -
          Mouse Genome 430A 2.0 Array 1425800_PM_at - -
          Affymetrix HT Mouse Genome 430A Array 1425800_PM_at - -
          Affymetrix HT Mouse Genome 430A Array 1425800_at - -
          Affymetrix HT Mouse Genome 430B Array 1439955_PM_at - -
          Affymetrix HT Mouse Genome 430B Array 1439955_at - -
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 558044 - -
          Array Name Probe IDs Targets Target Type
          Illumina mouse-6 v1.0 expression beadchip 2320064 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0002320064 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_1225080 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_1225080 - -
          mouse-6 v1.1 (Illumina) scl0231724.1_307-S - -
          Illumina Sentrix mouse-6.v1 bead array 2320064 - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl0231724.1_307-S - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 2320064 - -
          MouseRef-8 v2 Expression BeadChip ILMN_1225080 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_1225080 - -
          Illumina Sentrix Mouse-6 Expression BeadChip (3) scl0231724.1_307-S - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-784b477dbd-pq4vn:80/100.66.130.62:80.
          git-commit: d41a97e986437cfbf980934f14197a4b4d3eb741
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.31.0-2025.06.23-1.0