Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm01200471_m1
          See other HEMT1 GE Assays ›
          Gene Symbol:
          Hemt1
          Gene Name:
          hematopoietic cell transcript 1
          Gene Aliases:
          1700057K19Rik, HemT
          Chromosome Location:
          Chr.15: 74818880 - 74846164 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm01200471_m1
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Hemt1
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_010416.2 NP_034546.2 3 - 4 366 101
          GenBank mRNA AK005659.1 - 3 - 4 371 101
          AK005823.1 - 2 - 3 139 101

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC1500000545.mm.1

          Set Membership:

          Ambion siRNA available Amplicon length greater than or equal to 101 3' Most Mus musculus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Hemt1

          Gene Ontology Categories:

          Function(s) Process(es)

          biological_process
          molecular_function

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2050453 AK006826.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2050453 NM_010416.2 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2050453 AK005823.1 mouse_transcriptome
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 460251 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 970527 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2050453 AK006826.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2050453 NM_010416.2 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2050453 AK005823.1 mouse_transcriptome
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30339837 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30339796 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30339815 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30339776 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P08418420 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P08418411 - -
          Whole Mouse Genome G4122A A_52_P531140 AJ242831.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P531140 BC119337.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P531140 AJ011022.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P531140 NM_010416.2 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P531140 AJ011023.1 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P115705 - -
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P129492 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30339893 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30339817 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30339891 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30339884 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30339883 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30339886 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30339761 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30339860 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30339760 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30339861 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30339857 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30339776 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30339767 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30339836 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30339817 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30339815 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30339796 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10424673 - -
          Affymetrix Mouse Gene 1.0 ST Array 10424672 - -
          Affymetrix Mouse Gene 1.0 ST Array 10424671 - -
          Affymetrix Mouse Gene 1.1 ST Array 10424670 - -
          Mouse Genome 430 2.0 Array 1422400_a_at - -
          Mouse Genome 430 2.0 Array 1422400_PM_a_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 15202_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 15202_PM_at - -
          Affymetrix Mouse Clariom S Array PSR1500003754.mm.1 TC1500000545.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1500003754.mm.1 NM_010416.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1500003754.mm.1 NM_010416.2 Refseq
          Mouse Expression Set 430 1422400_PM_a_at - -
          Mouse Expression Set 430 1422400_a_at - -
          Murine Genome U74v2 Set 102949_PM_g_at - -
          Murine Genome U74v2 Set 102949_g_at - -
          Murine Genome U74v2 Set 102950_at - -
          Murine Genome U74v2 Set 102950_PM_at - -
          Murine Genome U74v2 Set 168868_PM_r_at - -
          Murine Genome U74v2 Set 168868_r_at - -
          Mouse Genome 430A 2.0 Array 1422400_a_at - -
          Mouse Genome 430A 2.0 Array 1422400_PM_a_at - -
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 932534 - -
          Array Name Probe IDs Targets Target Type
          MouseWG-6 v2 Expression BeadChip 102650750 - -
          Illumina mouse-6 v1.0 expression beadchip 6510131 - -
          Illumina mouse-6 v1.0 expression beadchip 102650750 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0006510131 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_3162214 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_2657905 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_2417754 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_3162214 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_2657905 - -
          mouse-6 v1.1 (Illumina) scl10675.1.1_191-S - -
          mouse-6 v1.1 (Illumina) GI_84662775-S - -
          mouse-6 v1.1 (Illumina) scl0015202.2_98-S - -
          Illumina Sentrix mouse-6.v1 bead array 102650750 - -
          Illumina Sentrix mouse-6.v1 bead array 6510131 - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl0015202.2_98-S - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 102650750 - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 6510131 - -
          MouseRef-8 v2 Expression BeadChip ILMN_3162214 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_2657905 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_3162214 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_2417754 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-65cb46898d-nlgdw:80/100.66.128.131:80.
          git-commit: e5493f66f4597fc66e516c8a379ca3b6bed48f5b
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.33.0-2025.07.30-1.0