Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Clariom S Array |
PSR0800016568.mm.1 |
TC0800002153.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0800016568.mm.1 |
NM_001039562.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0800016568.mm.1 |
NM_001039562.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0800016572.mm.1 |
TC0800002153.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0800016572.mm.1 |
NM_001039562.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0800016572.mm.1 |
NM_001039562.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0800016569.mm.1 |
TC0800002153.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0800016569.mm.1 |
NM_001039562.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0800016569.mm.1 |
NM_001039562.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0800016571.mm.1 |
TC0800002153.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0800016571.mm.1 |
NM_001039562.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0800016571.mm.1 |
NM_001039562.1 |
Refseq |