Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm01201456_g1
          See other POP5 GE Assays ›
          Gene Symbol:
          Pop5
          Gene Name:
          processing of precursor 5, ribonuclease P/MRP family (S. cerevisiae)
          Gene Aliases:
          1500019J17Rik, 2700077E03Rik, AW049989, Rnasep3
          Chromosome Location:
          Chr.5: 115235851 - 115240972 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm01201456_g1
          Size
          Availability Made To Order
          Catalog # 4351372
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Pop5
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_026398.4 NP_080674.1 5 - 6 663 73
          GenBank mRNA AK005290.1 - 4 - 5 419 73
          AK012421.1 - 4 - 5 413 73
          AK012523.1 - 421 73
          AK013257.1 - 3 - 4 400 73
          BC022670.1 - 5 - 6 687 4224973 73

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC0500001251.mm.1

          Set Membership:

          Ambion siRNA available Amplicon length between 71 and 85 3' Most Mammalian Gene Collection Detect Genomic DNA Mus musculus 1700 Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Pop5

          Gene Ontology Categories:

          Function(s) Process(es)

          rRNA processing
          tRNA processing
          RNA metabolic process
          RNA phosphodiester bond hydrolysis
          RNA phosphodiester bond hydrolysis, endonucleolytic
          ribonuclease MRP activity
          ribonuclease P activity
          ribonuclease activity
          hydrolase activity

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P24008565 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P24008560 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P24008581 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P24008562 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P24008561 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P24008564 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P24008583 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P24008580 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P24008582 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P24008563 - -
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2133431 NM_026398.4 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2090718 AK013257.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2090718 BC022670.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2090718 AK012421.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2090718 NM_026398.4 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2090718 AK005290.1 mouse_transcriptome
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 653434 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 125834 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 52158 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 515581 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2090718 AK013257.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2090718 BC022670.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2090718 AK012421.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2090718 NM_026398.4 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2090718 AK005290.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2133431 NM_026398.4 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P490198 AK012523.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P490198 AK013257.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P490198 BC022670.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P490198 AK012421.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P490198 NM_026398.4 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P490204 AK013257.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P490204 BC022670.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P490204 AK012421.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P490204 NM_026398.4 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P490204 AK005290.1 mouse_transcriptome
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24008570 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24008561 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24008537 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24008564 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24008540 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24008560 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24008580 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24008536 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24008557 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24008548 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24008575 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24008551 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24008549 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24008541 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24008573 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24008567 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24008568 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24008571 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24008581 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24008579 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24008545 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24008554 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24008569 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24008577 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24008538 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24008562 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24008543 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P01194370 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P01194375 - -
          Whole Mouse Genome G4122A A_51_P490204 AK013257.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P490204 BC022670.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P490204 AK012421.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P490204 NM_026398.4 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P490204 AK005290.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P490198 AK012523.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P490198 AK013257.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P490198 BC022670.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P490198 AK012421.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P490198 NM_026398.4 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P105718 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P04207221 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P04207220 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P04207215 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P04207225 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P24008564 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P24008565 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P24008561 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P24008562 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P24008580 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P24008583 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P24008582 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P24008581 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P24008563 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P24008560 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24008536 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24008579 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24008603 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24008599 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24008522 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24008584 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24008577 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24008546 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24008602 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24008564 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24008549 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24008571 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24008555 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24008597 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24008601 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24008552 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24008569 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24008585 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24008582 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24008586 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24008545 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24008537 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24008540 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24008596 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24008524 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24008570 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24008604 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24008548 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24008567 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24008551 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24008580 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24008541 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24008594 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24008558 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24008544 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24008560 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24008575 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24008529 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P04692837 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P04383349 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P24008560 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P04506787 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P04476406 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P24008603 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P04854660 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P04469979 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P24008554 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P04207214 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P24008599 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P24008546 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P24008604 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P24008552 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P24008570 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P04612805 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P24008575 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P24008557 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P04476419 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P24008558 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P24008538 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P04207597 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P24008537 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P04476414 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P24008567 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P04469983 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P04865429 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P24008531 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P04383348 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P24008551 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P24008601 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P04476405 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P04207222 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P24008594 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P24008540 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P24008578 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P24008529 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P24008530 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P24008543 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P24008589 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P24008536 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10524666 - -
          Affymetrix Mouse Gene 1.0 ST Array 10524663 - -
          Affymetrix Mouse Gene 1.0 ST Array 10524664 - -
          Affymetrix Mouse Gene 1.0 ST Array 10524660 - -
          Affymetrix Mouse Gene 1.0 ST Array 10524665 - -
          Affymetrix Mouse Gene 1.0 ST Array 10524667 - -
          Affymetrix Mouse Gene 1.0 ST Array 10524661 - -
          Affymetrix Mouse Gene 1.0 ST Array 10524662 - -
          Affymetrix HT MG-430 PM Array Plate 1417972_PM_s_at - -
          Affymetrix HT MG-430 PM Array Plate 1431373_PM_at - -
          Affymetrix Mouse Gene 1.1 ST Array 10524659 - -
          Mouse Genome 430 2.0 Array 1431373_at - -
          Mouse Genome 430 2.0 Array 1417972_PM_s_at - -
          Mouse Genome 430 2.0 Array 1417972_s_at - -
          Mouse Genome 430 2.0 Array 1431373_PM_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 117109_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 117109_PM_at - -
          Affymetrix Mouse Clariom S Array PSR0500010779.mm.1 TC0500001251.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0500010779.mm.1 NM_026398.4.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0500010779.mm.1 NM_026398.4 Refseq
          Affymetrix Mouse Clariom S Array PSR0500010778.mm.1 TC0500001251.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0500010778.mm.1 NM_026398.4.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0500010778.mm.1 NM_026398.4 Refseq
          Affymetrix Mouse Clariom S Array PSR0500010780.mm.1 TC0500001251.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0500010780.mm.1 NM_026398.4.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0500010780.mm.1 NM_026398.4 Refseq
          Mouse Expression Set 430 1417972_s_at - -
          Mouse Expression Set 430 1417972_PM_s_at - -
          Mouse Expression Set 430 1431373_at - -
          Mouse Expression Set 430 1431373_PM_at - -
          Mouse Genome 430A 2.0 Array 1431373_at - -
          Mouse Genome 430A 2.0 Array 1431373_PM_at - -
          Mouse Genome 430A 2.0 Array 1417972_PM_s_at - -
          Mouse Genome 430A 2.0 Array 1417972_s_at - -
          Affymetrix HT Mouse Genome 430A Array 1431373_at - -
          Affymetrix HT Mouse Genome 430A Array 1417972_PM_s_at - -
          Affymetrix HT Mouse Genome 430A Array 1417972_s_at - -
          Affymetrix HT Mouse Genome 430A Array 1431373_PM_at - -
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 920448 - -
          Array Name Probe IDs Targets Target Type
          MouseWG-6 v2 Expression BeadChip 1500110 - -
          Illumina mouse-6 v1.0 expression beadchip 5270619 - -
          Illumina mouse-6 v1.0 expression beadchip 1500110 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0001500110 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0005270619 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_2661019 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_1248878 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_1248878 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_2661019 - -
          mouse-6 v1.1 (Illumina) scl27367.4.1_8-S - -
          mouse-6 v1.1 (Illumina) scl0117109.5_26-S - -
          Illumina Sentrix mouse-6.v1 bead array 1500110 - -
          Illumina Sentrix mouse-6.v1 bead array 5270619 - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl27367.4.1_8-S - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl0117109.5_26-S - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 1500110 - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 5270619 - -
          MouseRef-8 v2 Expression BeadChip ILMN_2973928 - -
          MouseRef-8 v2 Expression BeadChip ILMN_2973925 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_2973928 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_2973925 - -
          Illumina Sentrix Mouse-6 Expression BeadChip (3) scl27367.4.1_8-S - -
          Illumina Sentrix Mouse-6 Expression BeadChip (3) scl0117109.5_26-S - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-74594944d-pk6zc:80/100.66.130.62:80.
          git-commit: ab2b2261c2971e97b1ae408e3ad10520b8413932
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.31.0-2025.06.23-1.0