Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm01203254_g1
          See other MRPS18C GE Assays ›
          Gene Symbol:
          Mrps18c
          Gene Name:
          mitochondrial ribosomal protein S18C
          Gene Aliases:
          1110037D14Rik
          Chromosome Location:
          Chr.5: 100798759 - 100804467 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm01203254_g1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Mrps18c
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_026826.1 NP_081102.1 5 - 6 365 121
          GenBank mRNA AK004139.1 - 5 - 6 378 121
          AK145935.1 - 5 - 6 382 121
          BC028442.1 - 5 - 6 365 1196057 121

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC0500000985.mm.1

          Set Membership:

          Ambion siRNA available Amplicon length greater than or equal to 101 3' Most Mammalian Gene Collection Detect Genomic DNA Recommended Assay Mus musculus Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Mrps18c

          Gene Ontology Categories:

          Function(s) Process(es)

          translation
          structural constituent of ribosome

          MicroArray Probes:

          Agilent Affymetrix Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P23931500 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P23931499 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P23931502 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P23931501 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P23931503 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P23931506 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P23931498 - -
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2027147 AK088290.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2027147 AK145935.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2027147 BC028442.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2027147 NM_026826.1 mouse_transcriptome
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 909154 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 313160 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 941849 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 822872 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P173458 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2027147 AK088290.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2027147 AK145935.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2027147 BC028442.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2027147 NM_026826.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P108484 AK088290.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P108484 AK145935.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P108484 BC028442.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P108484 NM_026826.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P108484 AK004139.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P108478 AK088290.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P108478 AK145935.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P108478 BC028442.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P108478 NM_026826.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P108478 AK004139.1 mouse_transcriptome
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23931495 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23931475 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23931486 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23931467 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23931484 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23931509 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23931526 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23931517 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23931496 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23931489 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23931471 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23931499 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23931493 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23931521 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23931531 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23931510 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23931492 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23931528 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23931468 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23931473 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23931501 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23931525 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23931476 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23931503 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23931534 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23931537 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23931507 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23931465 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23931513 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23931536 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23931494 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23931500 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23931523 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23931532 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23931483 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23931474 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23931478 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23931516 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23931497 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23931520 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23931522 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23931481 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23931487 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23931535 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P173458 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P05672215 - -
          Whole Mouse Genome G4122A A_51_P108484 AK088290.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P108484 AK145935.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P108484 BC028442.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P108484 NM_026826.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P108484 AK004139.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P108478 AK088290.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P108478 AK145935.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P108478 BC028442.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P108478 NM_026826.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P108478 AK004139.1 mouse_transcriptome
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P173458 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P04485893 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P04931667 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P23931499 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P23931500 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P23931503 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P23931502 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P23931498 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P23931506 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P23931501 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23931528 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23931531 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23931537 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23931536 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23931584 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23931510 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23931581 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23931595 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23931576 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23931590 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23931516 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23931583 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23931517 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23931523 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23931520 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23931518 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23931513 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23931503 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23931535 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23931525 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23931507 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23931587 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23931589 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23931532 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23931522 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23931578 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23931511 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23931534 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23931509 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23931540 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23931521 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23931528 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23931532 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P04539645 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23931522 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23931521 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P04485891 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23931516 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23931537 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P04459781 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P04844475 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23931508 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P04931667 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23931531 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23931538 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23931520 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23931536 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P04539644 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23931535 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23931509 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23931525 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23931511 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23931503 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23931526 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23931513 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23931518 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23931517 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10523542 - -
          Affymetrix Mouse Gene 1.0 ST Array 10523545 - -
          Affymetrix Mouse Gene 1.0 ST Array 10523544 - -
          Affymetrix Mouse Gene 1.0 ST Array 10523546 - -
          Affymetrix Mouse Gene 1.0 ST Array 10523543 - -
          Affymetrix HT MG-430 PM Array Plate 1427901_PM_at - -
          Affymetrix Mouse Gene 1.1 ST Array 10523541 - -
          Mouse Genome 430 2.0 Array 1445269_PM_at - -
          Mouse Genome 430 2.0 Array 1427901_PM_at - -
          Mouse Genome 430 2.0 Array 1427901_at - -
          Mouse Genome 430 2.0 Array 1445269_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 68735_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 68735_PM_at - -
          Affymetrix Mouse Clariom S Array PSR0500008321.mm.1 TC0500000985.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0500008321.mm.1 NM_026826.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0500008321.mm.1 NM_026826.1 Refseq
          Mouse Expression Set 430 1427901_at - -
          Mouse Expression Set 430 1427901_PM_at - -
          Mouse Genome 430A 2.0 Array 1427901_PM_at - -
          Mouse Genome 430A 2.0 Array 1427901_at - -
          Affymetrix HT Mouse Genome 430A Array 1427901_PM_at - -
          Affymetrix HT Mouse Genome 430A Array 1427901_at - -
          Array Name Probe IDs Targets Target Type
          MouseWG-6 v2 Expression BeadChip 102940373 - -
          MouseWG-6 v2 Expression BeadChip 6760044 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_2747345 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_2747345 - -
          MouseRef-8 v2 Expression BeadChip ILMN_2747345 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-97865cc7b-x8sd2:80/100.66.134.75:80.
          git-commit: d53bc399386730d6c5c6d01fe94b21e0ad3fdbfa
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.33.0-2025.07.30-1.0