Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Clariom S Array |
PSR0500008158.mm.1 |
TC0500000949.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0500008158.mm.1 |
NM_001024614.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0500008158.mm.1 |
NM_001024614.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0500008164.mm.1 |
TC0500000949.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0500008164.mm.1 |
NM_001024614.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0500008164.mm.1 |
NM_001024614.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0500008160.mm.1 |
TC0500000949.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0500008160.mm.1 |
NM_001024614.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0500008160.mm.1 |
NM_001024614.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0500008161.mm.1 |
TC0500000949.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0500008161.mm.1 |
NM_001024614.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0500008161.mm.1 |
NM_001024614.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0500008163.mm.1 |
TC0500000949.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0500008163.mm.1 |
NM_001024614.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0500008163.mm.1 |
NM_001024614.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0500008159.mm.1 |
TC0500000949.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0500008159.mm.1 |
NM_001024614.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0500008159.mm.1 |
NM_001024614.1 |
Refseq |