Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm01203737_m1
          See other 1700095B10RIK GE Assays ›
          Gene Symbol:
          1700095B10Rik
          Gene Name:
          RIKEN cDNA 1700095B10 gene
          Gene Aliases:
          -
          Chromosome Location:
          Chr.5: 112793292 - 112801701 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm01203737_m1
          Size
          Availability Made To Order
          Catalog # 4426961
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: 1700095B10Rik
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NR_040675.1 3 - 4 637 62
          NR_040676.1 3 - 4 540 62
          GenBank mRNA AK006812.1 - 3 - 4 620 62
          AK007075.1 - 3 - 4 543 62
          AK015608.1 - 3 - 4 541 62
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Transcript Location Amplicon Length
          GenBankmRNA BY706794.1 - 3 - 4 620 Chr. 5 - 112793309 - 112801369 [+] on Build GRCm38 62
          GenBankmRNA BY707145.1 - 3 - 4 541 Chr. 5 - 112793292 - 112801417 [+] on Build GRCm38 62

          Adjacent Gene Details

          Gene name Gene Symbol Alias Distance Relative Direction Location Chromsome
          Upstream predicted gene,
          30529
          Gm30529 -118.3kb Sense Chr. 5 - 112643830 - 112680538 on NCBI Build GRCm38
          Downstream adrenergic receptor kinase,
          beta 2
          Adrbk2 4833444A01Rik,
          AI851927,
          AW551196,
          Adrbk-2,
          Bark-2,
          GRK3
          108.8kb Anti-sense Chr. 5 - 112910478 - 113015951 on NCBI Build GRCm38

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC0500001187.mm.1

          Set Membership:

          Amplicon length less than or equal to 70 3' Most Mus musculus 1700 Probe spans exons

          MicroArray Probes:

          Agilent Affymetrix ABI

          Array Name Probe IDs Targets Target Type
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P104057 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P104057 - -
          Whole Mouse Genome G4122A A_52_P364998 AK006812.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P364998 AK007075.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P364998 AK015608.1 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P120823 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P104057 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23994345 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23994287 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P04307239 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23994292 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23994389 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23994369 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23994405 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P04945762 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23994302 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23994318 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23994269 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P04945763 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23994396 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23994274 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P04627986 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P04150194 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23994374 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23994421 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23994371 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P04877941 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P04113961 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P04889010 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P04507655 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23994305 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P04521553 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23994289 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P04507652 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23994340 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23994330 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23994416 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23994368 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P04627992 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23994347 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23994376 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23994378 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P04888985 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23994411 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23994382 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P04991592 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P04142068 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23994415 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23994386 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P04286136 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23994420 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23994402 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23994271 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P04888994 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P04991587 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P04627987 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P04888982 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P04279533 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P04991596 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23994339 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23994392 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P04734389 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23994294 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23994380 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23994393 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P04627991 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P04888997 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23994356 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23994291 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23994366 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P04279532 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23994365 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P04991588 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P04627983 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P04997271 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P04127510 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23994412 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23994342 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P04170048 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23994309 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P04673487 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P04521804 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23994262 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23994351 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P04127509 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23994300 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23994320 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23994325 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23994286 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23994307 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23994283 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23994304 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23994293 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P04945759 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P04889005 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23994332 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P04127513 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P04160574 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23994285 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P04142061 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23994265 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P04142064 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23994312 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23994414 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Clariom S Array PSR0500010157.mm.1 TC0500001187.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0500010157.mm.1 NR_040675.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0500010157.mm.1 NR_040675.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0500010157.mm.1 NR_040676.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0500010157.mm.1 NR_040676.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0500010156.mm.1 TC0500001187.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0500010156.mm.1 NR_040675.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0500010156.mm.1 NR_040675.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0500010156.mm.1 NR_040676.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0500010156.mm.1 NR_040676.1 Refseq
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 321424 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-7b7b59cd5b-z8v7b:80/100.66.130.62:80.
          git-commit: 87deb889a5bb48f859ebd9970e87be562fd9396c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.32.0-2025.07.28-1.0