Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm01204118_m1
          See other B930018H19RIK GE Assays ›
          Gene Symbol:
          B930018H19Rik
          Gene Name:
          RIKEN cDNA B930018H19 gene
          Gene Aliases:
          B930018H19
          Chromosome Location:
          Chr.8: 34589420 - 34640316 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm01204118_m1
          Size
          Availability Made To Order
          Catalog # 4426961
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: B930018H19Rik
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NR_040706.1 4 - 5 955 58
          GenBank mRNA AK047086.1 - 4 - 5 956 58

          Adjacent Gene Details

          Gene name Gene Symbol Alias Distance Relative Direction Location Chromsome
          Upstream predicted gene,
          33968
          Gm33968 -64.0kb Anti-sense Chr. 8 - 34520474 - 34525431 on NCBI Build GRCm38
          Downstream dual specificity phosphatase 4 Dusp4 2700078F24Rik,
          AI844617,
          BB104621,
          E130306H24Rik,
          MKP2
          215.7kb Anti-sense Chr. 8 - 34807610 - 34819894 on NCBI Build GRCm38

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC0800002033.mm.1

          Set Membership:

          Amplicon length less than or equal to 70 3' Most Mus musculus 1700 Probe spans exons

          MicroArray Probes:

          Agilent Affymetrix ABI

          Array Name Probe IDs Targets Target Type
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2222904 AK047086.1 mouse_transcriptome
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 212831 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 950083 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 312778 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 31500 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 402203 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 120490 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 232940 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 832122 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 944466 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 375939 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 673478 AK021043.1 genbank_mrna_mouse
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 19597 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 39217 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 26177 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 20639 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 15920 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P06474283 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P06474319 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P06474356 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P06474376 - -
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P126377 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_67_P06474319 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25774960 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06786961 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25774957 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25775152 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25775148 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25775146 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25774956 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25774948 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25774962 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25774964 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25775147 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25775157 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25775159 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25775151 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25774955 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25775163 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06816512 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06970963 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25775164 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25775161 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25775155 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06785234 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25774950 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Clariom S Array PSR0800015872.mm.1 TC0800002033.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0800015872.mm.1 NR_040706.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0800015872.mm.1 NR_040706.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0800015873.mm.1 TC0800002033.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0800015873.mm.1 NR_040706.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0800015873.mm.1 NR_040706.1 Refseq
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 438848 - -
          ABI Mouse Genome Survey Microarray 517306 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-6467d75d44-xp7n2:80/100.66.130.62:80.
          git-commit: 8ea1bebb48d33b8b7c0e2103582aa482e6db8664
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.32.0-2025.07.28-1.0