Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm01205939_m1
          See other CRYBA4 GE Assays ›
          Gene Symbol:
          Cryba4
          Gene Name:
          crystallin, beta A4
          Gene Aliases:
          -
          Chromosome Location:
          Chr.5: 112246492 - 112263555 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm01205939_m1
          Size
          Availability Made To Order
          Catalog # 4351372
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Cryba4
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_021351.1 NP_067326.1 4 - 5 461 70
          GenBank mRNA AJ272228.2 - 4 - 5 461 70
          AK132591.1 - 5 - 6 478 70
          AK162018.1 - 5 - 6 475 70
          BC056444.1 - 4 - 5 451 6824277 70
          BC058703.1 - 4 - 5 461 6824293 70

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC0500003013.mm.1

          Set Membership:

          Ambion siRNA available Amplicon length less than or equal to 70 3' Most Mammalian Gene Collection Mus musculus 1700 Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Cryba4

          Gene Ontology Categories:

          Function(s) Process(es)

          visual perception
          camera-type eye development
          structural constituent of eye lens

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P23990792 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P23990791 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P23990793 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P23990396 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P23990799 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P23990790 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P23990800 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P23990398 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P23990397 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P23990803 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P23990789 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P23990802 - -
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2027831 BC058703.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2027831 AK161601.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2027831 BC056444.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2000034 BC058703.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2000034 NM_021351.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2000034 AJ272228.2 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2000034 AK132591.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2000034 AK161601.1 mouse_transcriptome
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 742520 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 436425 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 843572 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 120373 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2000034 BC058703.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2000034 NM_021351.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2000034 AJ272228.2 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2000034 AK132591.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2000034 AK161601.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2027831 BC058703.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2027831 AK161601.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2027831 BC056444.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P285299 BC058703.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P285299 NM_021351.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P285299 AJ272228.2 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P285299 AK132591.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P285299 AK161601.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P285299 BC058703.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P285299 NM_021351.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P285299 AJ272228.2 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P285299 AK132591.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P285299 AK161601.1 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P134000 - -
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P117335 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P23990800 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P23990792 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P23990793 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P23990791 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P23990799 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P23990396 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P23990803 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P23990789 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P23990397 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P23990398 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P23990802 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P23990790 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23990863 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23990848 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23990869 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23990877 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23990900 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23990882 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23990893 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23990895 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23990883 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23990861 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23990857 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23990868 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23990846 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23990888 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23990859 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23990885 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23990866 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23990854 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23990870 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23990871 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23990879 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23990892 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23990844 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23990851 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23990898 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23990893 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P04541062 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23990845 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23990888 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P04541060 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23990844 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23990890 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23990900 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23990848 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23990892 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23990883 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23990891 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23990851 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23990882 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23990895 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23990854 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P04703486 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23990885 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23990905 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23990886 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23990856 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P04983918 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23990866 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23990863 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23990846 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10532520 - -
          Affymetrix Mouse Gene 1.0 ST Array 10532521 - -
          Affymetrix Mouse Gene 1.0 ST Array 10532522 - -
          Affymetrix Mouse Gene 1.0 ST Array 10532519 - -
          Affymetrix Mouse Gene 1.0 ST Array 10532523 - -
          Affymetrix Mouse Gene 1.0 ST Array 10532518 - -
          Affymetrix HT MG-430 PM Array Plate 1420686_PM_at - -
          Affymetrix Mouse Gene 1.1 ST Array 10532517 - -
          Mouse Genome 430 2.0 Array 1420686_PM_at - -
          Mouse Genome 430 2.0 Array 1420686_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 12959_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 12959_PM_at - -
          Affymetrix Mouse Clariom S Array PSR0500026736.mm.1 TC0500003013.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0500026736.mm.1 NM_021351.2 Refseq
          Affymetrix Mouse Clariom S Array PSR0500026735.mm.1 TC0500003013.mm.1 MTA TC
          Mouse Expression Set 430 1420686_PM_at - -
          Mouse Expression Set 430 1420686_at - -
          Mouse Genome 430A 2.0 Array 1420686_PM_at - -
          Mouse Genome 430A 2.0 Array 1420686_at - -
          Affymetrix HT Mouse Genome 430A Array 1420686_at - -
          Affymetrix HT Mouse Genome 430A Array 1420686_PM_at - -
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 480260 - -
          Array Name Probe IDs Targets Target Type
          Illumina mouse-6 v1.0 expression beadchip 6760603 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0006760603 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_1258732 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_1258732 - -
          mouse-6 v1.1 (Illumina) scl44577.6.1_56-S - -
          Illumina Sentrix mouse-6.v1 bead array 6760603 - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl44577.6.1_56-S - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 6760603 - -
          MouseRef-8 v2 Expression BeadChip ILMN_1258732 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_1258732 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-65cb46898d-nlgdw:80/100.66.128.131:80.
          git-commit: e5493f66f4597fc66e516c8a379ca3b6bed48f5b
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.33.0-2025.07.30-1.0