Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm01208301_g1
          See other TMEM132A GE Assays ›
          Gene Symbol:
          Tmem132a
          Gene Name:
          transmembrane protein 132A
          Gene Aliases:
          6720481D13Rik, Hspa5bp1, Orai1, R74613
          Chromosome Location:
          Chr.19: 10857829 - 10869935 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm01208301_g1
          Size
          Availability Made To Order
          Catalog # 4351372
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Tmem132a
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_133804.2 NP_598565.2 10 - 11 2169 76
          GenBank mRNA AK032956.1 - 10 - 11 2170 76
          AK048556.1 - 10 - 11 2170 76
          AK164646.1 - 10 - 11 2171 76
          AK173207.1 - 10 - 11 2157 76
          BC006896.1 - 10 - 11 2121 3599652 76

          Back To Top

          More Information


          Set Membership:

          Ambion siRNA available Amplicon length between 71 and 85 3' Most Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Tmem132a

          Gene Ontology Categories:

          Function(s) Process(es)

          negative regulation of programmed cell death
          molecular_function

          MicroArray Probes:

          Agilent Affymetrix Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P31953350 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P31953349 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P31953353 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P31953351 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P31953348 - -
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2113081 BC006896.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2113081 CF729397.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2113081 AK173207.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2113081 AK196272.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2113081 AK164646.1 mouse_transcriptome
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 386796 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 442921 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 76053 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 162215 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 13713 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 147252 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P141722 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P123533 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2113081 BC006896.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2113081 CF729397.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2113081 AK173207.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2113081 AK196272.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2113081 AK164646.1 mouse_transcriptome
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31953360 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31953339 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31953356 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31953335 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31953336 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31953357 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31953345 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31953343 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31953340 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31953344 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31953353 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31953341 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31953350 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31953337 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31953355 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31953346 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31953334 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31953347 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P03549788 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P141722 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P123533 - -
          Whole Mouse Genome G4122A A_51_P169281 BC006896.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P169281 CF729397.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P169281 AK173207.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P169281 AK048556.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P169281 AK164646.1 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P101641 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P123533 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P141722 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P14198550 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P14198544 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P14198543 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P14198529 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P14198540 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P14198535 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P14198551 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P31953349 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P31953348 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P31953353 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P31953350 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P31953351 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31953235 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31953299 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31953326 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31953401 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31953307 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31953270 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31953291 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31953233 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31953330 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31953260 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31953221 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31953305 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31953281 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31953360 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31953340 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31953216 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31953224 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31953206 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31953237 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31953205 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31953301 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31953334 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31953337 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31953353 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31953199 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31953271 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31953304 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31953339 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31953332 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31953403 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31953418 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31953201 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31953289 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31953341 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31953335 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31953196 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31953345 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31953230 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31953315 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31953274 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31953402 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31953284 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31953238 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31953347 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31953398 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31953306 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31953313 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31953355 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31953328 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31953227 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31953278 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31953412 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31953297 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31953319 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31953213 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31953293 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31953309 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31953408 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31953417 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31953296 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31953308 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31953322 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31953266 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31953323 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31953214 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31953239 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31953318 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31953267 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31953218 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31953343 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31953399 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31953303 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31953406 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31953280 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31953336 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31953415 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31953265 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31953357 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31953350 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31953261 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P14320149 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P14191541 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31953309 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31953350 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P14191539 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P14198529 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P14198906 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P14320147 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31953307 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P14453114 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31953347 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31953337 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31953343 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P14198542 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31953301 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31953306 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31953345 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31953305 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31953355 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31953318 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31953330 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31953328 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P14453120 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31953332 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P14320145 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31953360 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31953313 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31953357 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P14269402 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31953298 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31953315 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31953339 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P14215970 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31953296 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31953323 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31953322 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31953353 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31953303 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31953299 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31953308 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10466076 - -
          Affymetrix Mouse Gene 1.0 ST Array 10466084 - -
          Affymetrix Mouse Gene 1.0 ST Array 10466086 - -
          Affymetrix Mouse Gene 1.0 ST Array 10466080 - -
          Affymetrix Mouse Gene 1.0 ST Array 10466081 - -
          Affymetrix Mouse Gene 1.0 ST Array 10466083 - -
          Affymetrix Mouse Gene 1.0 ST Array 10466085 - -
          Affymetrix Mouse Gene 1.0 ST Array 10466078 - -
          Affymetrix Mouse Gene 1.0 ST Array 10466082 - -
          Affymetrix Mouse Gene 1.0 ST Array 10466079 - -
          Affymetrix Mouse Gene 1.0 ST Array 10466077 - -
          Affymetrix HT MG-430 PM Array Plate 1416845_PM_at - -
          Affymetrix Mouse Gene 1.1 ST Array 10466075 - -
          Mouse Genome 430 2.0 Array 1416845_PM_at - -
          Mouse Genome 430 2.0 Array 1416845_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 98170_PM_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 98170_at - -
          Mouse Expression Set 430 1416845_PM_at - -
          Mouse Expression Set 430 1416845_at - -
          Mouse Genome 430A 2.0 Array 1416845_at - -
          Mouse Genome 430A 2.0 Array 1416845_PM_at - -
          Affymetrix HT Mouse Genome 430A Array 1416845_at - -
          Affymetrix HT Mouse Genome 430A Array 1416845_PM_at - -
          Array Name Probe IDs Targets Target Type
          MouseWG-6 v2 Expression BeadChip 510433 - -
          Illumina mouse-6 v1.0 expression beadchip 510433 - -
          Illumina mouse-6 v1.0 expression beadchip 770711 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0000510433 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0000770711 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_2750588 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_1226606 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_2707798 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_1226606 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_2707798 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_2750588 - -
          mouse-6 v1.1 (Illumina) scl000043.1_1-S - -
          mouse-6 v1.1 (Illumina) scl52742.10_14-S - -
          Illumina Sentrix mouse-6.v1 bead array 770711 - -
          Illumina Sentrix mouse-6.v1 bead array 510433 - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl52742.10_14-S - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl000043.1_1-S - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 770711 - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 510433 - -
          MouseRef-8 v2 Expression BeadChip ILMN_1226606 - -
          MouseRef-8 v2 Expression BeadChip ILMN_2750588 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_2750588 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_1226606 - -
          Illumina Sentrix Mouse-6 Expression BeadChip (3) scl52742.10_14-S - -
          Illumina Sentrix Mouse-6 Expression BeadChip (3) scl000043.1_1-S - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-74594944d-pk6zc:80/100.66.130.62:80.
          git-commit: ab2b2261c2971e97b1ae408e3ad10520b8413932
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.31.0-2025.06.23-1.0