Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm01208563_g1
          See other TOPORSOS GE Assays ›
          Gene Symbol:
          Toporsos
          Gene Name:
          topoisomerase I binding, arginine/serine-rich, opposite strand
          Gene Aliases:
          2010003O02Rik
          Chromosome Location:
          Chr.4: 40269579 - 40270221 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm01208563_g1
          Size
          Availability Made To Order
          Catalog # 4426961
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Toporsos
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NR_045265.1 1 - 2 144 59
          GenBank mRNA AA614943.1 - 1 - 2 143 59
          AK008077.1 - 142 59
          AK019894.1 - 1 - 2 144 59
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Transcript Location Amplicon Length
          GenBankmRNA AA290355.1 - 1 - 2 141 Chr. 4 - 40269590 - 40270194 [+] on Build GRCm38 59
          GenBankmRNA W91647.1 - 1 - 2 125 Chr. 4 - 40269598 - 40270221 [+] on Build GRCm38 59
          GenBankmRNA BM508752.1 - 1 - 2 112 Chr. 4 - 40269611 - 40270224 [+] on Build GRCm38 59
          GenBankmRNA BY156097.1 - 1 - 2 143 Chr. 4 - 40269579 - 40270224 [+] on Build GRCm38 59
          GenBankmRNA BY716970.1 - 1 - 2 144 Chr. 4 - 40269579 - 40270224 [+] on Build GRCm38 59
          GenBankmRNA BB022034.1 - 1 - 2 115 Chr. 4 - 40269608 - 40270221 [+] on Build GRCm38 59
          GenBankmRNA AA190030.1 - 1 - 2 126 Chr. 4 - 40269597 - 40270228 [+] on Build GRCm38 59
          GenBankmRNA CR516257.1 - 1 - 2 Chr. 4 - 40269591 - 40270224 [-] on Build GRCm38 59
          GenBankmRNA W91651.1 - 1 - 2 125 Chr. 4 - 40269598 - 40270221 [+] on Build GRCm38 59
          GenBankmRNA BM508444.1 - 1 - 2 Chr. 4 - 40269641 - 40270221 [-] on Build GRCm38 59
          GenBankmRNA AV460479.1 - 1 - 2 131 Chr. 4 - 40269591 - 40270224 [+] on Build GRCm38 59
          GenBankmRNA CA461880.1 - 181 Chr. - - [-] on Build 59
          GenBankmRNA DT907618.1 - 1 - 2 141 Chr. 4 - 40269590 - 40270194 [+] on Build GRCm38 59
          GenBankmRNA W91372.1 - 1 - 2 137 Chr. 4 - 40269586 - 40270222 [+] on Build GRCm38 59
          GenBankmRNA AA510458.1 - 1 - 2 104 Chr. 4 - 40269619 - 40270217 [+] on Build GRCm38 59
          GenBankmRNA AA185915.1 - 1 - 2 119 Chr. 4 - 40269601 - 40270217 [+] on Build GRCm38 59
          GenBankmRNA CB318174.1 - 142 Chr. - - [-] on Build 59
          GenBankmRNA BU562161.1 - 157 Chr. - - [-] on Build 59
          GenBankmRNA W53999.1 - 126 Chr. - - [-] on Build 59

          Adjacent Gene Details

          Gene name Gene Symbol Alias Distance Relative Direction Location Chromsome
          Upstream DEAD (Asp-Glu-Ala-Asp) box polypeptide 58 Ddx58 6430573D20Rik,
          C330021E21,
          RIG-I,
          RLR-1
          -29.8kb Anti-sense Chr. 4 - 40203777 - 40239825 on NCBI Build GRCm38
          Downstream NADH dehydrogenase (ubiquinone) 1 beta subcomplex,
          6
          Ndufb6 Gm137 442.0 Anti-sense Chr. 4 - 40270663 - 40279368 on NCBI Build GRCm38

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC0400000344.mm.1

          Additional Information:

          This assay is in the 5'UTR region of the AK008077.1 transcripts.

          Set Membership:

          Amplicon length less than or equal to 70 3' Most Detect Genomic DNA Mus musculus Probe spans exons

          MicroArray Probes:

          Affymetrix

          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Clariom S Array PSR0400002099.mm.1 TC0400000344.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0400002099.mm.1 NR_045265.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0400002099.mm.1 NR_045265.1 Refseq

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-97865cc7b-9fgnx:80/100.66.128.131:80.
          git-commit: d53bc399386730d6c5c6d01fe94b21e0ad3fdbfa
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.33.0-2025.07.30-1.0