Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm01209070_m1
          See other ANKRD10 GE Assays ›
          Gene Symbol:
          Ankrd10
          Gene Name:
          ankyrin repeat domain 10
          Gene Aliases:
          4833425P12Rik, AW549277
          Chromosome Location:
          Chr.8: 11611581 - 11636136 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm01209070_m1
          Size
          Availability Made To Order
          Catalog # 4351372
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Ankrd10
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001167967.2 NP_001161439.1 4 - 5 834 84
          NM_001281974.1 NP_001268903.1 5 - 6 946 84
          NM_001281975.1 NP_001268904.1 5 - 6 996 84
          NM_133971.3 NP_598732.2 4 - 5 834 84
          NR_030781.2 5 - 6 825 84
          GenBank mRNA AK014768.1 - 5 - 6 825 84
          AK031808.1 - 4 - 5 832 84
          AK033623.1 - 4 - 5 832 84
          AK038156.1 - 4 - 5 832 84
          AK039879.1 - 2 - 3 279 84
          AK042500.1 - 4 - 5 832 84
          AK050983.1 - 4 - 5 835 84
          AK081754.1 - 5 - 6 994 84
          AK082875.1 - 4 - 5 831 84
          AK084456.1 - 5 - 6 944 84
          AK086012.1 - 2 - 3 1189 84
          AK088371.1 - 4 - 5 784 84
          BC002198.1 - 4 - 5 766 3489133 84
          BC033266.1 - 1 - 2 129 84

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC0800001701.mm.1

          Set Membership:

          Ambion siRNA available Amplicon length between 71 and 85 3' Most Mammalian Gene Collection Mus musculus Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Ankrd10

          Gene Ontology Categories:

          Function(s) Process(es)

          regulation of canonical Wnt signaling pathway
          molecular_function

          MicroArray Probes:

          Affymetrix

          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Clariom S Array PSR0800013818.mm.1 TC0800001701.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0800013818.mm.1 NM_001167967.2 Refseq
          Affymetrix Mouse Clariom S Array PSR0800013818.mm.1 NM_001281974.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0800013818.mm.1 NM_001281975.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0800013818.mm.1 NM_133971.3 Refseq
          Affymetrix Mouse Clariom S Array PSR0800013818.mm.1 NR_030781.2 Refseq
          Affymetrix Mouse Clariom S Array PSR0800013814.mm.1 TC0800001701.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0800013814.mm.1 NM_001167967.2 Refseq
          Affymetrix Mouse Clariom S Array PSR0800013814.mm.1 NR_030781.2 Refseq
          Affymetrix Mouse Clariom S Array PSR0800013817.mm.1 TC0800001701.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0800013817.mm.1 NM_001167967.2 Refseq
          Affymetrix Mouse Clariom S Array PSR0800013817.mm.1 NR_030781.2 Refseq

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-74594944d-pk6zc:80/100.66.130.62:80.
          git-commit: ab2b2261c2971e97b1ae408e3ad10520b8413932
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.31.0-2025.06.23-1.0