Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm01209897_m1
          See other 4932443I19RIK GE Assays ›
          Gene Symbol:
          4932443I19Rik
          Gene Name:
          RIKEN cDNA 4932443I19 gene
          Gene Aliases:
          A230061C02Rik, Gm1106
          Chromosome Location:
          Chr.8: 13705449 - 13743531 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm01209897_m1
          Size
          Availability Made To Order
          Catalog # 4351372
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: 4932443I19Rik
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001101519.1 NP_001094989.1 2 - 3 246 84
          GenBank mRNA AK030127.1 - 2 - 3 1450 84
          AK038767.1 - 2 - 3 248 84

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC0800000129.mm.1

          Set Membership:

          Amplicon length between 71 and 85 3' Most Mus musculus Probe spans exons

          Gene Ontology Categories:

          Function(s) Process(es)

          biological_process
          molecular_function

          MicroArray Probes:

          Agilent Affymetrix Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P1957209 AK038767.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P1957209 NM_001101519.1 mouse_transcriptome
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 336550 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 395318 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 8455 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 672994 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 176389 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 542045 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 96887 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 933413 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 939899 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 932875 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 307605 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 129775 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 593196 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 269746 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 561423 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 276614 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 127702 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 801177 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 28065 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 147842 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 165455 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 55019 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 119716 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 111785 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P139273 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P1957209 AK038767.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P1957209 NM_001101519.1 mouse_transcriptome
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25670243 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25670320 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25670189 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25670195 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25670085 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25670203 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25670188 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25670163 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25670198 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25670245 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25670088 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25670161 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25670316 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25670126 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25670164 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25670107 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25670226 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25670237 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25670082 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25670090 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25670294 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25670213 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25670282 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25670289 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25670121 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25670292 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25670186 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25670276 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25670192 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25670141 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25670231 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25670221 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25670210 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25670151 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25670248 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25670260 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25670256 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25670128 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25670074 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25670204 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25670327 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25670182 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25670147 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25670113 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25670129 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25670235 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25670283 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25670259 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25670269 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25670134 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25670140 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25670131 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25670132 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25670323 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25670262 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25670325 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25670239 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25670244 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25670274 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25670267 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25670180 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25670137 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25670266 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25670078 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25670144 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25670208 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25670115 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25670242 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25670093 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25670217 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25670076 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25670166 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25670111 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25670264 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25670097 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25670173 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25670154 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25670270 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25670212 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25670334 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25670218 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25670099 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25670081 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25670109 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25670156 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25670084 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25670123 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25670159 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25670087 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25670254 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25670272 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25670337 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25670193 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25670130 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25670153 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25670214 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25670285 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25670117 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25670319 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25670162 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25670223 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25670105 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25670257 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25670103 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25670332 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25670336 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25670155 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25670177 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25670220 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25670240 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25670171 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25670164 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25670161 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25670144 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25670103 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06676350 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25670119 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25670112 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25670155 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25670134 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25670153 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25670163 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25670130 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25670113 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25670141 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25670099 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25670128 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06676356 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06676358 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25670107 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25670147 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25670140 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25670137 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25670108 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25670156 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25670115 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25670111 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25670159 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25670131 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25670166 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06957485 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25670143 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25670097 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P07262538 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25670129 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25670151 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06957493 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P07262536 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P07081311 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P07261769 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P07262541 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25670126 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25670096 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25670132 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Clariom S Array PSR0800001208.mm.1 TC0800000129.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0800001208.mm.1 NM_001101519.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0800001208.mm.1 NM_001101519.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0800001210.mm.1 TC0800000129.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0800001210.mm.1 NM_001101519.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0800001210.mm.1 NM_001101519.1 Refseq
          Array Name Probe IDs Targets Target Type
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_2627818 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_1218088 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_2627818 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-74594944d-pk6zc:80/100.66.130.62:80.
          git-commit: ab2b2261c2971e97b1ae408e3ad10520b8413932
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.31.0-2025.06.23-1.0