Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm01210382_m1
          See other TRPM3 GE Assays ›
          Gene Symbol:
          Trpm3
          Gene Name:
          transient receptor potential cation channel, subfamily M, member 3
          Gene Aliases:
          6330504P12Rik, 9330180E14, AU018608, B930001P07Rik, LTRPC3, MLSN2
          Chromosome Location:
          Chr.19: 22138181 - 22989897 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm01210382_m1
          Size
          Availability Made To Order
          Catalog # 4351372
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Trpm3
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001035239.2 NP_001030316.1 8 - 9 1275 70
          NM_001035240.2 NP_001030317.1 8 - 9 1275 70
          NM_001035241.2 NP_001030318.1 8 - 9 1275 70
          NM_001035242.1 NP_001030319.1 8 - 9 1275 70
          NM_001035243.2 NP_001030320.1 8 - 9 1275 70
          NM_001035244.1 NP_001030321.1 9 - 10 1398 70
          NM_177341.4 NP_796315.2 8 - 9 1342 70
          GenBank mRNA AJ544532.1 - 8 - 9 1275 70
          AJ544533.1 - 8 - 9 1275 70
          AJ544534.1 - 8 - 9 1275 70
          AJ544535.1 - 8 - 9 1275 70
          AJ544536.1 - 8 - 9 1275 70
          AK044454.1 - 8 - 9 1344 70
          AK046908.1 - 9 - 10 1231 70
          AK051867.1 - 2 - 3 298 70
          AK053673.1 - 8 - 9 1160 70
          AK142504.1 - 9 - 10 1231 70
          AK173218.1 - 8 - 9 1346 70
          BC141400.1 - 8 - 9 1318 9056013 70
          BC141412.1 - 8 - 9 1318 9056025 70
          HM064407.1 - 8 - 9 1367 70
          HM064408.1 - 8 - 9 1367 70
          HM064409.1 - 8 - 9 1367 70
          HM064410.1 - 9 - 10 1442 70
          HM064411.1 - 8 - 9 1367 70
          JF706722.1 - 8 - 9 1338 70
          JX644970.1 - 8 - 9 1367 70
          JX644971.1 - 8 - 9 1367 70
          JX644972.1 - 8 - 9 1367 70
          JX644973.1 - 8 - 9 1367 70
          JX644974.1 - 8 - 9 1367 70
          JX644975.1 - 9 - 10 1442 70
          JX644976.1 - 8 - 9 1367 70
          JX644977.1 - 8 - 9 1367 70
          JX644978.1 - 8 - 9 1367 70
          JX644979.1 - 9 - 10 1442 70
          JX644980.1 - 9 - 10 1442 70
          JX644981.1 - 9 - 10 1442 70
          JX644982.1 - 9 - 10 1442 70
          JX644983.1 - 8 - 9 1367 70
          JX644984.1 - 8 - 9 1367 70
          JX644985.1 - 8 - 9 1367 70
          JX644986.1 - 8 - 9 1367 70

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC1900000356.mm.1

          Additional Information:

          This assay is in the 5'UTR region of the AK051867.1 transcripts.

          Set Membership:

          Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Mus musculus Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Trpm3

          Gene Ontology Categories:

          Function(s) Process(es)

          cation transport
          cation transmembrane transport
          cation channel activity

          MicroArray Probes:

          Affymetrix

          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Clariom S Array PSR1900003507.mm.1 TC1900000356.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1900003507.mm.1 NM_001035239.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1900003507.mm.1 NM_001035239.2 Refseq
          Affymetrix Mouse Clariom S Array PSR1900003507.mm.1 NM_001035240.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1900003507.mm.1 NM_001035240.2 Refseq
          Affymetrix Mouse Clariom S Array PSR1900003507.mm.1 NM_001035241.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1900003507.mm.1 NM_001035241.2 Refseq
          Affymetrix Mouse Clariom S Array PSR1900003507.mm.1 NM_001035242.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1900003507.mm.1 NM_001035242.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1900003507.mm.1 NM_001035243.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1900003507.mm.1 NM_001035243.2 Refseq
          Affymetrix Mouse Clariom S Array PSR1900003507.mm.1 NM_001035244.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1900003507.mm.1 NM_001035244.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1900003507.mm.1 NM_177341.4.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1900003507.mm.1 NM_177341.4 Refseq
          Affymetrix Mouse Clariom S Array PSR1900003508.mm.1 TC1900000356.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1900003508.mm.1 NM_001035239.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1900003508.mm.1 NM_001035239.2 Refseq
          Affymetrix Mouse Clariom S Array PSR1900003508.mm.1 NM_001035240.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1900003508.mm.1 NM_001035240.2 Refseq
          Affymetrix Mouse Clariom S Array PSR1900003508.mm.1 NM_001035241.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1900003508.mm.1 NM_001035241.2 Refseq
          Affymetrix Mouse Clariom S Array PSR1900003508.mm.1 NM_001035242.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1900003508.mm.1 NM_001035242.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1900003508.mm.1 NM_001035243.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1900003508.mm.1 NM_001035243.2 Refseq
          Affymetrix Mouse Clariom S Array PSR1900003508.mm.1 NM_177341.4.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1900003508.mm.1 NM_177341.4 Refseq
          Affymetrix Mouse Clariom S Array PSR1900003506.mm.1 TC1900000356.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1900003506.mm.1 NM_001035239.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1900003506.mm.1 NM_001035239.2 Refseq
          Affymetrix Mouse Clariom S Array PSR1900003506.mm.1 NM_001035240.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1900003506.mm.1 NM_001035240.2 Refseq
          Affymetrix Mouse Clariom S Array PSR1900003506.mm.1 NM_001035241.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1900003506.mm.1 NM_001035241.2 Refseq
          Affymetrix Mouse Clariom S Array PSR1900003506.mm.1 NM_001035242.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1900003506.mm.1 NM_001035242.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1900003506.mm.1 NM_001035243.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1900003506.mm.1 NM_001035243.2 Refseq
          Affymetrix Mouse Clariom S Array PSR1900003506.mm.1 NM_001035244.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1900003506.mm.1 NM_001035244.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1900003506.mm.1 NM_177341.4.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1900003506.mm.1 NM_177341.4 Refseq
          Affymetrix Mouse Clariom S Array PSR1900003503.mm.1 TC1900000356.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1900003503.mm.1 NM_001035239.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1900003503.mm.1 NM_001035239.2 Refseq
          Affymetrix Mouse Clariom S Array PSR1900003503.mm.1 NM_001035240.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1900003503.mm.1 NM_001035240.2 Refseq
          Affymetrix Mouse Clariom S Array PSR1900003503.mm.1 NM_001035241.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1900003503.mm.1 NM_001035241.2 Refseq
          Affymetrix Mouse Clariom S Array PSR1900003503.mm.1 NM_001035242.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1900003503.mm.1 NM_001035242.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1900003503.mm.1 NM_001035243.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1900003503.mm.1 NM_001035243.2 Refseq
          Affymetrix Mouse Clariom S Array PSR1900003503.mm.1 NM_001035244.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1900003503.mm.1 NM_001035244.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1900003503.mm.1 NM_177341.4.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1900003503.mm.1 NM_177341.4 Refseq

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-f85dfd666-px2hc:80/100.66.130.62:80.
          git-commit: 2ca7b3ab9a2195be9b417413331e4e963d93e17c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.32.0-2025.07.28-1.0