Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm01210631_m1
          See other EIF2S3Y GE Assays ›
          Gene Symbol:
          Eif2s3y
          Gene Name:
          eukaryotic translation initiation factor 2, subunit 3, structural gene Y-linked
          Gene Aliases:
          Eif-2gy, Spy, Tfy
          Chromosome Location:
          Chr.Y: 1010578 - 1046811 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm01210631_m1
          Size
          Availability Made To Order
          Catalog # 4351372
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Eif2s3y
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_012011.1 NP_036141.1 9 - 10 1014 135
          GenBank mRNA AJ006584.1 - 9 - 10 1014 135
          AK009448.1 - 9 - 10 1019 135
          BC031450.1 - 7 - 8 2089 135
          BC043656.1 - 9 - 10 1014 5368023 135

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC0Y00000006.mm.1

          Set Membership:

          Ambion siRNA available Amplicon length greater than or equal to 101 3' Most Mammalian Gene Collection Mus musculus 1700 Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Eif2s3y

          Gene Ontology Categories:

          Function(s) Process(es)

          translation
          translational initiation
          nucleotide binding
          translation initiation factor activity
          GTPase activity
          GTP binding

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2011877 BC043656.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2011877 AJ006584.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2011877 AK009448.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2011877 NM_012011.1 mouse_transcriptome
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 189383 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 964123 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 302849 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 683565 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 528411 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 690402 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 860199 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 271054 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 556883 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 45969 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 840100 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 5267 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 495246 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 481844 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 10983 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 60765 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2011877 BC043656.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2011877 AJ006584.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2011877 AK009448.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2011877 NM_012011.1 mouse_transcriptome
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P32825376 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P32825357 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P32825349 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P32825334 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P32825373 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P32825342 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P32825379 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P32825371 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P32825375 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P32825344 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P32825327 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P32825353 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P32825346 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P32825366 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P32825356 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P32825348 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P32825329 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P32825364 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P32825328 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P32825337 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P32825350 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P32825351 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P32825345 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P32825369 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P32825363 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P32825338 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P32825340 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P32825332 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P32825359 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P32825330 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P32825361 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P32825368 - -
          Whole Mouse Genome G4122A A_51_P479715 BC043656.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P479715 AJ006584.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P479715 AK009448.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P479715 NM_012011.1 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P134012 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P15232210 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P15229350 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P15232215 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P15232218 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P15229351 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32825398 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32825460 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32825389 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32825376 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32825380 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32825413 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32825332 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32825386 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32825479 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32825359 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32825396 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32825410 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32825478 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32825352 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32825451 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32825468 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32825353 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32825433 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32825404 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32825329 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32825486 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32825417 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32825446 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32825453 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32825496 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32825368 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32825459 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32825483 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32825435 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32825373 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32825424 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32825364 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32825471 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32825474 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32825458 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32825369 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32825338 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32825427 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32825438 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32825441 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32825436 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32825494 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32825379 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32825425 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32825375 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32825361 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32825491 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32825346 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32825366 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32825420 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32825340 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32825391 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32825430 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32825334 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32825348 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32825408 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32825455 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32825498 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32825326 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32825464 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32825485 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32825487 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32825350 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32825399 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32825377 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32825344 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32825448 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32825457 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32825476 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32825422 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32825356 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32825351 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32825382 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32825415 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32825500 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32825480 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32825371 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32825330 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32825467 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32825394 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32825443 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10608013 - -
          Affymetrix Mouse Gene 1.0 ST Array 10608003 - -
          Affymetrix Mouse Gene 1.0 ST Array 10608002 - -
          Affymetrix Mouse Gene 1.0 ST Array 10608007 - -
          Affymetrix Mouse Gene 1.0 ST Array 10608008 - -
          Affymetrix Mouse Gene 1.0 ST Array 10608011 - -
          Affymetrix Mouse Gene 1.0 ST Array 10608006 - -
          Affymetrix Mouse Gene 1.0 ST Array 10608012 - -
          Affymetrix Mouse Gene 1.0 ST Array 10608010 - -
          Affymetrix Mouse Gene 1.0 ST Array 10608009 - -
          Affymetrix Mouse Gene 1.0 ST Array 10608014 - -
          Affymetrix Mouse Gene 1.0 ST Array 10608005 - -
          Affymetrix Mouse Gene 1.0 ST Array 10608004 - -
          Affymetrix HT MG-430 PM Array Plate 1417210_PM_at - -
          Affymetrix HT MG-430 PM Array Plate 1431642_PM_at - -
          Affymetrix Mouse Gene 1.1 ST Array 10608001 - -
          Mouse Genome 430 2.0 Array 1457945_PM_at - -
          Mouse Genome 430 2.0 Array 1431642_PM_at - -
          Mouse Genome 430 2.0 Array 1417210_PM_at - -
          Mouse Genome 430 2.0 Array 1457945_at - -
          Mouse Genome 430 2.0 Array 1431642_at - -
          Mouse Genome 430 2.0 Array 1417210_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 26908_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 26908_PM_at - -
          Affymetrix Mouse Clariom S Array PSR0Y00000098.mm.1 TC0Y00000006.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0Y00000098.mm.1 NM_012011.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0Y00000098.mm.1 NM_012011.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0Y00000100.mm.1 TC0Y00000006.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0Y00000100.mm.1 NM_012011.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0Y00000100.mm.1 NM_012011.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0Y00000099.mm.1 TC0Y00000006.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0Y00000099.mm.1 NM_012011.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0Y00000099.mm.1 NM_012011.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0Y00000096.mm.1 TC0Y00000006.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0Y00000096.mm.1 NM_012011.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0Y00000096.mm.1 NM_012011.1 Refseq
          Mouse Expression Set 430 1417210_PM_at - -
          Mouse Expression Set 430 1417210_at - -
          Mouse Expression Set 430 1431642_PM_at - -
          Mouse Expression Set 430 1431642_at - -
          Murine Genome U74v2 Set 103674_PM_f_at - -
          Murine Genome U74v2 Set 103674_f_at - -
          Mouse Genome 430A 2.0 Array 1417210_at - -
          Mouse Genome 430A 2.0 Array 1417210_PM_at - -
          Affymetrix HT Mouse Genome 430A Array 1417210_at - -
          Affymetrix HT Mouse Genome 430A Array 1417210_PM_at - -
          Affymetrix HT Mouse Genome 430B Array 1431642_PM_at - -
          Affymetrix HT Mouse Genome 430B Array 1431642_at - -
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 931120 - -
          Array Name Probe IDs Targets Target Type
          Illumina mouse-6 v1.0 expression beadchip 6100707 - -
          Illumina mouse-6 v1.0 expression beadchip 102510025 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0006100707 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_1241001 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_1241001 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_1215914 - -
          mouse-6 v1.1 (Illumina) scl0026908.1_233-S - -
          Illumina Sentrix mouse-6.v1 bead array 102510025 - -
          Illumina Sentrix mouse-6.v1 bead array 6100707 - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl0026908.1_233-S - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 6100707 - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 102510025 - -
          MouseRef-8 v2 Expression BeadChip ILMN_1241001 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_1241001 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_1215914 - -
          Illumina Sentrix Mouse-6 Expression BeadChip (3) scl0026908.1_233-S - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-74594944d-pk6zc:80/100.66.130.62:80.
          git-commit: ab2b2261c2971e97b1ae408e3ad10520b8413932
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.31.0-2025.06.23-1.0