Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm01210795_m1
          See other CIB4 GE Assays ›
          Gene Symbol:
          Cib4
          Gene Name:
          calcium and integrin binding family member 4
          Gene Aliases:
          1700041E20Rik
          Chromosome Location:
          Chr.5: 30485583 - 30545837 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm01210795_m1
          Size
          Availability Made To Order
          Catalog # 4351372
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Cib4
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_028483.1 NP_082759.1 3 - 4 243 76
          GenBank mRNA AK006670.1 - 3 - 4 244 76
          BC099558.1 - 3 - 4 189 76
          BC118938.1 - 3 - 4 208 40091772 76

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC0500002158.mm.1

          Set Membership:

          Amplicon length between 71 and 85 3' Most Mammalian Gene Collection Mus musculus 1700 Probe spans exons

          Gene Ontology Categories:

          Function(s) Process(es)

          biological_process
          calcium ion binding
          metal ion binding

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P426373 BC099558.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P426373 NM_028483.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P426373 AK006670.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P426373 BC099558.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P426373 NM_028483.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P426373 AK006670.1 mouse_transcriptome
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 529513 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 134003 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 315522 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 772636 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 474072 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 686911 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 223656 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 732711 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 847612 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 229720 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 560479 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 169958 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 643510 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 517828 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 616288 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 470173 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 212868 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 898759 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 70132 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 733656 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 376049 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 498671 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 568509 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 3114 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 939396 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 768752 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 197694 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 107577 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 738728 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 173069 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 168835 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 679569 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 769789 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 264315 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 352159 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 200432 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 797621 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 87168 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 15441 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 85473 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 175392 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 142945 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 69389 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P155413 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P145213 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P151654 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P426373 BC099558.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P426373 NM_028483.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P426373 AK006670.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P426373 BC099558.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P426373 NM_028483.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P426373 AK006670.1 mouse_transcriptome
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P01064707 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P05520318 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P145213 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P151654 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P01064772 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P01064718 - -
          Whole Mouse Genome G4122A A_52_P987592 - -
          Whole Mouse Genome G4122A A_51_P426373 BC099558.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P426373 NM_028483.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P426373 AK006670.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P426373 BC099558.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P426373 NM_028483.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P426373 AK006670.1 mouse_transcriptome
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_67_P01064718 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P151654 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P145213 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23572828 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P04411579 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P04160865 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P04328911 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P04286195 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P04955504 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23572799 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23573119 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P04328910 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23572876 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P04427560 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23573055 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23573061 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23572897 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23573046 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23573114 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23572798 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P04755379 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P04113375 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P04682685 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23572834 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23573116 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P04542237 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23572832 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P04683912 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P04713235 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23573045 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P04341720 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P04552229 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23573042 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23573110 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23573080 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23572851 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23573118 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23573053 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P04955496 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P04152191 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P04552230 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P04542243 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P04682678 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P04542241 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23573117 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P04402231 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P04411584 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23573063 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23573093 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P04552231 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P04755377 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23573041 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23573079 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P04552220 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P04411580 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P04800634 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23572831 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P04871805 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23572899 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23573121 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23572896 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23572854 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23573083 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23573106 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23573096 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23572889 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P04152194 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P04755374 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23573135 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23573074 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23573070 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23573065 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P04825142 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P04152188 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P04913567 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23572852 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23572891 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P04868595 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P04411578 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P04478558 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P04198502 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P04438120 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23572830 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P04336399 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P04706666 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P04411577 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P04409701 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P04478540 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P04478539 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23572882 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P04542238 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23573133 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P04427548 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P04552217 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P04913565 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23572884 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23573084 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P04478559 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23572800 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23572845 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23572893 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P04755380 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23573077 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P04478561 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P04405961 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23573108 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23573044 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P04427555 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P04427554 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P04651393 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23572839 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P04984556 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P04650170 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10529025 - -
          Affymetrix Mouse Gene 1.0 ST Array 10529021 - -
          Affymetrix Mouse Gene 1.0 ST Array 10529022 - -
          Affymetrix Mouse Gene 1.0 ST Array 10529024 - -
          Affymetrix Mouse Gene 1.0 ST Array 10529020 - -
          Affymetrix Mouse Gene 1.0 ST Array 10529019 - -
          Affymetrix Mouse Gene 1.0 ST Array 10529023 - -
          Affymetrix HT MG-430 PM Array Plate 1454248_PM_at - -
          Affymetrix Mouse Gene 1.1 ST Array 10529018 - -
          Mouse Genome 430 2.0 Array 1458775_at - -
          Mouse Genome 430 2.0 Array 1454248_at - -
          Mouse Genome 430 2.0 Array 1458775_PM_at - -
          Mouse Genome 430 2.0 Array 1454248_PM_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 73259_PM_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 73259_at - -
          Affymetrix Mouse Clariom S Array PSR0500019632.mm.1 TC0500002158.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0500019632.mm.1 NM_028483.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0500019632.mm.1 NM_028483.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0500019633.mm.1 TC0500002158.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0500019633.mm.1 NM_028483.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0500019633.mm.1 NM_028483.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0500019629.mm.1 TC0500002158.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0500019629.mm.1 NM_028483.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0500019629.mm.1 NM_028483.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0500019630.mm.1 TC0500002158.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0500019630.mm.1 NM_028483.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0500019630.mm.1 NM_028483.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0500019631.mm.1 TC0500002158.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0500019631.mm.1 NM_028483.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0500019631.mm.1 NM_028483.1 Refseq
          Mouse Expression Set 430 1454248_PM_at - -
          Mouse Expression Set 430 1454248_at - -
          Affymetrix HT Mouse Genome 430B Array 1454248_PM_at - -
          Affymetrix HT Mouse Genome 430B Array 1454248_at - -
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 411076 - -
          Array Name Probe IDs Targets Target Type
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_2612107 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_2612107 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-74594944d-pk6zc:80/100.66.130.62:80.
          git-commit: ab2b2261c2971e97b1ae408e3ad10520b8413932
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.31.0-2025.06.23-1.0