Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm01210980_g1
          See other TM2D2 GE Assays ›
          Gene Symbol:
          Tm2d2
          Gene Name:
          TM2 domain containing 2
          Gene Aliases:
          2410018G23Rik, Blp1
          Chromosome Location:
          Chr.8: 25017211 - 25023260 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm01210980_g1
          Size
          Availability Made To Order
          Catalog # 4351372
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Tm2d2
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_027194.3 NP_081470.2 3 - 4 589 104
          GenBank mRNA AK010545.1 - 3 - 4 521 104
          AK045768.1 - 3 - 4 548 104
          AK080854.1 - 3 - 4 517 104
          AK083654.1 - 3 - 4 104
          AK085975.1 - 3 - 4 541 104
          AK086015.1 - 3 - 4 525 104
          AK086059.1 - 3 - 4 544 104
          AK088246.1 - 3 - 4 1054 104
          AK149407.1 - 3 - 4 518 104
          AK160039.1 - 3 - 4 526 104
          AK166012.1 - 3 - 4 542 104
          AK206565.1 - 2 - 3 168 104
          BC026789.1 - 3 - 4 516 4188168 104

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC0800000312.mm.1

          Set Membership:

          Ambion siRNA available Amplicon length greater than or equal to 101 3' Most Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Tm2d2

          Gene Ontology Categories:

          Function(s) Process(es)

          biological_process
          molecular_function

          MicroArray Probes:

          Agilent Affymetrix Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P25724849 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P25724846 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P25724848 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P25724847 - -
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P504588 BC026789.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P504588 CO044843.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P504588 AK086059.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P504588 AK088246.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P504588 AK149407.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P504588 BC026789.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P504588 CO044843.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P504588 AK086059.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P504588 AK088246.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P504588 AK149407.1 mouse_transcriptome
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 852519 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 887597 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 665618 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 52432 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P504588 BC026789.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P504588 CO044843.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P504588 AK086059.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P504588 AK088246.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P504588 AK149407.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P504588 BC026789.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P504588 CO044843.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P504588 AK086059.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P504588 AK088246.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P504588 AK149407.1 mouse_transcriptome
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25724855 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25724846 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25724865 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25724870 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25724874 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25724852 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25724872 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25724858 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25724847 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25724857 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25724866 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25724867 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25724856 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25724854 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25724850 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25724869 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25724876 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25724868 - -
          Whole Mouse Genome G4122A A_51_P504588 BC026789.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P504588 CO044843.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P504588 AK086059.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P504588 AK088246.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P504588 AK149407.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P504588 BC026789.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P504588 CO044843.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P504588 AK086059.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P504588 AK088246.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P504588 AK149407.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P493867 AK206565.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P493867 BC026789.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P493867 AK086059.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P493867 AK088246.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P493867 AK149407.1 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P109038 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P07281376 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P07281384 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P07281386 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P25724847 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P25724846 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P25724848 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P25724849 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25724877 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25724870 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25724949 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25724865 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25724932 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25724869 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25724936 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25724938 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25724850 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25724887 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25724872 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25724950 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25724931 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25724940 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25724866 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25724842 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25724884 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25724941 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25724867 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25724854 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25724880 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25724934 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25724856 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25724926 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25724925 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25724947 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25724939 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25724846 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25724882 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25724888 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25724857 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25724946 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25724852 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25724847 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25724868 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25724890 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25724878 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25724944 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25724854 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25724856 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25724888 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25724868 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25724865 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25724867 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P07137196 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25724855 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25724858 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25724866 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25724847 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P07397244 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25724880 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P07397242 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P07281375 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P07397238 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P07276254 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25724890 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25724850 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25724882 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25724852 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25724874 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P07397241 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25724887 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25724872 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25724864 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25724857 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10570978 - -
          Affymetrix Mouse Gene 1.0 ST Array 10570977 - -
          Affymetrix Mouse Gene 1.0 ST Array 10570976 - -
          Affymetrix Mouse Gene 1.0 ST Array 10570979 - -
          Affymetrix HT MG-430 PM Array Plate 1438181_PM_x_at - -
          Affymetrix HT MG-430 PM Array Plate 1456663_PM_x_at - -
          Affymetrix HT MG-430 PM Array Plate 1439234_PM_a_at - -
          Affymetrix HT MG-430 PM Array Plate 1439454_PM_x_at - -
          Affymetrix HT MG-430 PM Array Plate 1433473_PM_x_at - -
          Affymetrix HT MG-430 PM Array Plate 1437725_PM_x_at - -
          Affymetrix HT MG-430 PM Array Plate 1439235_PM_x_at - -
          Affymetrix HT MG-430 PM Array Plate 1451134_PM_a_at - -
          Affymetrix Mouse Gene 1.1 ST Array 10570975 - -
          Mouse Genome 430 2.0 Array 1433473_x_at - -
          Mouse Genome 430 2.0 Array 1451134_a_at - -
          Mouse Genome 430 2.0 Array 1439235_x_at - -
          Mouse Genome 430 2.0 Array 1433473_PM_x_at - -
          Mouse Genome 430 2.0 Array 1439234_PM_a_at - -
          Mouse Genome 430 2.0 Array 1439454_PM_x_at - -
          Mouse Genome 430 2.0 Array 1456663_x_at - -
          Mouse Genome 430 2.0 Array 1439234_a_at - -
          Mouse Genome 430 2.0 Array 1456663_PM_x_at - -
          Mouse Genome 430 2.0 Array 1438181_x_at - -
          Mouse Genome 430 2.0 Array 1451134_PM_a_at - -
          Mouse Genome 430 2.0 Array 1437725_x_at - -
          Mouse Genome 430 2.0 Array 1438181_PM_x_at - -
          Mouse Genome 430 2.0 Array 1439454_x_at - -
          Mouse Genome 430 2.0 Array 1437725_PM_x_at - -
          Mouse Genome 430 2.0 Array 1439235_PM_x_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 69742_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 69742_PM_at - -
          Affymetrix Mouse Clariom S Array PSR0800002323.mm.1 TC0800000312.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0800002323.mm.1 NM_027194.3.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0800002323.mm.1 NM_027194.3 Refseq
          Mouse Expression Set 430 1433473_x_at - -
          Mouse Expression Set 430 1437725_PM_x_at - -
          Mouse Expression Set 430 1438181_PM_x_at - -
          Mouse Expression Set 430 1439454_x_at - -
          Mouse Expression Set 430 1439235_x_at - -
          Mouse Expression Set 430 1451134_PM_a_at - -
          Mouse Expression Set 430 1439235_PM_x_at - -
          Mouse Expression Set 430 1451134_a_at - -
          Mouse Expression Set 430 1456663_PM_x_at - -
          Mouse Expression Set 430 1439234_PM_a_at - -
          Mouse Expression Set 430 1439454_PM_x_at - -
          Mouse Expression Set 430 1433473_PM_x_at - -
          Mouse Expression Set 430 1438181_x_at - -
          Mouse Expression Set 430 1456663_x_at - -
          Mouse Expression Set 430 1437725_x_at - -
          Mouse Expression Set 430 1439234_a_at - -
          Murine Genome U74v2 Set 98920_PM_g_at - -
          Murine Genome U74v2 Set 98919_PM_at - -
          Murine Genome U74v2 Set 98921_at - -
          Murine Genome U74v2 Set 98919_at - -
          Murine Genome U74v2 Set 98920_g_at - -
          Murine Genome U74v2 Set 98921_PM_at - -
          Mouse Genome 430A 2.0 Array 1456663_x_at - -
          Mouse Genome 430A 2.0 Array 1439234_PM_a_at - -
          Mouse Genome 430A 2.0 Array 1451134_a_at - -
          Mouse Genome 430A 2.0 Array 1438181_PM_x_at - -
          Mouse Genome 430A 2.0 Array 1439234_a_at - -
          Mouse Genome 430A 2.0 Array 1451134_PM_a_at - -
          Mouse Genome 430A 2.0 Array 1456663_PM_x_at - -
          Mouse Genome 430A 2.0 Array 1433473_x_at - -
          Mouse Genome 430A 2.0 Array 1433473_PM_x_at - -
          Mouse Genome 430A 2.0 Array 1439235_x_at - -
          Mouse Genome 430A 2.0 Array 1437725_PM_x_at - -
          Mouse Genome 430A 2.0 Array 1438181_x_at - -
          Mouse Genome 430A 2.0 Array 1439454_PM_x_at - -
          Mouse Genome 430A 2.0 Array 1437725_x_at - -
          Mouse Genome 430A 2.0 Array 1439454_x_at - -
          Mouse Genome 430A 2.0 Array 1439235_PM_x_at - -
          Affymetrix HT Mouse Genome 430A Array 1439454_x_at - -
          Affymetrix HT Mouse Genome 430A Array 1439234_a_at - -
          Affymetrix HT Mouse Genome 430A Array 1456663_PM_x_at - -
          Affymetrix HT Mouse Genome 430A Array 1437725_x_at - -
          Affymetrix HT Mouse Genome 430A Array 1437725_PM_x_at - -
          Affymetrix HT Mouse Genome 430A Array 1439235_PM_x_at - -
          Affymetrix HT Mouse Genome 430A Array 1439234_PM_a_at - -
          Affymetrix HT Mouse Genome 430A Array 1439235_x_at - -
          Affymetrix HT Mouse Genome 430A Array 1456663_x_at - -
          Affymetrix HT Mouse Genome 430A Array 1439454_PM_x_at - -
          Affymetrix HT Mouse Genome 430A Array 1433473_x_at - -
          Affymetrix HT Mouse Genome 430A Array 1451134_PM_a_at - -
          Affymetrix HT Mouse Genome 430A Array 1433473_PM_x_at - -
          Affymetrix HT Mouse Genome 430A Array 1438181_x_at - -
          Affymetrix HT Mouse Genome 430A Array 1438181_PM_x_at - -
          Affymetrix HT Mouse Genome 430A Array 1451134_a_at - -
          Array Name Probe IDs Targets Target Type
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_2679723 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_2679723 - -
          mouse-6 v1.1 (Illumina) scl33976.4_321-S - -
          Illumina Sentrix mouse-6.v1 bead array 780026 - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl33976.4_321-S - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 780026 - -
          MouseRef-8 v2 Expression BeadChip ILMN_2679723 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_2679723 - -
          Illumina Sentrix Mouse-6 Expression BeadChip (3) scl33976.4_321-S - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-6f7b98477d-hxtzg:80/100.66.130.62:80.
          git-commit: 5163bf559e7ad0ebe9326a88094353fc6888890d
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.31.0-Offline