Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Slc2a9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001012363.2 | NP_001012363.2 | 6 - 7 | 738 | 77 | |
NM_001102414.1 | NP_001095884.1 | 6 - 7 | 937 | 77 | ||
GenBank mRNA | AF469480.1 | - | 6 - 7 | 941 | 77 | |
AK156740.1 | - | 6 - 7 | 841 | 77 | ||
AK172234.1 | - | 6 - 7 | 841 | 77 | ||
AY776155.1 | - | 6 - 7 | 717 | 77 | ||
BC138213.1 | - | 7 - 8 | 873 | 8861232 | 77 |
Set Membership: |
![]() ![]() ![]() ![]() ![]() |