Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm01211308_m1
          See other 1700123M08RIK GE Assays ›
          Gene Symbol:
          1700123M08Rik
          Gene Name:
          RIKEN cDNA 1700123M08 gene
          Gene Aliases:
          1600025K15Rik
          Chromosome Location:
          Chr.4: 11966574 - 11994295 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm01211308_m1
          Size
          Availability Made To Order
          Catalog # 4426961
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: 1700123M08Rik
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NR_040577.1 4 - 5 636 99
          GenBank mRNA AK007255.1 - 4 - 5 637 99
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Transcript Location Amplicon Length
          GenBankmRNA BY707357.1 - 4 - 5 637 Chr. 4 - 11966574 - 11993999 [+] on Build GRCm38 99

          Adjacent Gene Details

          Gene name Gene Symbol Alias Distance Relative Direction Location Chromsome
          Upstream pyruvate dehyrogenase phosphatase catalytic subunit 1 Pdp1 Gm1024,
          Ppm2c
          -13.7kb Anti-sense Chr. 4 - 11958183 - 11966450 on NCBI Build GRCm38
          Downstream transmembrane protein 67 Tmem67 5330408M12Rik,
          B230117O07,
          b2b1163.1Clo,
          b2b1291.1Clo
          45.4kb Anti-sense Chr. 4 - 12039355 - 12089365 on NCBI Build GRCm38

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC0400000098.mm.1

          Set Membership:

          Amplicon length between 86 and 100 3' Most Mus musculus 1700 Probe spans exons

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 191622 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 16206 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 679750 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 452766 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 58321 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 647828 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 192343 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 935447 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 962449 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 554647 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 643359 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 830368 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 179043 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 148617 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 107941 - -
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P120976 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22732642 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22732650 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22732570 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03710224 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22732579 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22732644 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22732639 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22732638 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22732641 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22732643 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22732573 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22732647 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22732648 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03830638 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22732646 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22732652 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Clariom S Array PSR0400000694.mm.1 TC0400000098.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0400000694.mm.1 NR_040577.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0400000694.mm.1 NR_040577.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0400000690.mm.1 TC0400000098.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0400000690.mm.1 NR_040577.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0400000690.mm.1 NR_040577.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0400000693.mm.1 TC0400000098.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0400000693.mm.1 NR_040577.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0400000693.mm.1 NR_040577.1 Refseq
          Murine Genome U74v2 Set 171258_r_at - -
          Murine Genome U74v2 Set 171258_PM_r_at - -
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 382738 - -
          Array Name Probe IDs Targets Target Type
          MouseWG-6 v2 Expression BeadChip 105700372 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-784b477dbd-pq4vn:80/100.66.130.62:80.
          git-commit: d41a97e986437cfbf980934f14197a4b4d3eb741
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.31.0-2025.06.23-1.0