Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm01211558_m1
          See other NOA1 GE Assays ›
          Gene Symbol:
          Noa1
          Gene Name:
          nitric oxide associated 1
          Gene Aliases:
          2610024G14Rik, AU022345, AtNOS1, mAtNOS1
          Chromosome Location:
          Chr.5: 77294169 - 77310158 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm01211558_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Noa1
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_019836.3 NP_062810.1 4 - 5 1682 66
          GenBank mRNA AB041537.1 - 4 - 5 1654 66
          AK011533.1 - 4 - 5 1674 66
          AK017793.1 - 4 - 5 1655 66
          AK019975.1 - 4 - 5 1664 66
          AK128920.1 - 4 - 5 1654 66
          AK139955.1 - 4 - 5 1626 66
          AK157246.1 - 4 - 5 1682 66
          BC005689.1 - 4 - 5 735 66
          BC017154.1 - 4 - 5 1654 4910547 66

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC0500002650.mm.1

          Set Membership:

          Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Recommended Assay Mus musculus Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Noa1

          Gene Ontology Categories:

          Function(s) Process(es)

          apoptotic process
          regulation of cell death
          mitochondrial translation
          ribosome biogenesis
          regulation of cellular respiration
          nucleotide binding
          RNA binding
          GTPase activity
          GTP binding

          MicroArray Probes:

          Agilent Affymetrix Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P23820896 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P23820887 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P23820886 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P23820895 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P23820888 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P23820889 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P23820890 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P23820894 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P23820897 - -
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2066468 BC017154.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2066468 BC005689.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2066468 AK128920.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2066468 AK139955.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2066468 AK157246.1 mouse_transcriptome
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 707924 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 679537 AK012845.1 genbank_mrna_mouse
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 679537 AK017607.1 genbank_mrna_mouse
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 679537 AK019268.1 genbank_mrna_mouse
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 679537 AK133858.1 genbank_mrna_mouse
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 679537 BC046780.1 genbank_mrna_mouse
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 679537 BC052070.1 genbank_mrna_mouse
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 679537 NM_025676.3 refseq_mrna_mouse_NM
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 361176 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 447042 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 55527 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 51226 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 25953 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 273059 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 45610 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 109280 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P176715 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P149911 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2066468 BC017154.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2066468 BC005689.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2066468 AK128920.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2066468 AK139955.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2066468 AK157246.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P472488 BC017154.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P472488 BC005689.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P472488 AK128920.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P472488 AK139955.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P472488 AK157246.1 mouse_transcriptome
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23820881 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23820873 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23820880 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23820869 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23820890 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23820871 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23820876 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23820883 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23820860 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23820892 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23820877 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23820865 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23820868 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23820896 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23820862 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23820895 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23820863 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23820872 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23820866 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23820885 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23820888 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P149911 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P176715 - -
          Whole Mouse Genome G4122A A_51_P472488 BC017154.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P472488 BC005689.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P472488 AK128920.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P472488 AK139955.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P472488 AK157246.1 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P110040 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P149911 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P176715 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P04148016 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P04148015 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P23820886 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P23820890 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P23820888 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P23820896 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P23820897 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P23820887 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P23820895 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P23820889 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P23820894 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23820837 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23820876 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23820862 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23820781 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23820858 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23820819 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23820890 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23820759 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23820810 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23820833 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23820869 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23820790 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23820866 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23820854 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23820756 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23820868 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23820895 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23820842 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23820780 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23820888 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23820798 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23820755 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23820857 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23820865 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23820793 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23820873 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23820850 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23820827 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23820840 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23820763 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23820754 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23820765 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23820815 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23820776 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23820823 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23820778 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23820884 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23820779 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23820892 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23820863 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23820880 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23820813 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23820838 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23820786 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23820777 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23820896 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23820882 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23820811 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23820800 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23820847 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23820773 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23820812 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23820805 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23820782 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23820806 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23820829 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23820854 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23820840 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23820866 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23820859 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23820861 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23820850 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23820877 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23820856 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23820865 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23820873 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P04543968 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P04119608 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23820860 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23820857 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P04498828 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23820839 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23820868 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23820878 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P04543966 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23820871 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23820888 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P04308607 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23820876 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P04861692 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23820869 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23820847 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23820852 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23820842 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23820862 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P04119609 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P04148015 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23820884 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23820872 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P04990051 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23820844 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23820863 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P04134710 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10530840 - -
          Affymetrix Mouse Gene 1.0 ST Array 10530838 - -
          Affymetrix Mouse Gene 1.0 ST Array 10530834 - -
          Affymetrix Mouse Gene 1.0 ST Array 10530835 - -
          Affymetrix Mouse Gene 1.0 ST Array 10530839 - -
          Affymetrix Mouse Gene 1.0 ST Array 10530836 - -
          Affymetrix Mouse Gene 1.0 ST Array 10530833 - -
          Affymetrix Mouse Gene 1.0 ST Array 10530837 - -
          Affymetrix HT MG-430 PM Array Plate 1418222_PM_at - -
          Affymetrix Mouse Gene 1.1 ST Array 10530832 - -
          Mouse Genome 430 2.0 Array 1418222_at - -
          Mouse Genome 430 2.0 Array 1418222_PM_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 56412_PM_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 56412_at - -
          Affymetrix Mouse Clariom S Array PSR0500023428.mm.1 TC0500002650.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0500023428.mm.1 NM_019836.3.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0500023428.mm.1 NM_019836.3 Refseq
          Mouse Expression Set 430 1418222_at - -
          Mouse Expression Set 430 1418222_PM_at - -
          Mouse Genome 430A 2.0 Array 1418222_at - -
          Mouse Genome 430A 2.0 Array 1418222_PM_at - -
          Affymetrix HT Mouse Genome 430A Array 1418222_at - -
          Affymetrix HT Mouse Genome 430A Array 1418222_PM_at - -
          Array Name Probe IDs Targets Target Type
          MouseWG-6 v2 Expression BeadChip 3450079 - -
          MouseWG-6 v2 Expression BeadChip 7040408 - -
          Illumina mouse-6 v1.0 expression beadchip 3450079 - -
          Illumina mouse-6 v1.0 expression beadchip 7040408 - -
          Illumina mouse-6 v1.0 expression beadchip 4850068 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0004850068 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0007040408 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0003450079 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_2775352 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_1226758 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_1258552 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_2775352 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_1258552 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_1226758 - -
          mouse-6 v1.1 (Illumina) scl0004068.1_16-S - -
          mouse-6 v1.1 (Illumina) scl0003995.1_47-S - -
          mouse-6 v1.1 (Illumina) scl26446.8.1_29-S - -
          Illumina Sentrix mouse-6.v1 bead array 7040408 - -
          Illumina Sentrix mouse-6.v1 bead array 3450079 - -
          Illumina Sentrix mouse-6.v1 bead array 4850068 - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl26446.8.1_29-S - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl0004068.1_16-S - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl0003995.1_47-S - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 3450079 - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 4850068 - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 7040408 - -
          MouseRef-8 v2 Expression BeadChip ILMN_2599935 - -
          MouseRef-8 v2 Expression BeadChip ILMN_2835461 - -
          MouseRef-8 v2 Expression BeadChip ILMN_1258552 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_2599935 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_1258552 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_2835461 - -
          Illumina Sentrix Mouse-6 Expression BeadChip (3) scl26446.8.1_29-S - -
          Illumina Sentrix Mouse-6 Expression BeadChip (3) scl0004068.1_16-S - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-74594944d-pk6zc:80/100.66.130.62:80.
          git-commit: ab2b2261c2971e97b1ae408e3ad10520b8413932
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.31.0-2025.06.23-1.0