Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm01212290_m1
          See other URAD GE Assays ›
          Gene Symbol:
          Urad
          Gene Name:
          ureidoimidazoline (2-oxo-4-hydroxy-4-carboxy-5) decarboxylase
          Gene Aliases:
          EG231903, Prhoxnb
          Chromosome Location:
          Chr.5: 147314984 - 147322440 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm01212290_m1
          Best Coverage
          Size
          Availability Made To Order
          Catalog # 4351372
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Urad
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001039678.2 NP_001034767.1 1 - 2 284 96
          GenBank mRNA BC147779.1 - 1 - 2 284 9087499 96
          BC147790.1 - 1 - 2 284 9087508 96
          DQ225767.1 - 1 - 2 184 96

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC0500003625.mm.1

          Set Membership:

          Amplicon length between 86 and 100 3' Most Mammalian Gene Collection Recommended Assay Mus musculus Probe spans exons

          Gene Ontology Categories:

          Function(s) Process(es)

          purine nucleobase metabolic process
          allantoin biosynthetic process
          lyase activity
          carboxy-lyase activity

          MicroArray Probes:

          Agilent Affymetrix Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P467861 NM_001039678.2 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P467861 BC147779.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P467861 BC147790.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P467861 DQ225767.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P467861 NM_001039678.2 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P467861 BC147779.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P467861 BC147790.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P467861 DQ225767.1 mouse_transcriptome
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 312046 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 629058 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 259638 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 85504 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 81371 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P467861 NM_001039678.2 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P467861 BC147779.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P467861 BC147790.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P467861 DQ225767.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P467861 NM_001039678.2 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P467861 BC147779.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P467861 BC147790.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P467861 DQ225767.1 mouse_transcriptome
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P05783155 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P01246185 - -
          Whole Mouse Genome G4122A A_52_P467861 NM_001039678.2 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P467861 BC147779.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P467861 BC147790.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P467861 DQ225767.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P467861 NM_001039678.2 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P467861 BC147779.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P467861 BC147790.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P467861 DQ225767.1 mouse_transcriptome
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_67_P01246185 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24173112 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24173095 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24173106 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24173020 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24173060 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24173074 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24173109 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24173134 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24173032 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24173102 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24173004 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24173111 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24172973 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24173045 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24173008 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24173043 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24172989 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24173002 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24173041 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24173141 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24172975 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24173132 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24173087 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24172972 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24173093 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24173029 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24173024 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24173025 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24173081 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24172993 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24173103 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24172986 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24173062 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24173079 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24173010 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24173017 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24172978 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24172970 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24173114 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24173073 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24173097 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24173065 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24173028 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24173096 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24173035 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24173119 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24173069 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24173006 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24173075 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24173077 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24172995 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24172987 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24173131 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24173056 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24173005 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24173083 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24173038 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24172981 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24172984 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24173129 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24173138 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24173117 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24173137 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24173086 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24172991 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24173133 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24172982 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24173088 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24172992 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24173046 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24173125 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24173099 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24173058 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24173092 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24173011 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24172968 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24172999 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24173053 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24173026 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24173042 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24173064 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24173014 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P24173084 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P24173070 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P24173099 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P04635627 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P24173090 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P24173074 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P04861741 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P24173075 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P24173137 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P24173121 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P24173092 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P24173064 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P04539943 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P04931048 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P24173095 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P24173093 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P24173106 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P24173131 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P24173136 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P24173103 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P24173132 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P24173088 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P04679092 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P04727056 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P24173134 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P24173126 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P04508348 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P24173056 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P24173116 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P24173079 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P24173086 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P24173073 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P04936348 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P24173114 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P24173091 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P24173065 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P24173125 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P24173060 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P24173117 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P24173059 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P24173066 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P24173069 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P24173127 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P24173119 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P24173107 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P04579263 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P04605981 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P24173077 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P24173135 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P24173102 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P24173058 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P24173129 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P24173109 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10535777 - -
          Affymetrix Mouse Gene 1.0 ST Array 10535778 - -
          Affymetrix Mouse Gene 1.0 ST Array 10535779 - -
          Affymetrix Mouse Gene 1.1 ST Array 10535776 - -
          Affymetrix Mouse Clariom S Array PSR0500033263.mm.1 TC0500003625.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0500033263.mm.1 NM_001039678.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0500033263.mm.1 NM_001039678.2 Refseq
          Affymetrix Mouse Clariom S Array PSR0500033262.mm.1 TC0500003625.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0500033262.mm.1 NM_001039678.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0500033262.mm.1 NM_001039678.2 Refseq
          Affymetrix Mouse Clariom S Array PSR0500033264.mm.1 TC0500003625.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0500033264.mm.1 NM_001039678.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0500033264.mm.1 NM_001039678.2 Refseq
          Array Name Probe IDs Targets Target Type
          Illumina mouse-6 v1.1 expression beadchip ILMN_1229187 - -
          MouseRef-8 v2 Expression BeadChip ILMN_2819424 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_2819424 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-f85dfd666-px2hc:80/100.66.130.62:80.
          git-commit: 2ca7b3ab9a2195be9b417413331e4e963d93e17c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.32.0-2025.07.28-1.0