Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm01212793_m1
          See other POMK GE Assays ›
          Gene Symbol:
          Pomk
          Gene Name:
          protein-O-mannose kinase
          Gene Aliases:
          4930444A02Rik, Sgk196
          Chromosome Location:
          Chr.8: 25980604 - 25994121 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm01212793_m1
          Size
          Availability Made To Order
          Catalog # 4351372
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Pomk
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_029037.4 NP_083313.1 1 - 2 35 65
          GenBank mRNA AK015374.1 - 1 - 2 28 65
          AK032677.1 - 1 - 2 36 65
          AK036348.1 - 1 - 2 42 65
          AK043620.1 - 1 - 2 36 65
          AK138952.1 - 1 - 2 44 65
          AK160873.1 - 1 - 2 41 65
          AK170994.1 - 1 - 2 36 65

          Back To Top

          More Information


          Additional Information:

          This assay is in the 5'UTR region of the AK015374.1, AK032677.1, AK036348.1, AK043620.1, AK138952.1, AK160873.1, AK170994.1, NM_029037.4 transcripts.

          Set Membership:

          Ambion siRNA available Amplicon length less than or equal to 70 Mus musculus 1700 Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Pomk

          Gene Ontology Categories:

          Function(s) Process(es)

          neuron migration
          protein phosphorylation
          protein O-linked glycosylation
          brain development
          learning or memory
          phosphorylation
          sensory perception of pain
          carbohydrate phosphorylation
          neuromuscular process
          nucleotide binding
          protein kinase activity
          ATP binding
          phosphofructokinase activity
          phosphorylase kinase regulator activity
          cobinamide kinase activity
          phytol kinase activity
          kinase activity
          phosphatidylinositol phosphate kinase activity
          cyclin-dependent protein serine/threonine kinase regulator activity
          transferase activity
          transferase activity, transferring phosphorus-containing groups
          phosphotransferase activity, alcohol group as acceptor
          phenol kinase activity
          carbohydrate kinase activity
          cyclin-dependent protein kinase activating kinase regulator activity
          inositol tetrakisphosphate 2-kinase activity
          heptose 7-phosphate kinase activity
          aminoglycoside phosphotransferase activity
          phosphatidylinositol 3-kinase activity
          eukaryotic elongation factor-2 kinase regulator activity
          eukaryotic elongation factor-2 kinase activator activity
          LPPG:FO 2-phospho-L-lactate transferase activity
          cytidine kinase activity
          glycerate 2-kinase activity
          (S)-lactate 2-kinase activity
          phosphoserine:homoserine phosphotransferase activity
          L-seryl-tRNA(Sec) kinase activity
          phosphocholine transferase activity
          polynucleotide 5'-hydroxyl-kinase activity
          polynucleotide kinase activity
          GTP-dependent polynucleotide kinase activity
          CTP:farnesol kinase activity
          geraniol kinase activity
          geranylgeraniol kinase activity
          CTP:geranylgeraniol kinase activity
          prenol kinase activity
          1-phosphatidylinositol-5-kinase activity
          1-phosphatidylinositol-3-phosphate 4-kinase activity
          phosphatidylinositol-3,4-bisphosphate 5-kinase activity
          phosphatidylinositol bisphosphate kinase activity
          inositol-3,4,6-trisphosphate 1-kinase activity
          inositol 5-diphosphate pentakisphosphate 5-kinase activity
          inositol diphosphate tetrakisphosphate kinase activity

          MicroArray Probes:

          ABI

          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 660628 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-5777fb6d44-4pg6x:80/100.66.134.75:80.
          git-commit: 3a4c0ffdaddae765c0fdfab5c280296089abf14d
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.33.0-2025.07.30-1.0