Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm01212798_g1
          See other GBA GE Assays ›
          Gene Symbol:
          Gba
          Gene Name:
          glucosidase, beta, acid
          Gene Aliases:
          GBA1, GC, GCase, GLUC, betaGC
          Chromosome Location:
          Chr.3: 89202914 - 89208966 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm01212798_g1
          Size
          Availability Made To Order
          Catalog # 4351372
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Gba
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001077411.2 NP_001070879.1 6 - 7 884 67
          NM_008094.5 NP_032120.1 7 - 8 1053 67
          NR_122037.1 7 - 8 1053 67
          GenBank mRNA AK082767.1 - 6 - 7 882 67
          AK152745.1 - 6 - 7 865 67
          AK166198.1 - 7 - 8 1044 67
          BC006663.1 - 7 - 8 1038 3498551 67
          M24119.1 - 6 - 7 793 67

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC0300000750.mm.1

          Set Membership:

          Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Gba

          Gene Ontology Categories:

          Function(s) Process(es)

          carbohydrate metabolic process
          lipid metabolic process
          sphingolipid metabolic process
          glucosylceramide catabolic process
          metabolic process
          response to pH
          termination of signal transduction
          negative regulation of interleukin-6 production
          regulation of water loss via skin
          response to testosterone
          positive regulation of protein dephosphorylation
          negative regulation of MAP kinase activity
          skin morphogenesis
          response to estrogen
          sphingosine biosynthetic process
          ceramide biosynthetic process
          response to glucocorticoid
          cellular response to tumor necrosis factor
          response to thyroid hormone
          positive regulation of proteolysis involved in cellular protein catabolic process
          glucosylceramidase activity
          signaling receptor binding
          hydrolase activity
          hydrolase activity, acting on glycosyl bonds

          MicroArray Probes:

          Affymetrix

          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Clariom S Array PSR0300005609.mm.1 TC0300000750.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0300005609.mm.1 NM_001077411.2 Refseq
          Affymetrix Mouse Clariom S Array PSR0300005609.mm.1 NM_008094.5 Refseq
          Affymetrix Mouse Clariom S Array PSR0300005610.mm.1 TC0300000750.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0300005610.mm.1 NM_001077411.2 Refseq
          Affymetrix Mouse Clariom S Array PSR0300005610.mm.1 NM_008094.5 Refseq

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-74594944d-pk6zc:80/100.66.130.62:80.
          git-commit: ab2b2261c2971e97b1ae408e3ad10520b8413932
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.31.0-2025.06.23-1.0