Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm01213924_gH
          See other UGT2A3 GE Assays ›
          Gene Symbol:
          Ugt2a3
          Gene Name:
          UDP glucuronosyltransferase 2 family, polypeptide A3
          Gene Aliases:
          2010321J07Rik
          Chromosome Location:
          Chr.5: 87324972 - 87337195 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm01213924_gH
          Size
          Availability Made To Order
          Catalog # 4351372
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Ugt2a3
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_028094.3 NP_082370.2 4 - 5 1125 99
          GenBank mRNA AK050327.1 - 4 - 5 1126 99
          BC025795.1 - 4 - 5 1101 5133743 99

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC0500002695.mm.1

          Set Membership:

          Ambion siRNA available Amplicon length between 86 and 100 3' Most Mammalian Gene Collection Detect Genomic DNA Mus musculus 1700 Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Ugt2a3

          Gene Ontology Categories:

          Function(s) Process(es)

          metabolic process
          flavonoid biosynthetic process
          cellular response to hormone stimulus
          cellular glucuronidation
          flavonoid glucuronidation
          glucuronosyltransferase activity
          transferase activity
          glycosyltransferase activity
          hexosyltransferase activity

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P298023 BC025795.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P298023 AK050327.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P298023 NM_028094.3 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P298023 AK008601.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P298023 BC025795.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P298023 AK050327.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P298023 NM_028094.3 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P298023 AK008601.1 mouse_transcriptome
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 764724 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 418705 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 351209 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 249424 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 54593 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 39774 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P149987 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P178881 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P298023 BC025795.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P298023 AK050327.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P298023 NM_028094.3 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P298023 AK008601.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P298023 BC025795.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P298023 AK050327.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P298023 NM_028094.3 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P298023 AK008601.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P298023 BC025795.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P298023 AK050327.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P298023 NM_028094.3 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P298023 AK008601.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P298023 BC025795.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P298023 AK050327.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P298023 NM_028094.3 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P298023 AK008601.1 mouse_transcriptome
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23867368 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23867352 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23867360 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23867355 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23867353 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23867347 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23867357 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23867351 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23867349 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23867361 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23867364 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23867346 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P05644570 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P178881 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P05644581 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P149987 - -
          Whole Mouse Genome G4122A A_51_P298023 BC025795.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P298023 AK050327.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P298023 NM_028094.3 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P298023 AK008601.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P298023 BC025795.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P298023 AK050327.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P298023 NM_028094.3 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P298023 AK008601.1 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P132733 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P149987 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P178881 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P04735363 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P04735366 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23867368 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23867357 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23867299 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23867326 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23867283 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23867275 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23867285 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23867364 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23867315 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23867320 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23867287 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23867292 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23867322 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23867330 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23867355 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23867327 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23867341 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23867301 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23867339 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23867336 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23867347 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23867307 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23867282 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23867290 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23867324 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23867353 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23867294 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23867280 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23867318 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23867311 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23867329 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23867295 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23867304 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23867349 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23867351 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23867338 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23867319 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23867360 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23867373 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23867286 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23867337 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23867378 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P04835709 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23867355 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23867347 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23867357 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23867341 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23867368 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23867320 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P04735366 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23867336 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23867359 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23867349 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P04198097 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23867360 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P04198094 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P04538152 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23867324 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23867330 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23867353 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P04584084 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P04142279 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23867331 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23867329 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23867322 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23867327 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23867346 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P04584082 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P04835694 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23867364 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23867351 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23867334 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23867343 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10531071 - -
          Affymetrix Mouse Gene 1.0 ST Array 10531069 - -
          Affymetrix Mouse Gene 1.0 ST Array 10531068 - -
          Affymetrix Mouse Gene 1.0 ST Array 10531072 - -
          Affymetrix Mouse Gene 1.0 ST Array 10531070 - -
          Affymetrix Mouse Gene 1.0 ST Array 10531067 - -
          Affymetrix HT MG-430 PM Array Plate 1450133_PM_at - -
          Affymetrix Mouse Gene 1.1 ST Array 10531066 - -
          Mouse Genome 430 2.0 Array 1450133_at - -
          Mouse Genome 430 2.0 Array 1450133_PM_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 72094_PM_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 72094_at - -
          Affymetrix Mouse Clariom S Array PSR0500023793.mm.1 TC0500002695.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0500023793.mm.1 NM_028094.3.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0500023793.mm.1 NM_028094.3 Refseq
          Affymetrix Mouse Clariom S Array PSR0500023792.mm.1 TC0500002695.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0500023792.mm.1 NM_028094.3.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0500023792.mm.1 NM_028094.3 Refseq
          Mouse Expression Set 430 1450133_PM_at - -
          Mouse Expression Set 430 1450133_at - -
          Mouse Genome 430A 2.0 Array 1450133_PM_at - -
          Mouse Genome 430A 2.0 Array 1450133_at - -
          Affymetrix HT Mouse Genome 430A Array 1450133_PM_at - -
          Affymetrix HT Mouse Genome 430A Array 1450133_at - -
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 389661 - -
          Array Name Probe IDs Targets Target Type
          MouseWG-6 v2 Expression BeadChip 50138 - -
          Illumina mouse-6 v1.0 expression beadchip 50138 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0000050138 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_2714195 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_2714195 - -
          mouse-6 v1.1 (Illumina) scl26417.6.1_23-S - -
          Illumina Sentrix mouse-6.v1 bead array 50138 - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl26417.6.1_23-S - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 50138 - -
          MouseRef-8 v2 Expression BeadChip ILMN_2982764 - -
          MouseRef-8 v2 Expression BeadChip ILMN_2714195 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_2714195 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_2982764 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-7b745768ff-vzrjf:80/100.66.128.131:80.
          git-commit: 4b95038805a43672018eef8c57cc3f15d8b0c074
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: integration/2.33.0-meta-desc