Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm01214283_m1
          See other CHUK GE Assays ›
          Gene Symbol:
          Chuk
          Gene Name:
          conserved helix-loop-helix ubiquitous kinase
          Gene Aliases:
          AI256658, Chuk1, Fbx24, Fbxo24, IKBKA, IKK1, Ikka, NFKBIKA
          Chromosome Location:
          Chr.19: 44073334 - 44107506 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm01214283_m1
          Size
          Availability Made To Order
          Catalog # 4351372
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Chuk
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001162410.1 NP_001155882.1 13 - 14 1597 98
          NM_007700.2 NP_031726.2 13 - 14 1597 98
          GenBank mRNA AK018671.1 - 13 - 14 1576 98
          AK035326.1 - 13 - 14 1554 98
          AK150039.1 - 13 - 14 1576 98
          AK155328.1 - 13 - 14 1577 98
          AK155393.1 - 13 - 14 1598 98
          AK164236.1 - 13 - 14 1598 98
          AK193721.1 - 2 - 3 214 98
          BC018243.1 - 13 - 14 1589 4511275 98
          BC125266.1 - 2 - 3 196 98
          U12473.1 - 13 - 14 1565 98

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC1900001552.mm.1

          Set Membership:

          Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Mus musculus Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Chuk

          Gene Ontology Categories:

          Function(s) Process(es)

          morphogenesis of an epithelial sheet
          protein phosphorylation
          I-kappaB phosphorylation
          Rho protein signal transduction
          lactation
          response to organic substance
          phosphorylation
          osteoclast differentiation
          response to lipopolysaccharide
          mammary gland epithelial cell proliferation
          response to muscle stretch
          odontogenesis of dentin-containing tooth
          positive regulation of I-kappaB kinase/NF-kappaB signaling
          positive regulation of transcription by RNA polymerase II
          striated muscle cell differentiation
          mammary gland alveolus development
          cellular response to tumor necrosis factor
          nucleotide binding
          protein kinase activity
          protein serine/threonine kinase activity
          protein binding
          ATP binding
          IkappaB kinase activity
          kinase activity
          transferase activity
          transferase activity, transferring phosphorus-containing groups
          protein homodimerization activity
          protein heterodimerization activity
          scaffold protein binding

          MicroArray Probes:

          Affymetrix

          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Clariom S Array PSR1900013742.mm.1 TC1900001552.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1900013742.mm.1 NM_001162410.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1900013742.mm.1 NM_001162410.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1900013742.mm.1 NM_007700.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1900013742.mm.1 NM_007700.2 Refseq
          Affymetrix Mouse Clariom S Array PSR1900013740.mm.1 TC1900001552.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1900013740.mm.1 NM_001162410.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1900013740.mm.1 NM_001162410.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1900013740.mm.1 NM_007700.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1900013740.mm.1 NM_007700.2 Refseq
          Affymetrix Mouse Clariom S Array PSR1900013743.mm.1 TC1900001552.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1900013743.mm.1 NM_001162410.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1900013743.mm.1 NM_001162410.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1900013743.mm.1 NM_007700.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1900013743.mm.1 NM_007700.2 Refseq
          Affymetrix Mouse Clariom S Array PSR1900013741.mm.1 TC1900001552.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1900013741.mm.1 NM_001162410.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1900013741.mm.1 NM_001162410.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1900013741.mm.1 NM_007700.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1900013741.mm.1 NM_007700.2 Refseq
          Affymetrix Mouse Clariom S Array PSR1900013739.mm.1 TC1900001552.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1900013739.mm.1 NM_001162410.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1900013739.mm.1 NM_001162410.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1900013739.mm.1 NM_007700.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1900013739.mm.1 NM_007700.2 Refseq

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-74594944d-pk6zc:80/100.66.130.62:80.
          git-commit: ab2b2261c2971e97b1ae408e3ad10520b8413932
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.31.0-2025.06.23-1.0