Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm01215088_m1
          See other DIDO1 GE Assays ›
          Gene Symbol:
          Dido1
          Gene Name:
          death inducer-obliterator 1
          Gene Aliases:
          6720461J16Rik, D130048F08Rik, DATF-1, DIO-1, Datf1, dido
          Chromosome Location:
          Chr.2: 180657963 - 180710035 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm01215088_m1
          Size
          Availability Made To Order
          Catalog # 4351372
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Dido1
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001291432.1 NP_001278361.1 2 - 3 271 63
          NM_001291433.1 NP_001278362.1 2 - 3 323 63
          NM_011805.3 NP_035935.2 2 - 3 323 63
          NM_175551.4 NP_780760.2 2 - 3 323 63
          NM_177852.4 NP_808520.2 2 - 3 271 63
          NR_111965.1 1 - 2 181 63
          GenBank mRNA AJ238332.1 - 2 - 3 225 63
          AK014422.1 - 2 - 3 324 63
          AK042474.1 - 2 - 3 292 63
          AK129117.1 - 2 - 3 271 63
          AY425951.1 - 2 - 3 225 63
          AY425952.1 - 2 - 3 225 63
          BC060249.1 - 2 - 3 271 63
          BC096662.1 - 2 - 3 263 63
          BC138712.1 - 1 - 2 100 8861734 63
          BC138713.1 - 1 - 2 100 8861735 63
          BC145401.1 - 1 - 2 100 63
          BY713603.1 - 2 - 3 321 63

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC0200005415.mm.1

          Additional Information:

          This assay is in the 5'UTR region of the AJ238332.1, AK042474.1, AY425951.1, AY425952.1, BC096662.1, BC138712.1, BC138713.1, NM_001291432.1, NM_001291433.1, NM_011805.3, NM_175551.4, NM_177852.4 transcripts.

          Set Membership:

          Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Mus musculus Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Dido1

          Gene Ontology Categories:

          Function(s) Process(es)

          transcription, DNA-templated
          apoptotic process
          apoptotic signaling pathway
          RNA binding
          zinc ion binding
          metal ion binding

          MicroArray Probes:

          Affymetrix

          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Clariom S Array PSR0200046139.mm.1 TC0200005415.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0200046139.mm.1 NM_011805.3 Refseq
          Affymetrix Mouse Clariom S Array PSR0200046139.mm.1 NM_175551.4 Refseq
          Affymetrix Mouse Clariom S Array PSR0200046139.mm.1 NM_177852.4 Refseq
          Affymetrix Mouse Clariom S Array PSR0200046151.mm.1 TC0200005415.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0200046151.mm.1 NM_177852.4 Refseq

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-97865cc7b-9fgnx:80/100.66.128.131:80.
          git-commit: d53bc399386730d6c5c6d01fe94b21e0ad3fdbfa
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.33.0-2025.07.30-1.0