Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm01215180_g1
          See other RPS21 GE Assays ›
          Gene Symbol:
          Rps21
          Gene Name:
          ribosomal protein S21
          Gene Aliases:
          1810049N11Rik, 2410030A14Rik
          Chromosome Location:
          Chr.2: 180257373 - 180258444 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm01215180_g1
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Rps21
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_025587.2 NP_079863.1 5 - 6 319 66
          GenBank mRNA AA049954.1 - 4 - 5 245 66
          AK007846.1 - 5 - 6 282 66
          AK007850.1 - 5 - 6 274 66
          AK010637.1 - 5 - 6 287 66
          AK012488.1 - 5 - 6 282 66
          BC027563.1 - 5 - 6 267 3468174 66
          BC086912.1 - 5 - 6 284 6758965 66
          BU534365.1 - 5 - 6 322 66

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC0200002785.mm.1

          Set Membership:

          Ambion siRNA available Amplicon length less than or equal to 70 3' Most Mammalian Gene Collection Detect Genomic DNA Mus musculus Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Rps21

          Gene Ontology Categories:

          Function(s) Process(es)

          endonucleolytic cleavage in ITS1 to separate SSU-rRNA from 5.8S rRNA and LSU-rRNA from tricistronic rRNA transcript (SSU-rRNA, 5.8S rRNA, LSU-rRNA)
          endonucleolytic cleavage to generate mature 3'-end of SSU-rRNA from (SSU-rRNA, 5.8S rRNA, LSU-rRNA)
          translation
          ribosomal small subunit biogenesis
          RNA binding
          structural constituent of ribosome
          protein N-terminus binding

          MicroArray Probes:

          Agilent Affymetrix Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P21902493 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P21902498 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P21902496 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P21902494 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P21902495 - -
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P740562 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 48460 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P145796 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P740562 - -
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_52_P678800 AK010610.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P489959 AK007846.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P489959 AK007850.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P489959 AK010610.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P489959 AK010637.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P489959 AK012488.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_52_P740562 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21902506 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21902484 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21902500 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21902481 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21902497 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21902486 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21902498 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21902485 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21902493 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21902488 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21902490 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21902507 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21902495 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21902491 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21902504 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21902503 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21902482 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21902480 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21902501 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21902496 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P145796 - -
          Whole Mouse Genome G4122A A_51_P489959 AK007846.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P489959 AK007850.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P489959 AK010610.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P489959 AK010637.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P489959 AK012488.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P740562 - -
          Whole Mouse Genome G4122A A_52_P678800 AK010610.1 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P109687 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P145796 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P02178031 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P02178037 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P02178033 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P01351578 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P21902498 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P21902493 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P21902496 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P21902494 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P21902495 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21902456 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21902571 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21902495 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21902574 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21902498 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21902484 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21902468 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21902579 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21902504 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21902582 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21902482 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21902465 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21902490 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21902462 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21902458 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21902577 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21902493 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21902457 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21902585 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21902500 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21902481 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21902497 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21902503 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21902475 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21902463 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21902476 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21902477 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21902471 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21902486 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21902588 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21902581 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21902587 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01961849 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21902490 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21902473 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01469637 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01454681 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P02178037 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21902470 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P02178031 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P02178034 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21902472 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21902477 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21902481 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P02178033 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21902486 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21902482 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01669712 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21902489 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21902504 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21902484 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21902480 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21902478 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21902488 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P02108674 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21902500 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21902503 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10479365 - -
          Affymetrix Mouse Gene 1.0 ST Array 10479364 - -
          Affymetrix Mouse Gene 1.0 ST Array 10479363 - -
          Affymetrix Mouse Gene 1.0 ST Array 10479366 - -
          Affymetrix Mouse Gene 1.0 ST Array 10439238 - -
          Affymetrix Mouse Gene 1.0 ST Array 10479367 - -
          Affymetrix Mouse Gene 1.0 ST Array 10479368 - -
          Affymetrix HT MG-430 PM Array Plate 1433721_PM_x_at - -
          Affymetrix HT MG-430 PM Array Plate 1430288_PM_x_at - -
          Affymetrix HT MG-430 PM Array Plate 1433549_PM_x_at - -
          Affymetrix HT MG-430 PM Array Plate 1434358_PM_x_at - -
          Affymetrix Mouse Gene 1.1 ST Array 10439237 - -
          Affymetrix Mouse Gene 1.1 ST Array 10479362 - -
          Mouse Genome 430 2.0 Array 1433549_x_at - -
          Mouse Genome 430 2.0 Array 1434358_x_at - -
          Mouse Genome 430 2.0 Array 1430288_PM_x_at - -
          Mouse Genome 430 2.0 Array 1433549_PM_x_at - -
          Mouse Genome 430 2.0 Array 1430288_x_at - -
          Mouse Genome 430 2.0 Array 1433721_x_at - -
          Mouse Genome 430 2.0 Array 1433721_PM_x_at - -
          Mouse Genome 430 2.0 Array 1434358_PM_x_at - -
          Affymetrix Mouse Clariom S Array PSR0200022797.mm.1 TC0200002785.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0200022797.mm.1 NM_025587.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0200022797.mm.1 NM_025587.2 Refseq
          Affymetrix Mouse Clariom S Array PSR0200022795.mm.1 TC0200002785.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0200022795.mm.1 NM_025587.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0200022795.mm.1 NM_025587.2 Refseq
          Mouse Expression Set 430 1433549_x_at - -
          Mouse Expression Set 430 1434358_x_at - -
          Mouse Expression Set 430 1439415_x_at - -
          Mouse Expression Set 430 1433549_PM_x_at - -
          Mouse Expression Set 430 1433721_PM_x_at - -
          Mouse Expression Set 430 1434358_PM_x_at - -
          Mouse Expression Set 430 1433721_x_at - -
          Mouse Expression Set 430 1439415_PM_x_at - -
          Mouse Genome 430A 2.0 Array 1434358_PM_x_at - -
          Mouse Genome 430A 2.0 Array 1434358_x_at - -
          Mouse Genome 430A 2.0 Array 1433721_x_at - -
          Mouse Genome 430A 2.0 Array 1433721_PM_x_at - -
          Mouse Genome 430A 2.0 Array 1439415_PM_x_at - -
          Mouse Genome 430A 2.0 Array 1433549_PM_x_at - -
          Mouse Genome 430A 2.0 Array 1439415_x_at - -
          Mouse Genome 430A 2.0 Array 1433549_x_at - -
          Affymetrix HT Mouse Genome 430A Array 1433721_x_at - -
          Affymetrix HT Mouse Genome 430A Array 1433549_x_at - -
          Affymetrix HT Mouse Genome 430A Array 1434358_x_at - -
          Affymetrix HT Mouse Genome 430A Array 1433721_PM_x_at - -
          Affymetrix HT Mouse Genome 430A Array 1433549_PM_x_at - -
          Affymetrix HT Mouse Genome 430A Array 1434358_PM_x_at - -
          Affymetrix HT Mouse Genome 430B Array 1430288_x_at - -
          Affymetrix HT Mouse Genome 430B Array 1430288_PM_x_at - -
          Array Name Probe IDs Targets Target Type
          Illumina mouse-6 v1.0 expression beadchip 3120673 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0003120673 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_1240898 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_1240898 - -
          mouse-6 v1.1 (Illumina) scl066481.4_0-S - -
          Illumina Sentrix mouse-6.v1 bead array 3120673 - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl066481.4_0-S - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 3120673 - -
          MouseRef-8 v2 Expression BeadChip ILMN_1240898 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_1240898 - -
          Illumina Sentrix Mouse-6 Expression BeadChip (3) scl066481.4_0-S - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-6659c69d8f-ztd5s:80/100.66.134.6:80.
          git-commit: 1827ea18100b76eacea45a9906e00cdefce2d263
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.30.1-2025.05.22-1.0