Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm01215490_m1
          See other FGFR1 GE Assays ›
          Gene Symbol:
          Fgfr1
          Gene Name:
          fibroblast growth factor receptor 1
          Gene Aliases:
          AW208770, Eask, FLG, Fgfr-1, Flt-2, Hspy, MFR, bFGF-R-1, c-fgr
          Chromosome Location:
          Chr.8: 25513623 - 25575718 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm01215490_m1
          Size
          Availability Made To Order
          Catalog # 4351372
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Fgfr1
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001079908.2 NP_001073377.1 5 - 6 1374 79
          NM_001079909.2 NP_001073378.1 4 - 5 1113 79
          NM_010206.3 NP_034336.2 5 - 6 1380 79
          GenBank mRNA AF176552.1 - 3 - 4 405 79
          AK028354.1 - 4 - 5 553 79
          AK049704.1 - 5 - 6 1342 79
          BC010200.1 - 5 - 6 1273 3967874 79
          BC033447.1 - 3 - 4 376 4037899 79
          M28998.1 - 4 - 5 678 79
          M33760.1 - 3 - 4 397 79
          M65053.1 - 4 - 5 651 79
          U22324.1 - 4 - 5 678 79
          U23445.1 - 3 - 4 411 79
          X51893.1 - 5 - 6 735 79
          X57790.1 - 4 - 5 615 79

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC0800000321.mm.1

          Set Membership:

          Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Mus musculus Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Fgfr1

          Gene Ontology Categories:

          Function(s) Process(es)

          negative regulation of transcription by RNA polymerase II
          angiogenesis
          ureteric bud development
          in utero embryonic development
          organ induction
          positive regulation of mesenchymal cell proliferation
          chondrocyte differentiation
          protein phosphorylation
          brain development
          salivary gland morphogenesis
          sensory perception of sound
          positive regulation of cell population proliferation
          fibroblast growth factor receptor signaling pathway
          regulation of gene expression
          negative regulation of gene expression
          positive regulation of phospholipase C activity
          positive regulation of neuron projection development
          phosphorylation
          peptidyl-tyrosine phosphorylation
          stem cell population maintenance
          orbitofrontal cortex development
          motogenic signaling involved in postnatal olfactory bulb interneuron migration
          ventricular zone neuroblast division
          central nervous system neuron development
          lung development
          embryonic limb morphogenesis
          midbrain development
          neuron projection development
          fibroblast growth factor receptor signaling pathway involved in orbitofrontal cortex development
          regulation of cell population proliferation
          inner ear morphogenesis
          outer ear morphogenesis
          middle ear morphogenesis
          positive regulation of MAP kinase activity
          positive regulation of MAPK cascade
          positive regulation of neuron differentiation
          negative regulation of osteoblast differentiation
          positive regulation of cell cycle
          positive regulation of transcription by RNA polymerase II
          protein autophosphorylation
          paraxial mesoderm development
          regulation of lateral mesodermal cell fate specification
          cell maturation
          blood vessel morphogenesis
          generation of neurons
          mesenchymal cell differentiation
          regulation of sensory perception of pain
          positive regulation of cardiac muscle cell proliferation
          auditory receptor cell development
          branching involved in salivary gland morphogenesis
          lung-associated mesenchyme development
          regulation of branching involved in salivary gland morphogenesis by mesenchymal-epithelial signaling
          vasculogenesis involved in coronary vascular morphogenesis
          regulation of stem cell proliferation
          positive regulation of MAPKKK cascade by fibroblast growth factor receptor signaling pathway
          regulation of extrinsic apoptotic signaling pathway in absence of ligand
          nucleotide binding
          protein kinase activity
          protein tyrosine kinase activity
          transmembrane receptor protein tyrosine kinase activity
          fibroblast growth factor-activated receptor activity
          protein binding
          ATP binding
          heparin binding
          kinase activity
          transferase activity
          transferase activity, transferring phosphorus-containing groups
          fibroblast growth factor binding
          identical protein binding
          protein homodimerization activity
          protein-containing complex binding
          cell adhesion molecule binding

          MicroArray Probes:

          Affymetrix

          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Clariom S Array PSR0800002373.mm.1 TC0800000321.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0800002373.mm.1 NM_001079908.2 Refseq
          Affymetrix Mouse Clariom S Array PSR0800002373.mm.1 NM_001079909.2 Refseq
          Affymetrix Mouse Clariom S Array PSR0800002373.mm.1 NM_010206.3 Refseq

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-65cb46898d-nlgdw:80/100.66.128.131:80.
          git-commit: e5493f66f4597fc66e516c8a379ca3b6bed48f5b
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.33.0-2025.07.30-1.0