Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Clariom S Array |
PSR1600007666.mm.1 |
TC1600000989.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1600007666.mm.1 |
NM_001199210.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1600007666.mm.1 |
NM_027627.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1600007665.mm.1 |
TC1600000989.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1600007665.mm.1 |
NM_001199210.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1600007665.mm.1 |
NM_027627.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1600007667.mm.1 |
TC1600000989.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1600007667.mm.1 |
NM_001199210.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1600007667.mm.1 |
NM_027627.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1600007670.mm.1 |
TC1600000989.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1600007670.mm.1 |
NM_001199210.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1600007670.mm.1 |
NM_027627.3 |
Refseq |