Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm01215595_m1
          See other MRAP GE Assays ›
          Gene Symbol:
          Mrap
          Gene Name:
          melanocortin 2 receptor accessory protein
          Gene Aliases:
          1110025G12Rik, Falp, ORF61
          Chromosome Location:
          Chr.16: 90738324 - 90749776 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm01215595_m1
          Size
          Availability Made To Order
          Catalog # 4351372
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Mrap
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_029844.3 NP_084120.1 2 - 3 451 94
          GenBank mRNA AK003912.2 - 2 - 3 452 94
          AK158113.1 - 2 - 3 325 94
          AY079153.1 - 2 - 3 228 94
          BC027543.1 - 2 - 3 305 1477097 94

          Back To Top

          More Information


          Set Membership:

          Ambion siRNA available Amplicon length between 86 and 100 3' Most Mammalian Gene Collection Mus musculus 1700 Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Mrap

          Gene Ontology Categories:

          Function(s) Process(es)

          protein localization to cell surface
          brown fat cell differentiation
          corticotropin hormone receptor binding
          type 3 melanocortin receptor binding
          type 4 melanocortin receptor binding
          type 5 melanocortin receptor binding
          type 1 melanocortin receptor binding

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P346964 AK158113.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P346964 AY079153.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P346964 AK003912.2 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P346964 BC027543.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P346964 NM_029844.3 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P346964 AK158113.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P346964 AY079153.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P346964 AK003912.2 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P346964 BC027543.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P346964 NM_029844.3 mouse_transcriptome
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 414078 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 877090 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 709665 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 581822 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 20269 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 587506 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 138859 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P162400 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P116316 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P346964 AK158113.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P346964 AY079153.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P346964 AK003912.2 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P346964 BC027543.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P346964 NM_029844.3 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P346964 AK158113.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P346964 AY079153.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P346964 AK003912.2 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P346964 BC027543.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P346964 NM_029844.3 mouse_transcriptome
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30958589 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30958596 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30958595 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30958582 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30958587 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30958585 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30958584 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30958590 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30958588 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30958586 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30958598 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30958594 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P116316 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P162400 - -
          Whole Mouse Genome G4122A A_51_P346964 AK158113.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P346964 AY079153.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P346964 AK003912.2 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P346964 BC027543.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P346964 NM_029844.3 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P346964 AK158113.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P346964 AY079153.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P346964 AK003912.2 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P346964 BC027543.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P346964 NM_029844.3 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P112550 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P162400 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P116316 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P12677554 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P12677547 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P12677548 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P12677561 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P12677550 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30958603 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30958596 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30958660 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30958586 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30958671 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30958704 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30958701 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30958585 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30958717 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30958554 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30958711 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30958667 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30958608 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30958572 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30958587 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30958575 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30958633 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30958727 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30958576 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30958656 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30958665 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30958681 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30958631 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30958574 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30958696 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30958702 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30958690 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30958591 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30958619 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30958614 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30958719 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30958615 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30958652 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30958693 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30958555 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30958668 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30958565 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30958722 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30958647 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30958611 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30958617 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30958600 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30958610 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30958645 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30958549 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30958692 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30958649 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30958594 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30958550 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30958683 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30958638 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30958659 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30958582 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30958558 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30958553 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30958635 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30958694 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30958685 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30958584 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30958595 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30958700 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30958622 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30958678 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30958650 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30958680 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30958568 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30958687 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30958691 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30958675 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30958552 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30958714 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30958716 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30958570 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30958707 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30958569 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30958651 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30958556 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30958646 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30958598 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30958607 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30958605 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30958715 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30958698 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30958658 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30958654 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30958673 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30958621 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30958674 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30958708 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30958686 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30958629 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30958662 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30958585 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30958582 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P12775546 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30958610 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30958591 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P12677547 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P12855084 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30958614 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30958596 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30958594 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P12677554 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30958576 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P12726138 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30958586 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30958600 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30958619 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30958572 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30958589 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30958579 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30958570 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30958624 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P12868876 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30958590 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30958612 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30958605 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30958603 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30958575 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30958595 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P12727282 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30958587 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P12861786 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30958601 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30958569 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30958608 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30958604 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P12619610 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30958574 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30958602 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30958564 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30958617 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30958607 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30958565 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30958568 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P12677550 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P12619606 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P12775549 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P13013546 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30958598 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30958625 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10436806 - -
          Affymetrix Mouse Gene 1.0 ST Array 10436808 - -
          Affymetrix Mouse Gene 1.0 ST Array 10436805 - -
          Affymetrix Mouse Gene 1.0 ST Array 10436807 - -
          Affymetrix HT MG-430 PM Array Plate 1451966_PM_at - -
          Affymetrix HT MG-430 PM Array Plate 1451371_PM_at - -
          Affymetrix Mouse Gene 1.1 ST Array 10436804 - -
          Mouse Genome 430 2.0 Array 1451371_PM_at - -
          Mouse Genome 430 2.0 Array 1451966_at - -
          Mouse Genome 430 2.0 Array 1451371_at - -
          Mouse Genome 430 2.0 Array 1451966_PM_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 77037_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 77037_PM_at - -
          Mouse Expression Set 430 1451371_PM_at - -
          Mouse Expression Set 430 1451966_PM_at - -
          Mouse Expression Set 430 1451371_at - -
          Mouse Expression Set 430 1451966_at - -
          Murine Genome U74v2 Set 104325_at - -
          Murine Genome U74v2 Set 104325_PM_at - -
          Murine Genome U74v2 Set 171065_PM_at - -
          Murine Genome U74v2 Set 171065_at - -
          Mouse Genome 430A 2.0 Array 1451966_PM_at - -
          Mouse Genome 430A 2.0 Array 1451966_at - -
          Mouse Genome 430A 2.0 Array 1451371_PM_at - -
          Mouse Genome 430A 2.0 Array 1451371_at - -
          Affymetrix HT Mouse Genome 430A Array 1451371_PM_at - -
          Affymetrix HT Mouse Genome 430A Array 1451966_PM_at - -
          Affymetrix HT Mouse Genome 430A Array 1451371_at - -
          Affymetrix HT Mouse Genome 430A Array 1451966_at - -
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 752295 - -
          Array Name Probe IDs Targets Target Type
          MouseWG-6 v2 Expression BeadChip 2940114 - -
          Illumina mouse-6 v1.0 expression beadchip 2940114 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0002940114 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_1234241 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_1234241 - -
          mouse-6 v1.1 (Illumina) scl0001829.1_310-S - -
          Illumina Sentrix mouse-6.v1 bead array 2940114 - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl0001829.1_310-S - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 2940114 - -
          MouseRef-8 v2 Expression BeadChip ILMN_1234241 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_1234241 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-865d7754c7-mcq8j:80/100.66.130.62:80.
          git-commit: 7ff36f61b459eb8f02ba5a263f0e3ce367ca82d3
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.32.0-2025.07.28-1.0