Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm01215770_g1
          See other TIGD3 GE Assays ›
          Gene Symbol:
          Tigd3
          Gene Name:
          tigger transposable element derived 3
          Gene Aliases:
          -
          Chromosome Location:
          Chr.19: 5891138 - 5895166 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm01215770_g1
          Size
          Availability Made To Order
          Catalog # 4351372
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Tigd3
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_198634.1 NP_941036.1 1 - 2 138 92
          GenBank mRNA BC055784.1 - 1 - 2 138 6404652 92

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC1900000989.mm.1

          Additional Information:

          This assay is in the 5'UTR region of the BC055784.1, NM_198634.1 transcripts.

          Set Membership:

          Ambion siRNA available Amplicon length between 86 and 100 3' Most Mammalian Gene Collection Detect Genomic DNA Mus musculus 1700 Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Tigd3

          Gene Ontology Categories:

          Function(s) Process(es)

          biological_process
          molecular_function
          nucleic acid binding
          DNA binding

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P31931518 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P31931516 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P31931517 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P31931515 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P31931514 - -
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2021029 NM_198634.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2021029 BC053500.2 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2021029 BC055784.1 mouse_transcriptome
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 569830 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 82492 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P175946 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2021029 NM_198634.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2021029 BC053500.2 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2021029 BC055784.1 mouse_transcriptome
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31931523 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31931532 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31931518 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31931497 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31931516 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31931512 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31931510 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31931527 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31931534 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31931529 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31931507 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31931511 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31931517 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31931528 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31931521 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31931503 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31931502 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31931525 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31931505 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31931498 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31931514 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31931515 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31931536 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31931500 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31931531 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P175946 - -
          Whole Mouse Genome G4122A A_52_P82636 NM_198634.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P82636 BC053500.2 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P82636 BC055784.1 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P102493 - -
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P103154 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P175946 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P14392395 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P14392399 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P14392419 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P14392408 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P14392396 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P14392417 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P31931518 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P31931516 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P31931515 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P31931514 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P31931517 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31931442 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31931498 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31931453 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31931532 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31931480 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31931482 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31931500 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31931503 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31931505 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31931463 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31931469 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31931487 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31931449 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31931502 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31931521 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31931465 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31931459 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31931493 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31931450 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31931510 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31931507 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31931494 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31931461 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31931467 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31931525 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31931529 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31931514 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31931444 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31931512 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31931488 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31931535 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31931497 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31931517 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31931527 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31931443 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31931536 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31931447 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31931523 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31931480 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31931484 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31931482 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P14392405 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31931510 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P14392421 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P14225893 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31931488 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31931483 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31931535 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31931512 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31931523 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31931494 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31931508 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P14392396 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31931525 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31931529 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31931536 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P14392422 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31931497 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31931498 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31931503 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31931527 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31931490 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31931514 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31931502 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31931487 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31931521 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31931531 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10465261 - -
          Affymetrix Mouse Gene 1.0 ST Array 10465262 - -
          Affymetrix HT MG-430 PM Array Plate 1447540_PM_at - -
          Affymetrix Mouse Gene 1.1 ST Array 10465260 - -
          Mouse Genome 430 2.0 Array 1447540_at - -
          Mouse Genome 430 2.0 Array 1447540_PM_at - -
          Affymetrix Mouse Clariom S Array PSR1900009021.mm.1 TC1900000989.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1900009021.mm.1 NM_198634.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1900009021.mm.1 NM_198634.1 Refseq
          Mouse Expression Set 430 1447540_at - -
          Mouse Expression Set 430 1447540_PM_at - -
          Murine Genome U74v2 Set 138525_at - -
          Murine Genome U74v2 Set 138525_PM_at - -
          Affymetrix HT Mouse Genome 430B Array 1447540_at - -
          Affymetrix HT Mouse Genome 430B Array 1447540_PM_at - -
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 372121 - -
          Array Name Probe IDs Targets Target Type
          MouseWG-6 v2 Expression BeadChip 3850576 - -
          Illumina mouse-6 v1.0 expression beadchip 3850576 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0003850576 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_2742638 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_2742638 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_2525968 - -
          mouse-6 v1.1 (Illumina) scl0332359.2_2-S - -
          Illumina Sentrix mouse-6.v1 bead array 3850576 - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl0332359.2_2-S - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 3850576 - -
          MouseRef-8 v2 Expression BeadChip ILMN_1254037 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_1254037 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-786bfc498d-47c8f:80/100.66.130.62:80.
          git-commit: 9eb01563f6748445c58f67612c67ecf6b1da7421
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.32.0-2025.07.28-1.0