Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm01216082_gH
          See other FGFR3 GE Assays ›
          Gene Symbol:
          Fgfr3
          Gene Name:
          fibroblast growth factor receptor 3
          Gene Aliases:
          CD333, FR3, Fgfr-3, Flg-2, HBGFR, Mfr3, sam3
          Chromosome Location:
          Chr.5: 33721724 - 33737068 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm01216082_gH
          Size
          Availability Made To Order
          Catalog # 4351372
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Fgfr3
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001163215.2 NP_001156687.1 10 - 11 1732 66
          NM_001163216.2 NP_001156688.1 8 - 9 1447 66
          NM_001163217.2 NP_001156689.1 9 - 10 1507 66
          NM_001205270.1 NP_001192199.1 10 - 11 1709 66
          NM_008010.5 NP_032036.2 10 - 11 1706 66
          GenBank mRNA AF024638.1 - 8 - 9 1355 66
          AK143592.1 - 10 - 11 1642 66
          AK163495.1 - 8 - 9 1285 66
          BC053056.1 - 10 - 11 1616 5708838 66
          M81342.1 - 10 - 11 1624 66
          S56291.1 - 9 - 10 1451 66
          X58255.1 - 10 - 11 1610 66

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC0500000361.mm.1

          Set Membership:

          Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Fgfr3

          Gene Ontology Categories:

          Function(s) Process(es)

          negative regulation of transcription by RNA polymerase II
          MAPK cascade
          skeletal system development
          positive regulation of endothelial cell proliferation
          morphogenesis of an epithelium
          lens morphogenesis in camera-type eye
          protein phosphorylation
          apoptotic process
          cell-cell signaling
          positive regulation of cell population proliferation
          negative regulation of cell population proliferation
          fibroblast growth factor receptor signaling pathway
          positive regulation of phospholipase activity
          oligodendrocyte development
          phosphorylation
          peptidyl-tyrosine phosphorylation
          substantia nigra development
          central nervous system myelination
          cell differentiation
          bone mineralization
          forebrain development
          positive regulation of protein ubiquitination
          somatic stem cell population maintenance
          post-anal tail morphogenesis
          positive regulation of tyrosine phosphorylation of STAT protein
          positive regulation of MAPK cascade
          positive regulation of neuron apoptotic process
          positive regulation of phosphatidylinositol 3-kinase activity
          positive regulation of cell differentiation
          negative regulation of mitotic nuclear division
          negative regulation of smoothened signaling pathway
          protein autophosphorylation
          digestive tract morphogenesis
          negative regulation of developmental growth
          response to axon injury
          negative regulation of astrocyte differentiation
          inner ear development
          negative regulation of epithelial cell proliferation
          positive regulation of peptidyl-tyrosine phosphorylation
          cartilage development
          inner ear receptor cell differentiation
          bone morphogenesis
          axonogenesis involved in innervation
          alveolar secondary septum development
          lens fiber cell development
          positive regulation of ERK1 and ERK2 cascade
          bone maturation
          epithelial cell fate commitment
          positive regulation of MAPKKK cascade by fibroblast growth factor receptor signaling pathway
          cochlea development
          positive regulation of canonical Wnt signaling pathway
          fibroblast growth factor receptor apoptotic signaling pathway
          nucleotide binding
          protein kinase activity
          protein tyrosine kinase activity
          transmembrane receptor protein tyrosine kinase activity
          fibroblast growth factor-activated receptor activity
          protein binding
          ATP binding
          kinase activity
          transferase activity
          transferase activity, transferring phosphorus-containing groups
          fibroblast growth factor binding

          MicroArray Probes:

          Affymetrix

          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Clariom S Array PSR0500003416.mm.1 TC0500000361.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0500003416.mm.1 NM_001163215.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0500003416.mm.1 NM_001163215.2 Refseq
          Affymetrix Mouse Clariom S Array PSR0500003416.mm.1 NM_001163216.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0500003416.mm.1 NM_001163216.2 Refseq
          Affymetrix Mouse Clariom S Array PSR0500003416.mm.1 NM_001163217.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0500003416.mm.1 NM_001163217.2 Refseq
          Affymetrix Mouse Clariom S Array PSR0500003416.mm.1 NM_001205270.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0500003416.mm.1 NM_001205270.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0500003416.mm.1 NM_008010.5.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0500003416.mm.1 NM_008010.5 Refseq
          Affymetrix Mouse Clariom S Array PSR0500003415.mm.1 TC0500000361.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0500003415.mm.1 NM_001163215.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0500003415.mm.1 NM_001163215.2 Refseq
          Affymetrix Mouse Clariom S Array PSR0500003415.mm.1 NM_001163216.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0500003415.mm.1 NM_001163216.2 Refseq
          Affymetrix Mouse Clariom S Array PSR0500003415.mm.1 NM_001163217.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0500003415.mm.1 NM_001163217.2 Refseq
          Affymetrix Mouse Clariom S Array PSR0500003415.mm.1 NM_001205270.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0500003415.mm.1 NM_001205270.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0500003415.mm.1 NM_008010.5.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0500003415.mm.1 NM_008010.5 Refseq

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-74594944d-pk6zc:80/100.66.130.62:80.
          git-commit: ab2b2261c2971e97b1ae408e3ad10520b8413932
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.31.0-2025.06.23-1.0