Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm01216171_m1
          See other CCR5 GE Assays ›
          Gene Symbol:
          Ccr5
          Gene Name:
          chemokine (C-C motif) receptor 5
          Gene Aliases:
          AM4-7, CD195, Cmkbr5
          Chromosome Location:
          Chr.9: 124121543 - 124127183 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm01216171_m1
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Ccr5
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_009917.5 NP_034047.2 1 - 2 93 78
          GenBank mRNA AK141906.1 - 1 - 2 64 78
          AK154595.1 - 1 - 2 94 78
          AK155628.1 - 1 - 2 67 78
          AK155632.1 - 1 - 2 64 78
          AK155867.1 - 1 - 2 58 78
          BY198155.1 - 1 - 2 94 78
          D83648.1 - 1 - 2 63 78

          Back To Top

          More Information


          Additional Information:

          This assay is in the 5'UTR region of the AK141906.1, AK154595.1, AK155628.1, AK155867.1, D83648.1, NM_009917.5 transcripts.

          Set Membership:

          Ambion siRNA available Amplicon length between 71 and 85 G-protein coupled receptor Mus musculus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Ccr5

          Gene Ontology Categories:

          Function(s) Process(es)

          calcium ion transport
          chemotaxis
          defense response
          inflammatory response
          immune response
          signal transduction
          G-protein coupled receptor signaling pathway
          positive regulation of cytosolic calcium ion concentration
          cell-cell signaling
          positive regulation of gene expression
          release of sequestered calcium ion into cytosol by sarcoplasmic reticulum
          calcium-mediated signaling
          positive regulation of cell-cell adhesion
          negative regulation of cell migration
          negative regulation of axon extension
          positive regulation of fever generation
          positive regulation of interleukin-6 production
          positive regulation of tumor necrosis factor production
          positive regulation of neuron differentiation
          positive regulation of interleukin-1 beta secretion
          positive regulation of inflammatory response
          positive regulation of apoptotic process by virus
          chemokine-mediated signaling pathway
          response to cholesterol
          neuron death
          positive regulation of cell migration involved in sprouting angiogenesis
          negative regulation of macrophage apoptotic process
          negative regulation of neural precursor cell proliferation
          glycoprotein binding
          actin binding
          signal transducer activity
          G-protein coupled receptor activity
          chemokine receptor activity
          C-C chemokine receptor activity
          protein kinase binding
          C-C chemokine binding
          chemokine (C-C motif) ligand 5 binding

          MicroArray Probes:

          Affymetrix ABI

          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Clariom S Array PSR0900014086.mm.1 NM_009917.5.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0900014086.mm.1 NM_009917.5 Refseq
          Affymetrix Mouse Clariom S Array PSR0900014086.mm.1 TC0900001675.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0900014085.mm.1 NM_009917.5.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0900014085.mm.1 NM_009917.5 Refseq
          Affymetrix Mouse Clariom S Array PSR0900014085.mm.1 TC0900001675.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0900014088.mm.1 NM_009917.5.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0900014088.mm.1 NM_009917.5 Refseq
          Affymetrix Mouse Clariom S Array PSR0900014088.mm.1 TC0900001675.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0900014087.mm.1 NM_009917.5.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0900014087.mm.1 NM_009917.5 Refseq
          Affymetrix Mouse Clariom S Array PSR0900014087.mm.1 TC0900001675.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0900014078.mm.1 NM_009917.5.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0900014078.mm.1 NM_009917.5 Refseq
          Affymetrix Mouse Clariom S Array PSR0900014078.mm.1 TC0900001675.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0900014083.mm.1 NM_009917.5.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0900014083.mm.1 NM_009917.5 Refseq
          Affymetrix Mouse Clariom S Array PSR0900014083.mm.1 TC0900001675.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0900014082.mm.1 NM_009917.5.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0900014082.mm.1 NM_009917.5 Refseq
          Affymetrix Mouse Clariom S Array PSR0900014082.mm.1 TC0900001675.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0900014084.mm.1 NM_009917.5.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0900014084.mm.1 NM_009917.5 Refseq
          Affymetrix Mouse Clariom S Array PSR0900014084.mm.1 TC0900001675.mm.1 MTA TC
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 325323 - -
          ABI Mouse Genome Survey Microarray 847789 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-74594944d-pk6zc:80/100.66.130.62:80.
          git-commit: ab2b2261c2971e97b1ae408e3ad10520b8413932
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.31.0-2025.06.23-1.0