Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm01216286_g1
          See other ATRAID GE Assays ›
          Gene Symbol:
          Atraid
          Gene Name:
          all-trans retinoic acid induced differentiation factor
          Gene Aliases:
          0610007C21Rik, AI316792, Apr3, HSPC013, p18
          Chromosome Location:
          Chr.5: 31048640 - 31054633 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm01216286_g1
          Size
          Availability Made To Order
          Catalog # 4351372
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Atraid
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_027855.4 NP_082131.2 6 - 7 603 81
          NM_212470.3 NP_997635.1 6 - 7 506 81
          GenBank mRNA AB009018.1 - 6 - 7 586 81
          AK002276.1 - 6 - 7 579 81
          AK151307.1 - 6 - 7 597 81
          AK151366.1 - 6 - 7 597 81
          AK158882.1 - 4 - 5 1345 81
          AK169267.1 - 6 - 7 580 81
          BC049637.1 - 6 - 7 487 6585654 81
          BC057097.1 - 6 - 7 576 5714599 81

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC0500000310.mm.1

          Set Membership:

          Ambion siRNA available Amplicon length between 71 and 85 3' Most Mammalian Gene Collection Detect Genomic DNA Mus musculus 1700 Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Atraid

          Gene Ontology Categories:

          Function(s) Process(es)

          regulation of gene expression
          cell differentiation
          positive regulation of bone mineralization
          negative regulation of osteoblast proliferation
          positive regulation of osteoblast differentiation
          negative regulation of cellular protein catabolic process
          molecular_function

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P23576138 - -
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P1991552 BC049637.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P1991552 BC057097.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P1991552 AK169267.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P1991552 AK002276.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P1991552 AK151307.1 mouse_transcriptome
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P100908 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P1991552 BC049637.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P1991552 BC057097.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P1991552 AK169267.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P1991552 AK002276.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P1991552 AK151307.1 mouse_transcriptome
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23576140 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23576158 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23576159 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23576165 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23576154 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23576168 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23576148 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23576145 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23576153 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23576167 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23576164 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23576147 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23576160 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23576143 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23576155 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23576157 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23576151 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23576138 - -
          Whole Mouse Genome G4122A A_51_P256759 BC049637.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P256759 BC057097.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P256759 AK169267.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P256759 AK002276.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P256759 AK151307.1 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P121253 - -
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P133098 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P04131660 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P04385373 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P04604475 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P04604477 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P04385366 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P04604472 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P04131661 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P04385374 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P04604476 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P04385375 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P23576138 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23576200 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23576174 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23576223 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23576221 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23576164 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23576159 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23576188 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23576153 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23576197 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23576198 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23576240 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23576193 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23576192 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23576234 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23576202 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23576235 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23576143 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23576211 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23576209 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23576206 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23576175 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23576184 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23576215 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23576204 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23576219 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23576232 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23576155 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23576138 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23576186 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23576216 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23576208 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23576162 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23576213 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23576170 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23576178 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23576236 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23576167 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23576195 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23576177 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23576207 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23576157 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23576173 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23576158 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23576180 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23576194 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23576160 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23576218 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23576190 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23576145 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23576239 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23576168 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23576225 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23576231 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23576165 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P04131661 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23576147 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23576162 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P04604479 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23576161 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P04270071 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P04270077 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23576153 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23576160 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P04604473 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23576140 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P04604478 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23576148 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23576168 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P04385366 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P04497115 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P04270083 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P04385375 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23576157 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23576150 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23576167 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10520643 - -
          Affymetrix Mouse Gene 1.0 ST Array 10520644 - -
          Affymetrix Mouse Gene 1.0 ST Array 10520646 - -
          Affymetrix Mouse Gene 1.0 ST Array 10520645 - -
          Affymetrix Mouse Gene 1.0 ST Array 10520641 - -
          Affymetrix Mouse Gene 1.0 ST Array 10520642 - -
          Affymetrix Mouse Gene 1.0 ST Array 10520647 - -
          Affymetrix HT MG-430 PM Array Plate 1428380_PM_at - -
          Affymetrix Mouse Gene 1.1 ST Array 10520638 - -
          Mouse Genome 430 2.0 Array 1428380_at - -
          Mouse Genome 430 2.0 Array 1428380_PM_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 381629_PM_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 381629_at - -
          Affymetrix Mouse Clariom S Array PSR0500002654.mm.1 TC0500000310.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0500002654.mm.1 NM_027855.4 Refseq
          Affymetrix Mouse Clariom S Array PSR0500002654.mm.1 NM_212470.3 Refseq
          Affymetrix Mouse Clariom S Array PSR0500002653.mm.1 TC0500000310.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0500002653.mm.1 NM_027855.4 Refseq
          Affymetrix Mouse Clariom S Array PSR0500002653.mm.1 NM_212470.3 Refseq
          Affymetrix Mouse Clariom S Array PSR0500002655.mm.1 TC0500000310.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0500002655.mm.1 NM_027855.4 Refseq
          Affymetrix Mouse Clariom S Array PSR0500002655.mm.1 NM_212470.3 Refseq
          Affymetrix Mouse Clariom S Array PSR0500002656.mm.1 TC0500000310.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0500002656.mm.1 NM_027855.4 Refseq
          Affymetrix Mouse Clariom S Array PSR0500002656.mm.1 NM_212470.3 Refseq
          Mouse Expression Set 430 1428380_PM_at - -
          Mouse Expression Set 430 1428380_at - -
          Murine Genome U74v2 Set 160271_at - -
          Murine Genome U74v2 Set 160271_PM_at - -
          Mouse Genome 430A 2.0 Array 1428380_PM_at - -
          Mouse Genome 430A 2.0 Array 1428380_at - -
          Affymetrix HT Mouse Genome 430A Array 1428380_PM_at - -
          Affymetrix HT Mouse Genome 430A Array 1428380_at - -
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 918064 - -
          Array Name Probe IDs Targets Target Type
          MouseWG-6 v2 Expression BeadChip 102260551 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_2537239 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_1230777 - -
          MouseRef-8 v2 Expression BeadChip ILMN_3033922 - -
          MouseRef-8 v2 Expression BeadChip ILMN_3092673 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_3033922 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_3092673 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-6467d75d44-xp7n2:80/100.66.130.62:80.
          git-commit: 8ea1bebb48d33b8b7c0e2103582aa482e6db8664
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.32.0-2025.07.28-1.0