Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm01217349_m1
          See other NDUFAF6 GE Assays ›
          Gene Symbol:
          Ndufaf6
          Gene Name:
          NADH dehydrogenase (ubiquinone) complex I, assembly factor 6
          Gene Aliases:
          2310030N02Rik
          Chromosome Location:
          Chr.4: 11051042 - 11076235 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm01217349_m1
          Size
          Availability Made To Order
          Catalog # 4351372
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Ndufaf6
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001085493.2 NP_001078962.1 8 - 9 898 109
          GenBank mRNA AK009547.1 - 8 - 9 879 109
          BC048539.1 - 2 - 3 148 109

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC0400002200.mm.1

          Set Membership:

          Amplicon length greater than or equal to 101 3' Most Mammalian Gene Collection Mus musculus 1700 Probe spans exons

          Gene Ontology Categories:

          Function(s) Process(es)

          biosynthetic process
          mitochondrial respiratory chain complex I assembly
          molecular_function
          transferase activity

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P150525 NM_001085493.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P150525 AK009547.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P150525 BC048539.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P150525 NM_001085493.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P150525 AK009547.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P150525 BC048539.1 mouse_transcriptome
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 191942 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 936890 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 483936 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 913756 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 787548 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 854254 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 70096 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 195842 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 237367 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 765178 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 196657 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 16678 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 935228 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 844973 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 875954 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 157939 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 8549 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 61900 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P145689 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P150525 NM_001085493.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P150525 AK009547.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P150525 BC048539.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P150525 NM_001085493.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P150525 AK009547.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P150525 BC048539.1 mouse_transcriptome
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P05155144 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P145689 - -
          Whole Mouse Genome G4122A A_52_P150525 NM_001085493.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P150525 AK009547.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P150525 BC048539.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P150525 NM_001085493.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P150525 AK009547.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P150525 BC048539.1 mouse_transcriptome
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P145689 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22727798 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22727827 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22727874 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22727925 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22727770 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22727838 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22727893 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22727968 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22727944 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22727858 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22727954 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22727901 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22727942 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22727960 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22727876 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22727950 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22727782 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22727835 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22727756 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22727758 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22727757 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22727744 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22727866 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22727935 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22727921 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22727862 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22727963 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22727752 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22727829 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22727933 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22727749 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22727822 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22727924 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22727965 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22727839 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22727955 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22727898 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22727860 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22727745 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22727890 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22727850 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22727907 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22727922 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22727797 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22727946 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22727826 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22727780 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22727852 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22727776 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22727841 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22727751 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22727917 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22727878 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22727962 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22727964 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22727849 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22727773 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22727956 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22727904 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22727766 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22727761 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22727884 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22727790 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22727796 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22727832 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22727792 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22727919 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22727784 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22727937 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22727851 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22727945 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22727929 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22727789 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22727886 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22727892 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22727930 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22727879 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22727952 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22727742 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22727910 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22727762 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22727908 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22727913 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22727747 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22727741 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22727939 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22727916 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22727903 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22727857 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22727966 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22727778 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22727787 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22727781 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22727897 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22727738 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22727791 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22727941 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22727902 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22727772 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03816651 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22727766 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22727760 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22727891 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22727949 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03491960 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03491968 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03816655 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03727703 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03758283 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22727965 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03551055 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22727933 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22727958 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22727761 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22727919 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03444920 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03695735 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22727911 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03758280 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22727773 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22727944 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03618383 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22727920 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22727913 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22727755 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03879974 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22727897 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22727939 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03429042 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22727768 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22727937 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03352281 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03488119 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22727952 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22727968 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03491964 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22727948 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22727763 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03879971 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03368005 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03652251 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22727951 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22727930 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03546909 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03429619 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22727916 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22727764 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22727907 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22727886 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22727903 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03393788 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03444919 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22727775 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03491962 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03551059 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22727904 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22727890 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22727956 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22727954 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22727942 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03551057 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P04044935 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03305261 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22727888 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03758277 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03816647 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22727966 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22727899 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03879969 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03816653 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22727945 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22727912 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22727946 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22727963 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03491967 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22727909 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22727932 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22727955 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22727915 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22727893 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10511505 - -
          Affymetrix Mouse Gene 1.0 ST Array 10511504 - -
          Affymetrix Mouse Gene 1.0 ST Array 10511508 - -
          Affymetrix Mouse Gene 1.0 ST Array 10511506 - -
          Affymetrix Mouse Gene 1.0 ST Array 10511503 - -
          Affymetrix Mouse Gene 1.0 ST Array 10511509 - -
          Affymetrix Mouse Gene 1.0 ST Array 10511502 - -
          Affymetrix Mouse Gene 1.0 ST Array 10511507 - -
          Affymetrix HT MG-430 PM Array Plate 1429213_PM_at - -
          Affymetrix Mouse Gene 1.1 ST Array 10511501 - -
          Mouse Genome 430 2.0 Array 1429213_at - -
          Mouse Genome 430 2.0 Array 1429213_PM_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 76947_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 76947_PM_at - -
          Affymetrix Mouse Clariom S Array PSR0400018337.mm.1 TC0400002200.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0400018337.mm.1 NM_001085493.2 Refseq
          Affymetrix Mouse Clariom S Array PSR0400018339.mm.1 TC0400002200.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0400018339.mm.1 NM_001085493.2 Refseq
          Mouse Expression Set 430 1429213_at - -
          Mouse Expression Set 430 1429213_PM_at - -
          Murine Genome U74v2 Set 165986_PM_at - -
          Murine Genome U74v2 Set 165986_at - -
          Affymetrix HT Mouse Genome 430B Array 1429213_at - -
          Affymetrix HT Mouse Genome 430B Array 1429213_PM_at - -
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 631876 - -
          Array Name Probe IDs Targets Target Type
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_2656042 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_2656042 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-6c59565f7c-q2rrq:80/100.66.130.62:80.
          git-commit: 653a2953ec6bb2e3dd1e37289b36fc4e0ab15bfe
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.31.0-Offline