Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Clariom S Array |
PSR0400000126.mm.1 |
TC0400000018.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0400000126.mm.1 |
NM_001190322.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400000126.mm.1 |
NM_001285804.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400000126.mm.1 |
NM_181391.4 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400000126.mm.1 |
NR_104354.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400000126.mm.1 |
NR_104355.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400000126.mm.1 |
NR_104357.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400000125.mm.1 |
TC0400000018.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0400000125.mm.1 |
NM_001190322.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400000125.mm.1 |
NM_001285804.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400000125.mm.1 |
NM_181391.4 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400000125.mm.1 |
NR_104354.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400000125.mm.1 |
NR_104355.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400000125.mm.1 |
NR_104357.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400000131.mm.1 |
TC0400000018.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0400000131.mm.1 |
NM_001190322.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400000131.mm.1 |
NM_001190323.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400000131.mm.1 |
NM_001190324.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400000131.mm.1 |
NM_001285804.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400000131.mm.1 |
NM_181391.4 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400000131.mm.1 |
NR_104354.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400000131.mm.1 |
NR_104355.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400000131.mm.1 |
NR_104356.1 |
Refseq |