Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm01218169_g1
          See other HDGF GE Assays ›
          Gene Symbol:
          Hdgf
          Gene Name:
          hepatoma-derived growth factor
          Gene Aliases:
          AI118077, D3Ertd299e
          Chromosome Location:
          Chr.3: 87906321 - 87916132 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm01218169_g1
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Hdgf
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_008231.4 NP_032257.3 4 - 5 796 70
          GenBank mRNA AF251787.1 - 4 - 5 709 70
          AK005359.1 - 4 - 5 575 70
          AK017863.1 - 4 - 5 750 70
          AK029475.1 - 4 - 5 540 70
          AK076021.1 - 4 - 5 584 70
          BC005713.1 - 4 - 5 706 2609261 70
          BC021654.1 - 4 - 5 724 4163897 70
          D63707.1 - 4 - 5 550 70

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC0300000707.mm.1

          Set Membership:

          Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Mus musculus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Hdgf

          Gene Ontology Categories:

          Function(s) Process(es)

          negative regulation of transcription by RNA polymerase II
          transcription, DNA-templated
          regulation of transcription, DNA-templated
          transcription by RNA polymerase II
          cellular process
          protein localization to nucleus
          positive regulation of transcription by RNA polymerase II
          nucleotide binding
          RNA polymerase II cis-regulatory region sequence-specific DNA binding
          transcription corepressor binding
          DNA binding
          transcription corepressor activity
          RNA binding
          protein binding
          growth factor activity
          heparin binding

          MicroArray Probes:

          Affymetrix ABI

          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Clariom S Array PSR0300004743.mm.1 TC0300000707.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0300004743.mm.1 NM_008231.4.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0300004743.mm.1 NM_008231.4 Refseq
          Affymetrix Mouse Clariom S Array PSR0300004752.mm.1 TC0300000707.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0300004752.mm.1 NM_008231.4.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0300004752.mm.1 NM_008231.4 Refseq
          Affymetrix Mouse Clariom S Array PSR0300004741.mm.1 TC0300000707.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0300004741.mm.1 NM_008231.4.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0300004741.mm.1 NM_008231.4 Refseq
          Affymetrix Mouse Clariom S Array PSR0300004748.mm.1 TC0300000707.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0300004748.mm.1 NM_008231.4.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0300004748.mm.1 NM_008231.4 Refseq
          Affymetrix Mouse Clariom S Array PSR0300004749.mm.1 TC0300000707.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0300004749.mm.1 NM_008231.4.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0300004749.mm.1 NM_008231.4 Refseq
          Affymetrix Mouse Clariom S Array PSR0300004754.mm.1 TC0300000707.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0300004754.mm.1 NM_008231.4.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0300004754.mm.1 NM_008231.4 Refseq
          Affymetrix Mouse Clariom S Array PSR0300004755.mm.1 TC0300000707.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0300004755.mm.1 NM_008231.4.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0300004755.mm.1 NM_008231.4 Refseq
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 911925 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-74594944d-pk6zc:80/100.66.130.62:80.
          git-commit: ab2b2261c2971e97b1ae408e3ad10520b8413932
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.31.0-2025.06.23-1.0