Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm01218646_m1
          See other 1700027F09RIK GE Assays ›
          Gene Symbol:
          1700027F09Rik
          Gene Name:
          RIKEN cDNA 1700027F09 gene
          Gene Aliases:
          -
          Chromosome Location:
          Chr.5: 64454813 - 64467942 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm01218646_m1
          Size
          Availability Made To Order
          Catalog # 4426961
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: 1700027F09Rik
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NR_040681.1 2 - 3 295 68
          GenBank mRNA AK006413.1 - 2 - 3 295 68

          Adjacent Gene Details

          Gene name Gene Symbol Alias Distance Relative Direction Location Chromsome
          Upstream TBC1 domain family,
          member 1
          Tbc1d1 1110062G02Rik,
          AI385682,
          AW555803,
          Tbc1,
          mKIAA1108
          -103.3kb Anti-sense Chr. 5 - 64156310 - 64351486 on NCBI Build GRCm38
          Downstream RIKEN cDNA C030018K13 gene C030018K13Rik 14.4kb Anti-sense Chr. 5 - 64477008 - 64483967 on NCBI Build GRCm38

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC0500002468.mm.1

          Set Membership:

          Amplicon length less than or equal to 70 3' Most Mus musculus 1700 Probe spans exons

          MicroArray Probes:

          Agilent Affymetrix ABI

          Array Name Probe IDs Targets Target Type
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 341510 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 279426 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 881798 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 15502 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P05594828 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P04181096 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P04166016 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P04808812 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23751275 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23751233 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23751278 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23751244 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23751245 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23751255 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23751237 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23751267 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23751240 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23751248 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23751241 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23751251 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23751274 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23751268 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23751236 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23751234 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23751250 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23751272 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23751258 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P04350263 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P04745539 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23751254 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23751247 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23751239 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23751243 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23751257 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23751242 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P04745541 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23751230 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P04745543 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23751262 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Clariom S Array PSR0500022010.mm.1 TC0500002468.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0500022010.mm.1 NR_040681.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0500022010.mm.1 NR_040681.1 Refseq
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 699705 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-74594944d-pk6zc:80/100.66.130.62:80.
          git-commit: ab2b2261c2971e97b1ae408e3ad10520b8413932
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.31.0-2025.06.23-1.0