Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm01218657_g1
          See other 4930405L22RIK GE Assays ›
          Gene Symbol:
          4930405L22Rik
          Gene Name:
          RIKEN cDNA 4930405L22 gene
          Gene Aliases:
          -
          Chromosome Location:
          Chr.5: 45926822 - 45932409 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm01218657_g1
          Size
          Availability Made To Order
          Catalog # 4426961
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: 4930405L22Rik
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NR_110478.1 2 - 3 199 108
          GenBank mRNA AK019567.1 - 2 - 3 199 108
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Transcript Location Amplicon Length
          GenBankmRNA BY714632.1 - 2 - 3 199 Chr. 5 - 45926821 - 45932409 [-] on Build GRCm38 108

          Adjacent Gene Details

          Gene name Gene Symbol Alias Distance Relative Direction Location Chromsome
          Upstream ligand dependent nuclear receptor corepressor-like Lcorl Mlr1 -66.7kb Sense Chr. 5 - 45697184 - 45860115 on NCBI Build GRCm38
          Downstream slit homolog 2 (Drosophila) Slit2 Drad-1,
          E030015M03Rik,
          E130320P19Rik,
          Slil3,
          mKIAA4141,
          slit-2
          2.1mb Anti-sense Chr. 5 - 47983108 - 48306282 on NCBI Build GRCm38

          Back To Top

          More Information


          Set Membership:

          Amplicon length greater than or equal to 101 3' Most Detect Genomic DNA Mus musculus Probe spans exons

          MicroArray Probes:

          Agilent

          Array Name Probe IDs Targets Target Type
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 205259 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P157051 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P157051 - -
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P133779 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P157051 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23659182 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P04261608 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23659163 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23659179 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23659072 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23659168 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23659177 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P04703653 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23659170 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23659071 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23659078 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P04604588 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23659076 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23659162 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23659069 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23659167 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23659180 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23659176 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23659080 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P04703648 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23659175 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23659173 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23659067 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23659077 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23659169 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23659074 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-74594944d-pk6zc:80/100.66.130.62:80.
          git-commit: ab2b2261c2971e97b1ae408e3ad10520b8413932
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.31.0-2025.06.23-1.0