Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm01219080_g1
          See other 1810053B23RIK GE Assays ›
          Gene Symbol:
          1810053B23Rik
          Gene Name:
          RIKEN cDNA 1810053B23 gene
          Gene Aliases:
          1810008L18Rik, 1810056K14Rik
          Chromosome Location:
          Chr.16: 93343716 - 93359543 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm01219080_g1
          Size
          Availability Made To Order
          Catalog # 4426961
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: 1810053B23Rik
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NR_040486.1 3 - 4 656 71
          NR_040488.1 3 - 4 498 71
          NR_040489.1 3 - 4 496 71
          GenBank mRNA AI390631.1 - 3 - 4 488 71
          AK007854.1 - 3 - 4 657 71
          AK160158.1 - 3 - 4 499 71
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Transcript Location Amplicon Length
          GenBankmRNA BY707857.1 - 3 - 4 657 Chr. 16 - 93344020 - 93353189 [-] on Build GRCm38 71

          Adjacent Gene Details

          Gene name Gene Symbol Alias Distance Relative Direction Location Chromsome
          Upstream runt related transcription factor 1 Runx1 AML1,
          CBF-alpha-2,
          Cbfa2,
          Pebp2a2,
          Pebpa2b
          -517.7kb Sense Chr. 16 - 92601466 - 92826074 on NCBI Build GRCm38
          Downstream microRNA 802 Mir802 Mirn802,
          mir-802,
          mmu-mir-802
          24.7kb Anti-sense Chr. 16 - 93369720 - 93369816 on NCBI Build GRCm38

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC1600002103.mm.1

          Set Membership:

          Amplicon length between 71 and 85 3' Most Detect Genomic DNA Mus musculus 1700 Probe spans exons

          MicroArray Probes:

          Agilent Affymetrix ABI

          Array Name Probe IDs Targets Target Type
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 154785 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 562641 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 183446 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 55719 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 188894 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 746033 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 650119 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 84994 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2002772 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P08687880 - -
          Whole Mouse Genome G4122A A_52_P236288 AK007854.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P236288 AK160158.1 mouse_transcriptome
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_67_P08687880 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30975176 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30975197 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30975171 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P13054090 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P12646938 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P12595818 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P12593213 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30975135 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30975216 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30975210 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P12593214 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30975193 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30975251 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P12578156 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30975203 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30975214 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30975169 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P12523177 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30975143 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30975139 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30975189 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30975172 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30975182 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P12905525 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30975166 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30975252 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P12952512 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P12578158 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30975188 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30975198 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30975202 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P12860868 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30975167 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P12908687 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30975246 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30975181 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30975136 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30975141 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P12571246 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P12578161 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30975145 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30975132 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30975142 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30975098 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30975205 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P12571264 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30975196 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30975131 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30975184 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30975158 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30975200 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30975207 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30975178 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P12571261 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P12952515 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30975101 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30975161 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30975194 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30975137 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P12955313 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix HT MG-430 PM Array Plate 1431742_PM_at - -
          Mouse Genome 430 2.0 Array 1431742_at - -
          Mouse Genome 430 2.0 Array 1431742_PM_at - -
          Affymetrix Mouse Clariom S Array PSR1600016148.mm.1 TC1600002103.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1600016148.mm.1 NR_040486.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1600016148.mm.1 NR_040486.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1600016148.mm.1 NR_040488.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1600016148.mm.1 NR_040488.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1600016148.mm.1 NR_040489.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1600016148.mm.1 NR_040489.1 Refseq
          Mouse Expression Set 430 1431742_at - -
          Mouse Expression Set 430 1431742_PM_at - -
          Affymetrix HT Mouse Genome 430B Array 1431742_PM_at - -
          Affymetrix HT Mouse Genome 430B Array 1431742_at - -
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 361766 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-74594944d-pk6zc:80/100.66.130.62:80.
          git-commit: ab2b2261c2971e97b1ae408e3ad10520b8413932
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.31.0-2025.06.23-1.0