Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm01219097_m1
          See other A630023A22RIK GE Assays ›
          Gene Symbol:
          A630023A22Rik
          Gene Name:
          RIKEN cDNA A630023A22 gene
          Gene Aliases:
          AI461879
          Chromosome Location:
          Chr.14: 34051126 - 34102754 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm01219097_m1
          Size
          Availability Made To Order
          Catalog # 4426961
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: A630023A22Rik
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001251843.1 NP_001238772.1 3 - 4 579 67
          GenBank mRNA AK041587.1 - 3 - 4 518 67
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Transcript Location Amplicon Length
          GenBankmRNA BB638275.1 - 3 - 4 Chr. 14 - 34052633 - 34102693 [-] on Build GRCm38 67

          Adjacent Gene Details

          Gene name Gene Symbol Alias Distance Relative Direction Location Chromsome
          Upstream anthrax toxin receptor-like Antxrl 1700112N15Rik,
          4933430J11
          Anti-sense Chr. 14 - 34052713 - 34076323 on NCBI Build GRCm38
          Downstream annexin A8 Anxa8 AI466995,
          Anx8
          32.2kb Anti-sense Chr. 14 - 34085979 - 34100571 on NCBI Build GRCm38

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC1400001718.mm.1

          Set Membership:

          Amplicon length less than or equal to 70 3' Most Mus musculus 1700 Probe spans exons

          Gene Ontology Categories:

          Function(s) Process(es)

          biological_process
          molecular_function

          MicroArray Probes:

          Agilent Affymetrix ABI

          Array Name Probe IDs Targets Target Type
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2274370 AI461879.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2274370 AK041587.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2274370 BB229670.2 mouse_transcriptome
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 663042 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 209442 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 340840 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 813058 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 593968 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 498718 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 720062 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 39272 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 240869 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 594521 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P154593 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2274370 AI461879.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2274370 AK041587.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2274370 BB229670.2 mouse_transcriptome
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P08079579 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P154593 - -
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P100914 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P154593 - -
          Agilent Mouse cDNA Microarray (G4104A) 1248058 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P11207887 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29528099 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29528102 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29528101 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P11307599 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29528103 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix HT MG-430 PM Array Plate 1455536_PM_at - -
          Mouse Genome 430 2.0 Array 1455536_at - -
          Mouse Genome 430 2.0 Array 1455536_PM_at - -
          Affymetrix Mouse Clariom S Array PSR1400013380.mm.1 TC1400001718.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1400013380.mm.1 NM_001251843.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1400013380.mm.1 NM_001251843.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1400013381.mm.1 TC1400001718.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1400013381.mm.1 NM_001251843.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1400013381.mm.1 NM_001251843.1 Refseq
          Mouse Expression Set 430 1455536_at - -
          Mouse Expression Set 430 1455536_PM_at - -
          Affymetrix HT Mouse Genome 430B Array 1455536_at - -
          Affymetrix HT Mouse Genome 430B Array 1455536_PM_at - -
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 748528 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-65cb46898d-nlgdw:80/100.66.128.131:80.
          git-commit: e5493f66f4597fc66e516c8a379ca3b6bed48f5b
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.33.0-2025.07.30-1.0